Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49005607-49006049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKAG1
Entrez Gene ID
5571
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 1
Gene Aliases
AMPKG
UniGene
Hs.530862
Gene Chromosome Location
Chr.12: 49002272-49018975 on Build GRCh38
Transcripts
NM_001206709.1 NM_001206710.1 NM_002733.4 XM_005269019.4 XM_005269020.2 XM_006719499.2 XM_011538562.2 XM_017019622.1 XM_017019623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1263045 c.167A>T p.Q56L Chr.12: 49005744-49005744 on Build GRCh38
5942493 c.92C>T p.S31F Chr.12: 49005819-49005819 on Build GRCh38
1222012 c.64C>A p.P22T Chr.12: 49005847-49005847 on Build GRCh38
939953 c.118G>C p.D40H Chr.12: 49005793-49005793 on Build GRCh38
2006537 c.82G>A p.V28M Chr.12: 49005829-49005829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027197732 Chr.12: 49005674-49005674 on Build GRCh38
rs34905927 Chr.12: 49005845-49005846 on Build GRCh38
rs754094570 Chr.12: 49005896-49005896 on Build GRCh38
rs758972457 Chr.12: 49005853-49005854 on Build GRCh38
rs759401511 Chr.12: 49005723-49005723 on Build GRCh38
rs771144897 Chr.12: 49005781-49005781 on Build GRCh38
rs887515928 Chr.12: 49005684-49005684 on Build GRCh38
rs201861851 Chr.12: 49005808-49005808 on Build GRCh38
rs777059418 Chr.12: 49005815-49005815 on Build GRCh38
rs200066830 Chr.12: 49005830-49005830 on Build GRCh38
rs748723454 Chr.12: 49005819-49005819 on Build GRCh38
rs913648904 Chr.12: 49006004-49006004 on Build GRCh38
rs753756784 Chr.12: 49005862-49005863 on Build GRCh38
rs756484181 Chr.12: 49005634-49005634 on Build GRCh38
rs925147824 Chr.12: 49005976-49005976 on Build GRCh38
rs558948513 Chr.12: 49005953-49005953 on Build GRCh38
rs754101147 Chr.12: 49005802-49005802 on Build GRCh38
rs752600036 Chr.12: 49005855-49005856 on Build GRCh38
rs767450614 Chr.12: 49005854-49005855 on Build GRCh38
rs749769122 Chr.12: 49005697-49005697 on Build GRCh38
rs1004037978 Chr.12: 49005862-49005862 on Build GRCh38
rs200836335 Chr.12: 49005823-49005823 on Build GRCh38
rs201241007 Chr.12: 49005835-49005835 on Build GRCh38
rs576756433 Chr.12: 49005752-49005752 on Build GRCh38
rs765123296 Chr.12: 49005735-49005735 on Build GRCh38
rs747621596 Chr.12: 49005713-49005713 on Build GRCh38
rs755400968 Chr.12: 49005805-49005805 on Build GRCh38
rs912536310 Chr.12: 49005992-49005992 on Build GRCh38
rs979231165 Chr.12: 49005974-49005974 on Build GRCh38
rs760744571 Chr.12: 49005858-49005859 on Build GRCh38
rs773852651 Chr.12: 49005796-49005798 on Build GRCh38
rs767637720 Chr.12: 49005754-49005754 on Build GRCh38
rs772986682 Chr.12: 49005716-49005716 on Build GRCh38
rs771722251 Chr.12: 49005715-49005715 on Build GRCh38
rs780347186 Chr.12: 49005641-49005641 on Build GRCh38
rs200706669 Chr.12: 49005776-49005776 on Build GRCh38
rs367864375 Chr.12: 49005858-49005858 on Build GRCh38
rs755485935 Chr.12: 49005703-49005703 on Build GRCh38
rs776381191 Chr.12: 49005833-49005833 on Build GRCh38
rs779495352 Chr.12: 49005818-49005818 on Build GRCh38
rs772105380 Chr.12: 49005869-49005869 on Build GRCh38
rs763001186 Chr.12: 49005719-49005719 on Build GRCh38
rs199910078 Chr.12: 49005810-49005810 on Build GRCh38
rs890700504 Chr.12: 49005655-49005655 on Build GRCh38
rs201626731 Chr.12: 49005859-49005859 on Build GRCh38
rs756328911 Chr.12: 49005784-49005784 on Build GRCh38
rs759966006 Chr.12: 49005904-49005904 on Build GRCh38
rs201797468 Chr.12: 49005881-49005881 on Build GRCh38
rs762941105 Chr.12: 49005868-49005868 on Build GRCh38
rs773211996 Chr.12: 49005879-49005879 on Build GRCh38
rs770730517 Chr.12: 49005720-49005720 on Build GRCh38
rs980701183 Chr.12: 49005909-49005909 on Build GRCh38
rs200952727 Chr.12: 49005886-49005886 on Build GRCh38
rs779585113 Chr.12: 49005707-49005707 on Build GRCh38
rs775414575 Chr.12: 49005743-49005743 on Build GRCh38
rs776341939 Chr.12: 49005721-49005721 on Build GRCh38
rs763951666 Chr.12: 49005860-49005861 on Build GRCh38
rs541535909 Chr.12: 49006006-49006006 on Build GRCh38
rs957734934 Chr.12: 49005977-49005977 on Build GRCh38
rs750653558 Chr.12: 49005629-49005629 on Build GRCh38
rs201786139 Chr.12: 49005829-49005829 on Build GRCh38
rs766741379 Chr.12: 49005795-49005795 on Build GRCh38
rs559919544 Chr.12: 49005681-49005681 on Build GRCh38
rs184621052 Chr.12: 49005980-49005980 on Build GRCh38
rs1001982003 Chr.12: 49005691-49005691 on Build GRCh38
rs1051920816 Chr.12: 49005652-49005652 on Build GRCh38

View Full Product Details