Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16013783-16014304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPB7
Entrez Gene ID
27129
Gene Name
heat shock protein family B (small) member 7
Gene Aliases
cvHSP
UniGene
Hs.502612
Gene Chromosome Location
Chr.1: 16014028-16018796 on Build GRCh38
Transcripts
NM_014424.4 XM_011541249.1 XM_011541250.2

SNP Detail

SNP ID Chromosome Location
rs763098635 Chr.1: 16014180-16014180 on Build GRCh38
rs15399 Chr.1: 16014109-16014109 on Build GRCh38
rs748243096 Chr.1: 16013850-16013850 on Build GRCh38
rs888221716 Chr.1: 16013922-16013922 on Build GRCh38
rs1048334 Chr.1: 16014266-16014266 on Build GRCh38
rs375698047 Chr.1: 16014221-16014221 on Build GRCh38
rs947718158 Chr.1: 16014092-16014092 on Build GRCh38
rs1048514 Chr.1: 16014117-16014117 on Build GRCh38
rs992752371 Chr.1: 16014151-16014151 on Build GRCh38
rs1006770485 Chr.1: 16014004-16014004 on Build GRCh38
rs904491226 Chr.1: 16014241-16014241 on Build GRCh38
rs147287304 Chr.1: 16014193-16014193 on Build GRCh38
rs980351580 Chr.1: 16014093-16014093 on Build GRCh38
rs186865506 Chr.1: 16014274-16014274 on Build GRCh38
rs1005200376 Chr.1: 16013950-16013952 on Build GRCh38
rs891863689 Chr.1: 16013873-16013873 on Build GRCh38
rs561067026 Chr.1: 16013849-16013849 on Build GRCh38
rs929733008 Chr.1: 16013813-16013813 on Build GRCh38
rs1044270957 Chr.1: 16014202-16014205 on Build GRCh38
rs938865645 Chr.1: 16013858-16013858 on Build GRCh38
rs946150564 Chr.1: 16014222-16014222 on Build GRCh38
rs985621987 Chr.1: 16014060-16014060 on Build GRCh38
rs115358604 Chr.1: 16014085-16014085 on Build GRCh38
rs528255912 Chr.1: 16013882-16013882 on Build GRCh38
rs879818126 Chr.1: 16014105-16014105 on Build GRCh38
rs1018213902 Chr.1: 16014017-16014017 on Build GRCh38
rs778165520 Chr.1: 16013908-16013908 on Build GRCh38
rs112782458 Chr.1: 16014251-16014251 on Build GRCh38
rs924497212 Chr.1: 16013819-16013819 on Build GRCh38
rs111926460 Chr.1: 16014027-16014027 on Build GRCh38
rs998451074 Chr.1: 16014002-16014002 on Build GRCh38
rs909571652 Chr.1: 16013842-16013842 on Build GRCh38
rs372561157 Chr.1: 16013876-16013876 on Build GRCh38
rs951153300 Chr.1: 16014084-16014084 on Build GRCh38
rs1763601 Chr.1: 16013965-16013965 on Build GRCh38
rs182944962 Chr.1: 16014155-16014155 on Build GRCh38
rs1048043853 Chr.1: 16013836-16013836 on Build GRCh38
rs965181559 Chr.1: 16014023-16014023 on Build GRCh38
rs1057119582 Chr.1: 16013865-16013865 on Build GRCh38
rs140738233 Chr.1: 16013811-16013811 on Build GRCh38
rs927567497 Chr.1: 16014103-16014103 on Build GRCh38
rs1044158103 Chr.1: 16013895-16013895 on Build GRCh38
rs562588627 Chr.1: 16013837-16013837 on Build GRCh38
rs758637015 Chr.1: 16013851-16013851 on Build GRCh38
rs11552854 Chr.1: 16014083-16014083 on Build GRCh38
rs11556 Chr.1: 16014170-16014170 on Build GRCh38
rs113839411 Chr.1: 16014028-16014028 on Build GRCh38
rs954228876 Chr.1: 16014059-16014059 on Build GRCh38
rs755410633 Chr.1: 16013991-16013991 on Build GRCh38
rs1043539289 Chr.1: 16014225-16014225 on Build GRCh38
rs138425527 Chr.1: 16014067-16014067 on Build GRCh38
rs565232277 Chr.1: 16014243-16014243 on Build GRCh38
rs897366283 Chr.1: 16013995-16013995 on Build GRCh38
rs891999116 Chr.1: 16014198-16014198 on Build GRCh38
rs866355344 Chr.1: 16013883-16013883 on Build GRCh38
rs142764481 Chr.1: 16014087-16014087 on Build GRCh38
rs370053748 Chr.1: 16013854-16013854 on Build GRCh38
rs938960597 Chr.1: 16014106-16014106 on Build GRCh38
rs150110485 Chr.1: 16013889-16013889 on Build GRCh38

View Full Product Details