Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89263553-89263922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP5
Entrez Gene ID
115362
Gene Name
guanylate binding protein 5
Gene Aliases
GBP-5
UniGene
Hs.513726
Gene Chromosome Location
Chr.1: 89258950-89272861 on Build GRCh38
Transcripts
NM_001134486.2 NM_052942.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1503960 c.1205C>T p.S402L Chr.1: 89263893-89263893 on Build GRCh38
5519963 c.1362G>A p.Q454Q Chr.1: 89263736-89263736 on Build GRCh38
1644630 c.1340G>A p.R447Q Chr.1: 89263758-89263758 on Build GRCh38
1344765 c.1218G>A p.S406S Chr.1: 89263880-89263880 on Build GRCh38
913013 c.1217C>T p.S406L Chr.1: 89263881-89263881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150292659 Chr.1: 89263695-89263695 on Build GRCh38
rs762956117 Chr.1: 89263654-89263654 on Build GRCh38
rs575024461 Chr.1: 89263782-89263783 on Build GRCh38
rs140630714 Chr.1: 89263579-89263579 on Build GRCh38
rs773275071 Chr.1: 89263644-89263644 on Build GRCh38
rs762472979 Chr.1: 89263874-89263874 on Build GRCh38
rs761247788 Chr.1: 89263859-89263859 on Build GRCh38
rs896872337 Chr.1: 89263728-89263728 on Build GRCh38
rs200944849 Chr.1: 89263774-89263774 on Build GRCh38
rs1001781034 Chr.1: 89263761-89263761 on Build GRCh38
rs561595452 Chr.1: 89263730-89263730 on Build GRCh38
rs772380698 Chr.1: 89263694-89263694 on Build GRCh38
rs143662900 Chr.1: 89263756-89263756 on Build GRCh38
rs565649317 Chr.1: 89263602-89263602 on Build GRCh38
rs755985311 Chr.1: 89263895-89263895 on Build GRCh38
rs757073705 Chr.1: 89263753-89263753 on Build GRCh38
rs756146590 Chr.1: 89263758-89263758 on Build GRCh38
rs556565480 Chr.1: 89263884-89263884 on Build GRCh38
rs371196248 Chr.1: 89263780-89263780 on Build GRCh38
rs138753771 Chr.1: 89263806-89263806 on Build GRCh38
rs770026145 Chr.1: 89263594-89263594 on Build GRCh38
rs770370985 Chr.1: 89263835-89263835 on Build GRCh38
rs766762535 Chr.1: 89263709-89263709 on Build GRCh38
rs752318061 Chr.1: 89263746-89263746 on Build GRCh38
rs148078713 Chr.1: 89263795-89263795 on Build GRCh38
rs116350128 Chr.1: 89263700-89263700 on Build GRCh38
rs757955006 Chr.1: 89263747-89263747 on Build GRCh38
rs868778489 Chr.1: 89263863-89263863 on Build GRCh38
rs750275362 Chr.1: 89263893-89263893 on Build GRCh38
rs1034629134 Chr.1: 89263771-89263771 on Build GRCh38
rs763884452 Chr.1: 89263749-89263749 on Build GRCh38
rs781151989 Chr.1: 89263754-89263754 on Build GRCh38
rs993943901 Chr.1: 89263729-89263729 on Build GRCh38
rs547270035 Chr.1: 89263595-89263595 on Build GRCh38
rs777055940 Chr.1: 89263716-89263716 on Build GRCh38
rs572642839 Chr.1: 89263603-89263603 on Build GRCh38
rs553250350 Chr.1: 89263892-89263892 on Build GRCh38
rs141794180 Chr.1: 89263821-89263821 on Build GRCh38
rs374220083 Chr.1: 89263823-89263823 on Build GRCh38
rs766536843 Chr.1: 89263658-89263658 on Build GRCh38
rs763724428 Chr.1: 89263880-89263880 on Build GRCh38
rs998692412 Chr.1: 89263726-89263726 on Build GRCh38
rs751275007 Chr.1: 89263750-89263750 on Build GRCh38
rs529905548 Chr.1: 89263632-89263632 on Build GRCh38
rs961286105 Chr.1: 89263822-89263822 on Build GRCh38
rs543453504 Chr.1: 89263793-89263793 on Build GRCh38
rs759954294 Chr.1: 89263829-89263829 on Build GRCh38
rs774838379 Chr.1: 89263867-89263867 on Build GRCh38
rs761620026 Chr.1: 89263888-89263888 on Build GRCh38
rs186974399 Chr.1: 89263881-89263881 on Build GRCh38
rs927032590 Chr.1: 89263586-89263586 on Build GRCh38
rs760157864 Chr.1: 89263735-89263735 on Build GRCh38
rs777932690 Chr.1: 89263799-89263799 on Build GRCh38
rs764639414 Chr.1: 89263740-89263740 on Build GRCh38
rs778829194 Chr.1: 89263759-89263759 on Build GRCh38

View Full Product Details