Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110362645-110363179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM15
Entrez Gene ID
64783
Gene Name
RNA binding motif protein 15
Gene Aliases
OTT, OTT1, SPEN
UniGene
Hs.435947
Gene Chromosome Location
Chr.1: 110338426-110373005 on Build GRCh38
Transcripts
NM_001201546.1 NM_001201547.1 NM_001201548.1 NM_001201549.1 NM_001319220.1 NM_004696.2 XM_005271317.4 XM_006711033.3

Gene Symbol
SLC16A4
Entrez Gene ID
9122
Gene Name
solute carrier family 16 member 4
Gene Aliases
MCT4, MCT5
UniGene
Hs.351306
Gene Chromosome Location
Chr.1: 110362851-110391970 on Build GRCh38
Transcripts
NM_001201546.1 NM_001201547.1 NM_001201548.1 NM_001201549.1 NM_001319220.1 NM_004696.2 XM_005271317.4 XM_006711033.3

SNP Detail

SNP ID Chromosome Location
rs969240758 Chr.1: 110363133-110363133 on Build GRCh38
rs1046002 Chr.1: 110362953-110362953 on Build GRCh38
rs563341656 Chr.1: 110363020-110363020 on Build GRCh38
rs1043241425 Chr.1: 110363141-110363141 on Build GRCh38
rs530932273 Chr.1: 110362800-110362800 on Build GRCh38
rs984662304 Chr.1: 110363028-110363028 on Build GRCh38
rs998617975 Chr.1: 110362945-110362945 on Build GRCh38
rs908714886 Chr.1: 110363030-110363030 on Build GRCh38
rs998982137 Chr.1: 110363154-110363154 on Build GRCh38
rs1055632484 Chr.1: 110363116-110363116 on Build GRCh38
rs564408231 Chr.1: 110362838-110362838 on Build GRCh38
rs933025955 Chr.1: 110362927-110362930 on Build GRCh38
rs1021365817 Chr.1: 110362887-110362887 on Build GRCh38
rs565669121 Chr.1: 110363120-110363120 on Build GRCh38
rs931497570 Chr.1: 110362747-110362747 on Build GRCh38
rs1046344449 Chr.1: 110362767-110362767 on Build GRCh38
rs1019266431 Chr.1: 110362952-110362952 on Build GRCh38
rs903034285 Chr.1: 110363150-110363150 on Build GRCh38
rs764800093 Chr.1: 110362748-110362748 on Build GRCh38
rs114264244 Chr.1: 110362702-110362702 on Build GRCh38
rs992464723 Chr.1: 110362709-110362709 on Build GRCh38
rs938035333 Chr.1: 110363107-110363107 on Build GRCh38
rs532942779 Chr.1: 110362963-110362963 on Build GRCh38
rs1051345982 Chr.1: 110362929-110362929 on Build GRCh38
rs529090663 Chr.1: 110362944-110362944 on Build GRCh38
rs116156830 Chr.1: 110363045-110363045 on Build GRCh38
rs906564309 Chr.1: 110362789-110362789 on Build GRCh38
rs902896116 Chr.1: 110362890-110362890 on Build GRCh38
rs982177080 Chr.1: 110363052-110363052 on Build GRCh38
rs1011860930 Chr.1: 110362869-110362869 on Build GRCh38
rs947331453 Chr.1: 110363131-110363131 on Build GRCh38
rs117813270 Chr.1: 110362814-110362814 on Build GRCh38
rs540536780 Chr.1: 110362764-110362764 on Build GRCh38
rs887103207 Chr.1: 110363094-110363094 on Build GRCh38
rs184996136 Chr.1: 110362836-110362836 on Build GRCh38
rs550097633 Chr.1: 110362997-110362997 on Build GRCh38
rs778806011 Chr.1: 110362751-110362751 on Build GRCh38
rs975643969 Chr.1: 110362974-110362974 on Build GRCh38
rs534616410 Chr.1: 110363121-110363121 on Build GRCh38
rs913545324 Chr.1: 110362888-110362888 on Build GRCh38
rs528342530 Chr.1: 110362754-110362754 on Build GRCh38
rs115866365 Chr.1: 110362995-110362995 on Build GRCh38
rs995981686 Chr.1: 110363060-110363060 on Build GRCh38
rs11120 Chr.1: 110363061-110363061 on Build GRCh38
rs757091268 Chr.1: 110362734-110362734 on Build GRCh38

View Full Product Details