Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155728904-155729441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAP3
Entrez Gene ID
7818
Gene Name
death associated protein 3
Gene Aliases
DAP-3, MRP-S29, MRPS29, bMRP-10
UniGene
Hs.516746
Gene Chromosome Location
Chr.1: 155688104-155739010 on Build GRCh38
Transcripts
NM_001199849.1 NM_001199850.1 NM_001199851.1 NM_004632.3 NM_033657.2 XM_005245480.2 XM_005245481.2 XM_005245482.4 XM_011509972.2 XM_017002289.1 XM_017002290.1 XM_017002291.1 XM_017002292.1 XM_017002293.1 XM_017002294.1 XM_017002295.1 XM_017002296.1 XM_017002297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7122217 c.635_638delAGAG p.R213fs*17 Chr.1: 155729073-155729076 on Build GRCh38
3728009 c.638G>C p.R213T Chr.1: 155729076-155729076 on Build GRCh38
2210927 c.616G>C p.E206Q Chr.1: 155729054-155729054 on Build GRCh38
1202989 c.755C>A p.S252Y Chr.1: 155729278-155729278 on Build GRCh38
6450896 c.843G>A p.P281P Chr.1: 155729366-155729366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779341692 Chr.1: 155729300-155729300 on Build GRCh38
rs866962755 Chr.1: 155729201-155729201 on Build GRCh38
rs761731075 Chr.1: 155729158-155729160 on Build GRCh38
rs757669207 Chr.1: 155729215-155729215 on Build GRCh38
rs867839385 Chr.1: 155729227-155729227 on Build GRCh38
rs765094969 Chr.1: 155729175-155729175 on Build GRCh38
rs754890180 Chr.1: 155729325-155729325 on Build GRCh38
rs372817713 Chr.1: 155729170-155729170 on Build GRCh38
rs761333313 Chr.1: 155729259-155729259 on Build GRCh38
rs750472379 Chr.1: 155729311-155729311 on Build GRCh38
rs771596425 Chr.1: 155729130-155729130 on Build GRCh38
rs774925517 Chr.1: 155729134-155729134 on Build GRCh38
rs777044295 Chr.1: 155729010-155729010 on Build GRCh38
rs764225657 Chr.1: 155729293-155729293 on Build GRCh38
rs758210669 Chr.1: 155729155-155729155 on Build GRCh38
rs760795857 Chr.1: 155729073-155729073 on Build GRCh38
rs367639956 Chr.1: 155729376-155729376 on Build GRCh38
rs1034113965 Chr.1: 155728979-155728979 on Build GRCh38
rs762691201 Chr.1: 155729241-155729241 on Build GRCh38
rs59322867 Chr.1: 155729015-155729015 on Build GRCh38
rs375689451 Chr.1: 155729365-155729365 on Build GRCh38
rs771173696 Chr.1: 155729124-155729124 on Build GRCh38
rs759923685 Chr.1: 155729127-155729127 on Build GRCh38
rs992573179 Chr.1: 155728993-155728993 on Build GRCh38
rs545683945 Chr.1: 155729061-155729061 on Build GRCh38
rs755127068 Chr.1: 155729086-155729086 on Build GRCh38
rs547638092 Chr.1: 155728988-155728988 on Build GRCh38
rs748340933 Chr.1: 155729014-155729014 on Build GRCh38
rs1001669542 Chr.1: 155728949-155728949 on Build GRCh38
rs200984755 Chr.1: 155729217-155729217 on Build GRCh38
rs552936757 Chr.1: 155729214-155729214 on Build GRCh38
rs375216873 Chr.1: 155729154-155729154 on Build GRCh38
rs768025714 Chr.1: 155729006-155729006 on Build GRCh38
rs753566559 Chr.1: 155729062-155729062 on Build GRCh38
rs766481911 Chr.1: 155729272-155729272 on Build GRCh38
rs145328544 Chr.1: 155729218-155729218 on Build GRCh38
rs983130426 Chr.1: 155729378-155729378 on Build GRCh38
rs779538302 Chr.1: 155729346-155729347 on Build GRCh38
rs749717538 Chr.1: 155729115-155729115 on Build GRCh38
rs368325239 Chr.1: 155729152-155729152 on Build GRCh38
rs377654022 Chr.1: 155729262-155729262 on Build GRCh38
rs1025761729 Chr.1: 155728997-155728997 on Build GRCh38
rs1025391605 Chr.1: 155729287-155729287 on Build GRCh38
rs776260956 Chr.1: 155729005-155729006 on Build GRCh38
rs752381978 Chr.1: 155729057-155729057 on Build GRCh38
rs747840157 Chr.1: 155729343-155729343 on Build GRCh38
rs868612639 Chr.1: 155729229-155729229 on Build GRCh38
rs144563957 Chr.1: 155729187-155729187 on Build GRCh38
rs145213844 Chr.1: 155729367-155729367 on Build GRCh38
rs781370831 Chr.1: 155729088-155729088 on Build GRCh38
rs368412712 Chr.1: 155729021-155729021 on Build GRCh38
rs904694853 Chr.1: 155728932-155728932 on Build GRCh38
rs747183061 Chr.1: 155729002-155729002 on Build GRCh38
rs754116737 Chr.1: 155729297-155729297 on Build GRCh38
rs751701842 Chr.1: 155729383-155729384 on Build GRCh38
rs756999098 Chr.1: 155729076-155729076 on Build GRCh38
rs748394462 Chr.1: 155729100-155729100 on Build GRCh38
rs536698370 Chr.1: 155729307-155729307 on Build GRCh38
rs774705445 Chr.1: 155729026-155729026 on Build GRCh38
rs771532009 Chr.1: 155729022-155729022 on Build GRCh38
rs376225469 Chr.1: 155729138-155729138 on Build GRCh38
rs772816024 Chr.1: 155729264-155729264 on Build GRCh38
rs367821120 Chr.1: 155729104-155729104 on Build GRCh38
rs373171130 Chr.1: 155729383-155729383 on Build GRCh38
rs1043335872 Chr.1: 155729186-155729186 on Build GRCh38
rs371226776 Chr.1: 155729266-155729266 on Build GRCh38
rs775974700 Chr.1: 155729228-155729228 on Build GRCh38
rs778968624 Chr.1: 155729079-155729079 on Build GRCh38
rs370718586 Chr.1: 155729203-155729203 on Build GRCh38
rs780589326 Chr.1: 155729320-155729320 on Build GRCh38
rs200267442 Chr.1: 155729126-155729126 on Build GRCh38
rs201155259 Chr.1: 155729195-155729195 on Build GRCh38
rs766903055 Chr.1: 155729056-155729056 on Build GRCh38
rs370880846 Chr.1: 155729414-155729414 on Build GRCh38
rs367572641 Chr.1: 155729156-155729156 on Build GRCh38
rs147699097 Chr.1: 155729239-155729239 on Build GRCh38
rs57692591 Chr.1: 155729241-155729241 on Build GRCh38
rs996473331 Chr.1: 155729204-155729204 on Build GRCh38
rs1040848596 Chr.1: 155729174-155729174 on Build GRCh38
rs919814951 Chr.1: 155729129-155729129 on Build GRCh38
rs931233006 Chr.1: 155729132-155729132 on Build GRCh38
rs959451578 Chr.1: 155729345-155729345 on Build GRCh38
rs4933 Chr.1: 155729327-155729327 on Build GRCh38
rs371263118 Chr.1: 155729059-155729059 on Build GRCh38
rs569405423 Chr.1: 155729306-155729306 on Build GRCh38
rs766241879 Chr.1: 155729370-155729370 on Build GRCh38
rs756299657 Chr.1: 155729284-155729284 on Build GRCh38
rs750376416 Chr.1: 155729185-155729186 on Build GRCh38
rs201265692 Chr.1: 155729149-155729149 on Build GRCh38
rs975191251 Chr.1: 155729072-155729072 on Build GRCh38
rs776387238 Chr.1: 155729242-155729242 on Build GRCh38
rs141810300 Chr.1: 155729396-155729396 on Build GRCh38
rs1802736 Chr.1: 155729282-155729282 on Build GRCh38
rs766232102 Chr.1: 155729341-155729342 on Build GRCh38
rs750306311 Chr.1: 155729083-155729083 on Build GRCh38
rs768606269 Chr.1: 155729006-155729006 on Build GRCh38
rs373985177 Chr.1: 155729220-155729220 on Build GRCh38
rs749435379 Chr.1: 155729213-155729213 on Build GRCh38
rs951124976 Chr.1: 155729298-155729298 on Build GRCh38
rs753795801 Chr.1: 155729399-155729399 on Build GRCh38
rs760328530 Chr.1: 155729136-155729136 on Build GRCh38
rs751517938 Chr.1: 155729283-155729283 on Build GRCh38

View Full Product Details