Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 16371638-16372051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF16B
Entrez Gene ID
55614
Gene Name
kinesin family member 16B
Gene Aliases
C20orf23, KISC20ORF, SNX23
UniGene
Hs.101774
Gene Chromosome Location
Chr.20: 16272104-16573457 on Build GRCh38
Transcripts
NM_001199865.1 NM_001199866.1 NM_024704.4 XM_005260750.3 XM_005260751.4 XM_005260752.3 XM_005260753.3 XM_005260754.3 XM_005260755.3 XM_006723588.3 XM_017027926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5190878 c.3440C>T p.T1147M Chr.20: 16371672-16371672 on Build GRCh38
6092611 c.3424A>T p.S1142C Chr.20: 16371688-16371688 on Build GRCh38
5017576 c.3361delT p.Y1121fs*16 Chr.20: 16371751-16371751 on Build GRCh38
312329 c.3380A>G p.Q1127R Chr.20: 16371732-16371732 on Build GRCh38
312330 c.3380A>G p.Q1127R Chr.20: 16371732-16371732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998581712 Chr.20: 16371967-16371967 on Build GRCh38
rs900740869 Chr.20: 16371887-16371887 on Build GRCh38
rs75989969 Chr.20: 16371699-16371699 on Build GRCh38
rs780448065 Chr.20: 16371748-16371748 on Build GRCh38
rs577518747 Chr.20: 16372009-16372009 on Build GRCh38
rs773248482 Chr.20: 16371783-16371783 on Build GRCh38
rs767967206 Chr.20: 16371727-16371727 on Build GRCh38
rs368053049 Chr.20: 16371689-16371689 on Build GRCh38
rs752147583 Chr.20: 16371736-16371738 on Build GRCh38
rs755303332 Chr.20: 16371872-16371872 on Build GRCh38
rs3746783 Chr.20: 16371903-16371903 on Build GRCh38
rs368915275 Chr.20: 16372005-16372005 on Build GRCh38
rs1022921626 Chr.20: 16372018-16372019 on Build GRCh38
rs749503207 Chr.20: 16371901-16371901 on Build GRCh38
rs887710135 Chr.20: 16371885-16371885 on Build GRCh38
rs367836942 Chr.20: 16371743-16371745 on Build GRCh38
rs776553582 Chr.20: 16371798-16371798 on Build GRCh38
rs924462028 Chr.20: 16371669-16371669 on Build GRCh38
rs747644686 Chr.20: 16371779-16371779 on Build GRCh38
rs766313563 Chr.20: 16371708-16371708 on Build GRCh38
rs8123195 Chr.20: 16371756-16371756 on Build GRCh38
rs1029748519 Chr.20: 16371952-16371952 on Build GRCh38
rs768869303 Chr.20: 16371806-16371806 on Build GRCh38
rs749011870 Chr.20: 16371672-16371672 on Build GRCh38
rs1009029948 Chr.20: 16371837-16371837 on Build GRCh38
rs973263654 Chr.20: 16371935-16371935 on Build GRCh38
rs904299415 Chr.20: 16371943-16371943 on Build GRCh38
rs547335822 Chr.20: 16371693-16371693 on Build GRCh38
rs985490352 Chr.20: 16371991-16371991 on Build GRCh38
rs754590689 Chr.20: 16371772-16371772 on Build GRCh38
rs768031772 Chr.20: 16371674-16371674 on Build GRCh38
rs1031346123 Chr.20: 16371975-16371975 on Build GRCh38
rs138136533 Chr.20: 16371671-16371671 on Build GRCh38
rs1023950537 Chr.20: 16371874-16371874 on Build GRCh38
rs746988228 Chr.20: 16371784-16371784 on Build GRCh38
rs750960072 Chr.20: 16371740-16371740 on Build GRCh38
rs73898703 Chr.20: 16371812-16371812 on Build GRCh38
rs759257065 Chr.20: 16371803-16371803 on Build GRCh38
rs924137344 Chr.20: 16372026-16372026 on Build GRCh38
rs778442592 Chr.20: 16371776-16371776 on Build GRCh38
rs775550223 Chr.20: 16371709-16371709 on Build GRCh38
rs772076054 Chr.20: 16371683-16371683 on Build GRCh38
rs538666717 Chr.20: 16371726-16371726 on Build GRCh38
rs147513419 Chr.20: 16371908-16371908 on Build GRCh38
rs189218104 Chr.20: 16371836-16371836 on Build GRCh38
rs773975721 Chr.20: 16371680-16371680 on Build GRCh38
rs370814736 Chr.20: 16371666-16371666 on Build GRCh38
rs62198125 Chr.20: 16372023-16372023 on Build GRCh38
rs370655324 Chr.20: 16371807-16371807 on Build GRCh38
rs745913611 Chr.20: 16371972-16371972 on Build GRCh38
rs376560120 Chr.20: 16371723-16371723 on Build GRCh38
rs911462794 Chr.20: 16371998-16371998 on Build GRCh38
rs769931372 Chr.20: 16371984-16371984 on Build GRCh38
rs934014625 Chr.20: 16371725-16371725 on Build GRCh38
rs756440126 Chr.20: 16371743-16371743 on Build GRCh38
rs770833387 Chr.20: 16371796-16371796 on Build GRCh38
rs961438940 Chr.20: 16371910-16371910 on Build GRCh38
rs192867504 Chr.20: 16371664-16371664 on Build GRCh38
rs375035068 Chr.20: 16371718-16371718 on Build GRCh38
rs771738893 Chr.20: 16371781-16371781 on Build GRCh38
rs555178031 Chr.20: 16371859-16371859 on Build GRCh38
rs946838247 Chr.20: 16371788-16371788 on Build GRCh38

View Full Product Details