Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 3577593-3578107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS7
Entrez Gene ID
6201
Gene Name
ribosomal protein S7
Gene Aliases
DBA8, S7
UniGene
Hs.534346
Gene Chromosome Location
Chr.2: 3575263-3580919 on Build GRCh38
Transcripts
NM_001011.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
347243 c.326G>T p.R109L Chr.2: 3577744-3577744 on Build GRCh38
1180315 c.293G>T p.R98M Chr.2: 3577711-3577711 on Build GRCh38
2943339 c.339G>T p.K113N Chr.2: 3577757-3577757 on Build GRCh38
4637434 c.346C>T p.R116C Chr.2: 3577764-3577764 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs549269458 Chr.2: 3577917-3577917 on Build GRCh38
rs556354465 Chr.2: 3577990-3577990 on Build GRCh38
rs985083792 Chr.2: 3577914-3577914 on Build GRCh38
rs538065869 Chr.2: 3577957-3577957 on Build GRCh38
rs565581519 Chr.2: 3577883-3577883 on Build GRCh38
rs940539187 Chr.2: 3577654-3577654 on Build GRCh38
rs189350242 Chr.2: 3577900-3577900 on Build GRCh38
rs567481729 Chr.2: 3577921-3577921 on Build GRCh38
rs754079338 Chr.2: 3577811-3577812 on Build GRCh38
rs940634091 Chr.2: 3578015-3578015 on Build GRCh38
rs371056451 Chr.2: 3577734-3577734 on Build GRCh38
rs760034391 Chr.2: 3577790-3577790 on Build GRCh38
rs767948960 Chr.2: 3577795-3577795 on Build GRCh38
rs878858480 Chr.2: 3577723-3577723 on Build GRCh38
rs925623452 Chr.2: 3577945-3577945 on Build GRCh38
rs866345076 Chr.2: 3578078-3578078 on Build GRCh38
rs373405472 Chr.2: 3577702-3577702 on Build GRCh38
rs757471985 Chr.2: 3577816-3577816 on Build GRCh38
rs73910594 Chr.2: 3577671-3577671 on Build GRCh38
rs770440343 Chr.2: 3577774-3577774 on Build GRCh38
rs140632181 Chr.2: 3577656-3577657 on Build GRCh38
rs780246648 Chr.2: 3577860-3577860 on Build GRCh38
rs376999255 Chr.2: 3577683-3577683 on Build GRCh38
rs1046597460 Chr.2: 3577997-3577997 on Build GRCh38
rs886671303 Chr.2: 3578014-3578014 on Build GRCh38
rs765824593 Chr.2: 3577659-3577659 on Build GRCh38
rs757321602 Chr.2: 3577698-3577698 on Build GRCh38
rs779035901 Chr.2: 3577817-3577817 on Build GRCh38
rs1028801485 Chr.2: 3577809-3577809 on Build GRCh38
rs73137100 Chr.2: 3577839-3577839 on Build GRCh38
rs756479043 Chr.2: 3577679-3577679 on Build GRCh38
rs967312345 Chr.2: 3577899-3577899 on Build GRCh38
rs796957262 Chr.2: 3577858-3577859 on Build GRCh38
rs752062898 Chr.2: 3577666-3577666 on Build GRCh38
rs937052493 Chr.2: 3577964-3577964 on Build GRCh38
rs909117610 Chr.2: 3577996-3578000 on Build GRCh38
rs758408723 Chr.2: 3578085-3578085 on Build GRCh38
rs1037635275 Chr.2: 3578029-3578029 on Build GRCh38
rs1001502341 Chr.2: 3577719-3577719 on Build GRCh38
rs963415880 Chr.2: 3577787-3577787 on Build GRCh38
rs773794878 Chr.2: 3577778-3577778 on Build GRCh38
rs578036764 Chr.2: 3578018-3578018 on Build GRCh38
rs991651919 Chr.2: 3577965-3577965 on Build GRCh38
rs150686417 Chr.2: 3577689-3577689 on Build GRCh38
rs1036179828 Chr.2: 3577662-3577662 on Build GRCh38
rs753130074 Chr.2: 3577675-3577675 on Build GRCh38
rs199547240 Chr.2: 3577831-3577834 on Build GRCh38
rs750595791 Chr.2: 3577818-3577818 on Build GRCh38
rs756635684 Chr.2: 3577985-3577985 on Build GRCh38
rs868655899 Chr.2: 3578041-3578041 on Build GRCh38
rs764456684 Chr.2: 3577810-3577812 on Build GRCh38
rs371489826 Chr.2: 3577834-3577837 on Build GRCh38
rs949577458 Chr.2: 3577994-3577994 on Build GRCh38
rs565011438 Chr.2: 3577834-3577834 on Build GRCh38
rs772868042 Chr.2: 3577761-3577761 on Build GRCh38
rs538796604 Chr.2: 3578027-3578027 on Build GRCh38
rs1013799447 Chr.2: 3577658-3577658 on Build GRCh38
rs753220088 Chr.2: 3577799-3577799 on Build GRCh38
rs796846849 Chr.2: 3577653-3577654 on Build GRCh38
rs1016239955 Chr.2: 3577705-3577705 on Build GRCh38
rs767858631 Chr.2: 3577672-3577672 on Build GRCh38
rs754061886 Chr.2: 3577813-3577813 on Build GRCh38
rs549865430 Chr.2: 3577811-3577811 on Build GRCh38
rs981224165 Chr.2: 3577846-3577846 on Build GRCh38
rs113127192 Chr.2: 3577803-3577803 on Build GRCh38
rs61730447 Chr.2: 3577735-3577735 on Build GRCh38
rs140005491 Chr.2: 3577863-3577863 on Build GRCh38
rs776562361 Chr.2: 3577732-3577732 on Build GRCh38
rs746941533 Chr.2: 3577701-3577701 on Build GRCh38
rs762541439 Chr.2: 3577769-3577769 on Build GRCh38
rs769173610 Chr.2: 3577918-3577919 on Build GRCh38
rs778900653 Chr.2: 3577700-3577700 on Build GRCh38

View Full Product Details