Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 53750577-53751134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB3
Entrez Gene ID
51130
Gene Name
ankyrin repeat and SOCS box containing 3
Gene Aliases
ASB-3
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53669980-53787009 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001164165.1

Gene Symbol
GPR75-ASB3
Entrez Gene ID
100302652
Gene Name
GPR75-ASB3 readthrough
Gene Aliases
-
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53670293-53860033 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001164165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1021568 c.292C>A p.L98I Chr.2: 53750846-53750846 on Build GRCh38
1021569 c.279A>C p.K93N Chr.2: 53750859-53750859 on Build GRCh38
6158390 c.379C>A p.Q127K Chr.2: 53750873-53750873 on Build GRCh38
7180214 c.354C>G p.F118L Chr.2: 53750898-53750898 on Build GRCh38
3582227 c.282C>T p.I94I Chr.2: 53750856-53750856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914256871 Chr.2: 53750713-53750713 on Build GRCh38
rs768434575 Chr.2: 53750938-53750939 on Build GRCh38
rs111382659 Chr.2: 53750753-53750753 on Build GRCh38
rs947023211 Chr.2: 53751087-53751087 on Build GRCh38
rs768257899 Chr.2: 53750803-53750803 on Build GRCh38
rs141354765 Chr.2: 53750856-53750856 on Build GRCh38
rs1011933052 Chr.2: 53751027-53751027 on Build GRCh38
rs991121872 Chr.2: 53750683-53750683 on Build GRCh38
rs76764843 Chr.2: 53750737-53750737 on Build GRCh38
rs370386828 Chr.2: 53750771-53750771 on Build GRCh38
rs369339617 Chr.2: 53750808-53750808 on Build GRCh38
rs914516420 Chr.2: 53751060-53751060 on Build GRCh38
rs981325487 Chr.2: 53750637-53750637 on Build GRCh38
rs1056419716 Chr.2: 53751002-53751002 on Build GRCh38
rs768364245 Chr.2: 53750991-53750991 on Build GRCh38
rs750137018 Chr.2: 53750833-53750833 on Build GRCh38
rs199556292 Chr.2: 53750758-53750758 on Build GRCh38
rs923805244 Chr.2: 53750935-53750935 on Build GRCh38
rs754038869 Chr.2: 53750839-53750839 on Build GRCh38
rs759692761 Chr.2: 53750937-53750937 on Build GRCh38
rs878972589 Chr.2: 53751037-53751037 on Build GRCh38
rs951270040 Chr.2: 53750600-53750600 on Build GRCh38
rs781685416 Chr.2: 53750977-53750977 on Build GRCh38
rs141518506 Chr.2: 53750853-53750853 on Build GRCh38
rs746968570 Chr.2: 53750842-53750842 on Build GRCh38
rs760579801 Chr.2: 53750926-53750926 on Build GRCh38
rs771864728 Chr.2: 53750907-53750907 on Build GRCh38
rs200223999 Chr.2: 53750755-53750755 on Build GRCh38
rs1051976549 Chr.2: 53750841-53750841 on Build GRCh38
rs534088626 Chr.2: 53750882-53750882 on Build GRCh38
rs375297031 Chr.2: 53750674-53750674 on Build GRCh38
rs112301241 Chr.2: 53750852-53750852 on Build GRCh38
rs769277048 Chr.2: 53750786-53750786 on Build GRCh38
rs748933905 Chr.2: 53750879-53750879 on Build GRCh38
rs775236787 Chr.2: 53750792-53750792 on Build GRCh38
rs756222866 Chr.2: 53750746-53750746 on Build GRCh38
rs746239682 Chr.2: 53750987-53750987 on Build GRCh38
rs747410152 Chr.2: 53750782-53750782 on Build GRCh38
rs771292908 Chr.2: 53750867-53750867 on Build GRCh38
rs138055351 Chr.2: 53750883-53750883 on Build GRCh38
rs765029653 Chr.2: 53750670-53750670 on Build GRCh38
rs774061607 Chr.2: 53750809-53750809 on Build GRCh38
rs751195509 Chr.2: 53750938-53750938 on Build GRCh38
rs896391762 Chr.2: 53751008-53751008 on Build GRCh38
rs896192463 Chr.2: 53750634-53750634 on Build GRCh38
rs556121614 Chr.2: 53751028-53751028 on Build GRCh38
rs765755013 Chr.2: 53751000-53751000 on Build GRCh38
rs928541088 Chr.2: 53750668-53750668 on Build GRCh38
rs964124410 Chr.2: 53750627-53750627 on Build GRCh38
rs775087671 Chr.2: 53750735-53750737 on Build GRCh38
rs778363197 Chr.2: 53750858-53750858 on Build GRCh38
rs767405027 Chr.2: 53750741-53750741 on Build GRCh38
rs1026802244 Chr.2: 53750897-53750897 on Build GRCh38
rs139014010 Chr.2: 53750824-53750824 on Build GRCh38
rs574218764 Chr.2: 53750958-53750958 on Build GRCh38
rs752081124 Chr.2: 53750960-53750960 on Build GRCh38
rs540542893 Chr.2: 53750800-53750800 on Build GRCh38
rs150341971 Chr.2: 53751041-53751041 on Build GRCh38
rs772637804 Chr.2: 53750768-53750768 on Build GRCh38
rs371519069 Chr.2: 53750855-53750855 on Build GRCh38
rs753702521 Chr.2: 53750931-53750931 on Build GRCh38
rs368579825 Chr.2: 53750837-53750837 on Build GRCh38
rs983709154 Chr.2: 53750779-53750779 on Build GRCh38
rs761420380 Chr.2: 53750886-53750886 on Build GRCh38
rs758532530 Chr.2: 53750953-53750953 on Build GRCh38
rs548972431 Chr.2: 53750605-53750605 on Build GRCh38
rs773171418 Chr.2: 53750912-53750912 on Build GRCh38
rs565186466 Chr.2: 53751088-53751088 on Build GRCh38
rs187097477 Chr.2: 53750685-53750685 on Build GRCh38
rs370379125 Chr.2: 53750760-53750760 on Build GRCh38
rs201763724 Chr.2: 53750866-53750866 on Build GRCh38
rs781455954 Chr.2: 53750870-53750870 on Build GRCh38
rs750299583 Chr.2: 53750744-53750744 on Build GRCh38
rs35388971 Chr.2: 53750898-53750899 on Build GRCh38
rs889327244 Chr.2: 53750846-53750846 on Build GRCh38
rs1045103229 Chr.2: 53751033-53751033 on Build GRCh38
rs766185188 Chr.2: 53750930-53750930 on Build GRCh38
rs191197445 Chr.2: 53751104-53751104 on Build GRCh38
rs761838522 Chr.2: 53750817-53750817 on Build GRCh38
rs529218513 Chr.2: 53750602-53750602 on Build GRCh38
rs115068554 Chr.2: 53750968-53750968 on Build GRCh38
rs760550005 Chr.2: 53750836-53750836 on Build GRCh38
rs115817165 Chr.2: 53751050-53751050 on Build GRCh38

View Full Product Details