Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219147557-219147940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHEJ1
Entrez Gene ID
79840
Gene Name
non-homologous end joining factor 1
Gene Aliases
XLF
UniGene
Hs.225988
Gene Chromosome Location
Chr.2: 219075324-219160865 on Build GRCh38
Transcripts
NM_024782.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7245614 c.448G>A p.V150M Chr.2: 219147738-219147738 on Build GRCh38
6341028 c.409C>A p.R137S Chr.2: 219147777-219147777 on Build GRCh38
4091604 c.461C>T p.A154V Chr.2: 219147725-219147725 on Build GRCh38
6466977 c.529G>T p.D177Y Chr.2: 219147657-219147657 on Build GRCh38
1016569 c.518C>T p.T173M Chr.2: 219147668-219147668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777703611 Chr.2: 219147829-219147829 on Build GRCh38
rs759426248 Chr.2: 219147673-219147673 on Build GRCh38
rs769689300 Chr.2: 219147758-219147758 on Build GRCh38
rs985784651 Chr.2: 219147671-219147671 on Build GRCh38
rs375019866 Chr.2: 219147878-219147878 on Build GRCh38
rs773876034 Chr.2: 219147772-219147772 on Build GRCh38
rs148337310 Chr.2: 219147633-219147633 on Build GRCh38
rs774053051 Chr.2: 219147846-219147846 on Build GRCh38
rs141996352 Chr.2: 219147735-219147735 on Build GRCh38
rs61753340 Chr.2: 219147786-219147786 on Build GRCh38
rs761028730 Chr.2: 219147787-219147787 on Build GRCh38
rs369523341 Chr.2: 219147623-219147623 on Build GRCh38
rs775191124 Chr.2: 219147686-219147686 on Build GRCh38
rs763611486 Chr.2: 219147703-219147703 on Build GRCh38
rs954430930 Chr.2: 219147876-219147876 on Build GRCh38
rs111985333 Chr.2: 219147835-219147835 on Build GRCh38
rs377105168 Chr.2: 219147639-219147639 on Build GRCh38
rs755272050 Chr.2: 219147809-219147809 on Build GRCh38
rs1030120036 Chr.2: 219147869-219147869 on Build GRCh38
rs754215865 Chr.2: 219147807-219147807 on Build GRCh38
rs766636405 Chr.2: 219147726-219147726 on Build GRCh38
rs376861632 Chr.2: 219147736-219147736 on Build GRCh38
rs114020286 Chr.2: 219147581-219147581 on Build GRCh38
rs781232983 Chr.2: 219147740-219147740 on Build GRCh38
rs768115441 Chr.2: 219147718-219147718 on Build GRCh38
rs773823999 Chr.2: 219147777-219147777 on Build GRCh38
rs140633489 Chr.2: 219147699-219147699 on Build GRCh38
rs188077600 Chr.2: 219147820-219147820 on Build GRCh38
rs752122425 Chr.2: 219147626-219147626 on Build GRCh38
rs763590891 Chr.2: 219147622-219147622 on Build GRCh38
rs762342375 Chr.2: 219147715-219147715 on Build GRCh38
rs758387082 Chr.2: 219147738-219147738 on Build GRCh38
rs949893152 Chr.2: 219147886-219147886 on Build GRCh38
rs7585742 Chr.2: 219147854-219147854 on Build GRCh38
rs770861190 Chr.2: 219147668-219147668 on Build GRCh38
rs371400946 Chr.2: 219147677-219147677 on Build GRCh38
rs749513419 Chr.2: 219147648-219147648 on Build GRCh38
rs1045525434 Chr.2: 219147892-219147892 on Build GRCh38
rs74477376 Chr.2: 219147913-219147913 on Build GRCh38
rs751006580 Chr.2: 219147713-219147713 on Build GRCh38
rs755940345 Chr.2: 219147830-219147830 on Build GRCh38
rs762489890 Chr.2: 219147619-219147619 on Build GRCh38
rs766853125 Chr.2: 219147799-219147799 on Build GRCh38
rs780359781 Chr.2: 219147642-219147642 on Build GRCh38
rs768404777 Chr.2: 219147836-219147836 on Build GRCh38
rs752936625 Chr.2: 219147814-219147814 on Build GRCh38
rs765605992 Chr.2: 219147813-219147813 on Build GRCh38
rs759356112 Chr.2: 219147712-219147712 on Build GRCh38
rs757708569 Chr.2: 219147630-219147630 on Build GRCh38
rs148382683 Chr.2: 219147768-219147768 on Build GRCh38
rs374664084 Chr.2: 219147721-219147721 on Build GRCh38
rs756563868 Chr.2: 219147722-219147722 on Build GRCh38
rs370231557 Chr.2: 219147769-219147769 on Build GRCh38
rs376407655 Chr.2: 219147649-219147649 on Build GRCh38
rs746945256 Chr.2: 219147659-219147659 on Build GRCh38
rs778613948 Chr.2: 219147624-219147628 on Build GRCh38
rs146783338 Chr.2: 219147656-219147656 on Build GRCh38
rs576486559 Chr.2: 219147592-219147592 on Build GRCh38
rs754126015 Chr.2: 219147729-219147729 on Build GRCh38
rs375230208 Chr.2: 219147776-219147776 on Build GRCh38
rs202229437 Chr.2: 219147745-219147745 on Build GRCh38
rs762584679 Chr.2: 219147687-219147687 on Build GRCh38
rs775211651 Chr.2: 219147762-219147762 on Build GRCh38
rs902982250 Chr.2: 219147839-219147839 on Build GRCh38
rs779859313 Chr.2: 219147833-219147833 on Build GRCh38
rs1055710780 Chr.2: 219147800-219147800 on Build GRCh38
rs750747940 Chr.2: 219147638-219147638 on Build GRCh38
rs776313502 Chr.2: 219147672-219147672 on Build GRCh38

View Full Product Details