Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81061205-81061803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf46
Entrez Gene ID
123775
Gene Name
chromosome 16 open reading frame 46
Gene Aliases
-
UniGene
Hs.658684
Gene Chromosome Location
Chr.16: 81053497-81077267 on Build GRCh38
Transcripts
NM_001100873.1 NM_152337.2 XM_005255792.2 XM_011522848.2 XM_011522849.2 XM_017022916.1 XM_017022917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4502795 c.622C>T p.L208F Chr.16: 81061727-81061727 on Build GRCh38
4129384 c.863T>C p.I288T Chr.16: 81061486-81061486 on Build GRCh38
3421185 c.690G>T p.K230N Chr.16: 81061659-81061659 on Build GRCh38
974001 c.910C>A p.L304M Chr.16: 81061439-81061439 on Build GRCh38
4406347 c.642G>A p.L214L Chr.16: 81061707-81061707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143084248 Chr.16: 81061339-81061339 on Build GRCh38
rs763450505 Chr.16: 81061424-81061424 on Build GRCh38
rs370784988 Chr.16: 81061520-81061520 on Build GRCh38
rs76579573 Chr.16: 81061419-81061419 on Build GRCh38
rs748536475 Chr.16: 81061254-81061254 on Build GRCh38
rs772149274 Chr.16: 81061769-81061769 on Build GRCh38
rs575038585 Chr.16: 81061712-81061712 on Build GRCh38
rs868170751 Chr.16: 81061532-81061532 on Build GRCh38
rs753955003 Chr.16: 81061311-81061311 on Build GRCh38
rs768061988 Chr.16: 81061570-81061571 on Build GRCh38
rs537563624 Chr.16: 81061308-81061308 on Build GRCh38
rs112523458 Chr.16: 81061614-81061614 on Build GRCh38
rs765765776 Chr.16: 81061343-81061343 on Build GRCh38
rs761447624 Chr.16: 81061400-81061400 on Build GRCh38
rs147533255 Chr.16: 81061397-81061397 on Build GRCh38
rs9930623 Chr.16: 81061280-81061280 on Build GRCh38
rs2287995 Chr.16: 81061494-81061494 on Build GRCh38
rs752747522 Chr.16: 81061645-81061645 on Build GRCh38
rs138763234 Chr.16: 81061711-81061711 on Build GRCh38
rs577390951 Chr.16: 81061360-81061360 on Build GRCh38
rs537023465 Chr.16: 81061521-81061521 on Build GRCh38
rs1055414252 Chr.16: 81061241-81061241 on Build GRCh38
rs764639607 Chr.16: 81061640-81061640 on Build GRCh38
rs759487918 Chr.16: 81061314-81061314 on Build GRCh38
rs371039198 Chr.16: 81061734-81061734 on Build GRCh38
rs758276184 Chr.16: 81061646-81061646 on Build GRCh38
rs1032463722 Chr.16: 81061315-81061315 on Build GRCh38
rs375296318 Chr.16: 81061702-81061702 on Build GRCh38
rs542536304 Chr.16: 81061459-81061459 on Build GRCh38
rs766606694 Chr.16: 81061262-81061262 on Build GRCh38
rs766062970 Chr.16: 81061515-81061515 on Build GRCh38
rs753172995 Chr.16: 81061344-81061344 on Build GRCh38
rs553141790 Chr.16: 81061398-81061398 on Build GRCh38
rs899719788 Chr.16: 81061273-81061273 on Build GRCh38
rs535897034 Chr.16: 81061394-81061394 on Build GRCh38
rs553402061 Chr.16: 81061382-81061382 on Build GRCh38
rs761061877 Chr.16: 81061596-81061596 on Build GRCh38
rs759107267 Chr.16: 81061721-81061721 on Build GRCh38
rs748192210 Chr.16: 81061694-81061694 on Build GRCh38
rs143656674 Chr.16: 81061472-81061472 on Build GRCh38
rs774191102 Chr.16: 81061399-81061399 on Build GRCh38
rs1001016422 Chr.16: 81061310-81061310 on Build GRCh38
rs774548786 Chr.16: 81061328-81061328 on Build GRCh38
rs557650910 Chr.16: 81061738-81061738 on Build GRCh38
rs10459872 Chr.16: 81061346-81061346 on Build GRCh38
rs760017648 Chr.16: 81061484-81061484 on Build GRCh38
rs959018661 Chr.16: 81061531-81061531 on Build GRCh38
rs779697905 Chr.16: 81061461-81061461 on Build GRCh38
rs139506410 Chr.16: 81061495-81061495 on Build GRCh38
rs1030598778 Chr.16: 81061501-81061501 on Build GRCh38
rs777430599 Chr.16: 81061545-81061545 on Build GRCh38
rs776679658 Chr.16: 81061417-81061417 on Build GRCh38
rs755731515 Chr.16: 81061460-81061460 on Build GRCh38
rs749487394 Chr.16: 81061690-81061690 on Build GRCh38
rs907989539 Chr.16: 81061433-81061433 on Build GRCh38
rs34941481 Chr.16: 81061512-81061512 on Build GRCh38
rs140831378 Chr.16: 81061626-81061626 on Build GRCh38
rs753410237 Chr.16: 81061516-81061516 on Build GRCh38
rs995361182 Chr.16: 81061278-81061278 on Build GRCh38
rs888275177 Chr.16: 81061416-81061416 on Build GRCh38
rs764521396 Chr.16: 81061425-81061425 on Build GRCh38
rs369373244 Chr.16: 81061420-81061420 on Build GRCh38
rs758272225 Chr.16: 81061542-81061542 on Build GRCh38
rs7198494 Chr.16: 81061486-81061486 on Build GRCh38
rs148918343 Chr.16: 81061437-81061437 on Build GRCh38
rs778474371 Chr.16: 81061770-81061770 on Build GRCh38
rs753076132 Chr.16: 81061742-81061742 on Build GRCh38
rs762868784 Chr.16: 81061298-81061298 on Build GRCh38
rs773616881 Chr.16: 81061301-81061301 on Build GRCh38
rs917869299 Chr.16: 81061642-81061642 on Build GRCh38
rs780292731 Chr.16: 81061675-81061675 on Build GRCh38
rs981408414 Chr.16: 81061522-81061522 on Build GRCh38
rs142721987 Chr.16: 81061347-81061347 on Build GRCh38
rs745310372 Chr.16: 81061289-81061289 on Build GRCh38
rs757483318 Chr.16: 81061573-81061573 on Build GRCh38
rs184045841 Chr.16: 81061361-81061361 on Build GRCh38
rs146937896 Chr.16: 81061773-81061773 on Build GRCh38
rs755158909 Chr.16: 81061762-81061762 on Build GRCh38
rs1053550861 Chr.16: 81061724-81061724 on Build GRCh38
rs754151927 Chr.16: 81061759-81061759 on Build GRCh38
rs575214348 Chr.16: 81061746-81061746 on Build GRCh38
rs771196552 Chr.16: 81061412-81061412 on Build GRCh38
rs748197489 Chr.16: 81061763-81061763 on Build GRCh38
rs748008410 Chr.16: 81061469-81061469 on Build GRCh38
rs74827829 Chr.16: 81061584-81061584 on Build GRCh38
rs952203418 Chr.16: 81061336-81061336 on Build GRCh38
rs570890676 Chr.16: 81061567-81061567 on Build GRCh38
rs368711170 Chr.16: 81061297-81061297 on Build GRCh38
rs931276281 Chr.16: 81061717-81061717 on Build GRCh38
rs746831628 Chr.16: 81061665-81061665 on Build GRCh38
rs201595004 Chr.16: 81061783-81061783 on Build GRCh38
rs372673508 Chr.16: 81061468-81061468 on Build GRCh38
rs769864952 Chr.16: 81061291-81061291 on Build GRCh38
rs758812004 Chr.16: 81061345-81061345 on Build GRCh38
rs142706633 Chr.16: 81061587-81061587 on Build GRCh38
rs749304514 Chr.16: 81061464-81061464 on Build GRCh38
rs757512912 Chr.16: 81061431-81061431 on Build GRCh38
rs780653712 Chr.16: 81061362-81061362 on Build GRCh38
rs377170723 Chr.16: 81061482-81061482 on Build GRCh38
rs751971056 Chr.16: 81061428-81061428 on Build GRCh38
rs542547879 Chr.16: 81061729-81061729 on Build GRCh38
rs748015552 Chr.16: 81061529-81061529 on Build GRCh38
rs754360557 Chr.16: 81061228-81061228 on Build GRCh38
rs150112740 Chr.16: 81061709-81061709 on Build GRCh38
rs773716766 Chr.16: 81061594-81061594 on Build GRCh38
rs146750423 Chr.16: 81061564-81061564 on Build GRCh38
rs755504169 Chr.16: 81061242-81061242 on Build GRCh38
rs778851312 Chr.16: 81061693-81061693 on Build GRCh38
rs766150963 Chr.16: 81061604-81061604 on Build GRCh38
rs804885 Chr.16: 81061758-81061758 on Build GRCh38
rs762274829 Chr.16: 81061497-81061497 on Build GRCh38
rs775643584 Chr.16: 81061572-81061572 on Build GRCh38
rs201993583 Chr.16: 81061753-81061753 on Build GRCh38
rs764170243 Chr.16: 81061651-81061651 on Build GRCh38
rs757009701 Chr.16: 81061666-81061666 on Build GRCh38
rs867110811 Chr.16: 81061533-81061533 on Build GRCh38
rs767747223 Chr.16: 81061509-81061509 on Build GRCh38
rs200068614 Chr.16: 81061260-81061260 on Build GRCh38
rs776203898 Chr.16: 81061284-81061284 on Build GRCh38
rs959254609 Chr.16: 81061608-81061608 on Build GRCh38
rs200487806 Chr.16: 81061710-81061710 on Build GRCh38
rs575984281 Chr.16: 81061307-81061307 on Build GRCh38
rs762210126 Chr.16: 81061590-81061590 on Build GRCh38
rs755823631 Chr.16: 81061513-81061513 on Build GRCh38
rs368244002 Chr.16: 81061477-81061477 on Build GRCh38
rs936305112 Chr.16: 81061716-81061716 on Build GRCh38
rs767990825 Chr.16: 81061591-81061591 on Build GRCh38
rs757089640 Chr.16: 81061756-81061756 on Build GRCh38
rs768535543 Chr.16: 81061299-81061299 on Build GRCh38
rs568051084 Chr.16: 81061243-81061243 on Build GRCh38
rs758723933 Chr.16: 81061259-81061259 on Build GRCh38
rs1000567140 Chr.16: 81061493-81061493 on Build GRCh38
rs140378740 Chr.16: 81061571-81061571 on Build GRCh38
rs2287994 Chr.16: 81061467-81061467 on Build GRCh38
rs563994596 Chr.16: 81061479-81061479 on Build GRCh38
rs769358468 Chr.16: 81061784-81061784 on Build GRCh38
rs201809453 Chr.16: 81061473-81061473 on Build GRCh38
rs748550570 Chr.16: 81061410-81061410 on Build GRCh38
rs771489469 Chr.16: 81061775-81061775 on Build GRCh38
rs754567961 Chr.16: 81061618-81061618 on Build GRCh38
rs777140628 Chr.16: 81061782-81061782 on Build GRCh38
rs746655394 Chr.16: 81061272-81061272 on Build GRCh38
rs756510520 Chr.16: 81061744-81061749 on Build GRCh38

View Full Product Details