Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 77199106-77199682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MON1B
Entrez Gene ID
22879
Gene Name
MON1 homolog B, secretory trafficking associated
Gene Aliases
HSRG1, SAND2, SRG1
UniGene
Hs.513743
Gene Chromosome Location
Chr.16: 77190919-77199646 on Build GRCh38
Transcripts
NM_001286639.1 NM_001286640.1 NM_014940.3 NM_001129979.1

Gene Symbol
SYCE1L
Entrez Gene ID
100130958
Gene Name
synaptonemal complex central element protein 1 like
Gene Aliases
MRP2
UniGene
Hs.734779
Gene Chromosome Location
Chr.16: 77199452-77213079 on Build GRCh38
Transcripts
NM_001286639.1 NM_001286640.1 NM_014940.3 NM_001129979.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6581928 c.31G>A p.E11K Chr.16: 77199482-77199482 on Build GRCh38
4129374 c.10A>G p.K4E Chr.16: 77199461-77199461 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974831608 Chr.16: 77199518-77199518 on Build GRCh38
rs927014480 Chr.16: 77199296-77199296 on Build GRCh38
rs1002763653 Chr.16: 77199392-77199392 on Build GRCh38
rs1055717836 Chr.16: 77199486-77199486 on Build GRCh38
rs966668005 Chr.16: 77199430-77199430 on Build GRCh38
rs754218754 Chr.16: 77199510-77199510 on Build GRCh38
rs557773263 Chr.16: 77199630-77199630 on Build GRCh38
rs1025136002 Chr.16: 77199382-77199382 on Build GRCh38
rs894013950 Chr.16: 77199655-77199655 on Build GRCh38
rs989601357 Chr.16: 77199174-77199174 on Build GRCh38
rs1040616072 Chr.16: 77199634-77199634 on Build GRCh38
rs148882587 Chr.16: 77199240-77199240 on Build GRCh38
rs899053487 Chr.16: 77199468-77199468 on Build GRCh38
rs748765438 Chr.16: 77199148-77199148 on Build GRCh38
rs927625931 Chr.16: 77199632-77199632 on Build GRCh38
rs1033963396 Chr.16: 77199395-77199395 on Build GRCh38
rs913873268 Chr.16: 77199301-77199301 on Build GRCh38
rs1011141686 Chr.16: 77199415-77199415 on Build GRCh38
rs772486821 Chr.16: 77199384-77199384 on Build GRCh38
rs886735129 Chr.16: 77199379-77199379 on Build GRCh38
rs1054849137 Chr.16: 77199642-77199642 on Build GRCh38
rs761238363 Chr.16: 77199503-77199503 on Build GRCh38
rs1046168724 Chr.16: 77199465-77199465 on Build GRCh38
rs943863132 Chr.16: 77199331-77199331 on Build GRCh38
rs770974432 Chr.16: 77199448-77199448 on Build GRCh38
rs79760539 Chr.16: 77199559-77199559 on Build GRCh38
rs777470215 Chr.16: 77199141-77199141 on Build GRCh38
rs552313156 Chr.16: 77199511-77199511 on Build GRCh38
rs912407344 Chr.16: 77199328-77199328 on Build GRCh38
rs774461393 Chr.16: 77199308-77199308 on Build GRCh38
rs757888203 Chr.16: 77199438-77199438 on Build GRCh38
rs926651288 Chr.16: 77199162-77199162 on Build GRCh38
rs1036440947 Chr.16: 77199466-77199466 on Build GRCh38
rs576698148 Chr.16: 77199318-77199318 on Build GRCh38
rs962114381 Chr.16: 77199547-77199547 on Build GRCh38
rs11544783 Chr.16: 77199548-77199548 on Build GRCh38
rs111615388 Chr.16: 77199222-77199222 on Build GRCh38
rs1011078003 Chr.16: 77199656-77199656 on Build GRCh38
rs966741546 Chr.16: 77199499-77199499 on Build GRCh38
rs533039786 Chr.16: 77199429-77199429 on Build GRCh38
rs561643410 Chr.16: 77199341-77199341 on Build GRCh38
rs908248831 Chr.16: 77199457-77199457 on Build GRCh38
rs761911708 Chr.16: 77199477-77199477 on Build GRCh38
rs906740660 Chr.16: 77199388-77199388 on Build GRCh38
rs1020144577 Chr.16: 77199390-77199390 on Build GRCh38
rs186695481 Chr.16: 77199307-77199307 on Build GRCh38
rs572639117 Chr.16: 77199616-77199616 on Build GRCh38
rs988281089 Chr.16: 77199461-77199461 on Build GRCh38
rs373375255 Chr.16: 77199566-77199566 on Build GRCh38
rs1028843016 Chr.16: 77199352-77199352 on Build GRCh38
rs989255565 Chr.16: 77199404-77199404 on Build GRCh38
rs564208768 Chr.16: 77199368-77199368 on Build GRCh38
rs199890788 Chr.16: 77199512-77199512 on Build GRCh38
rs8398 Chr.16: 77199441-77199441 on Build GRCh38
rs1003894573 Chr.16: 77199265-77199265 on Build GRCh38
rs116855347 Chr.16: 77199197-77199197 on Build GRCh38
rs895465627 Chr.16: 77199326-77199326 on Build GRCh38
rs1039995682 Chr.16: 77199215-77199215 on Build GRCh38
rs572007822 Chr.16: 77199350-77199350 on Build GRCh38
rs1034593868 Chr.16: 77199530-77199530 on Build GRCh38
rs369924114 Chr.16: 77199456-77199456 on Build GRCh38
rs1003925793 Chr.16: 77199380-77199380 on Build GRCh38
rs931232480 Chr.16: 77199445-77199445 on Build GRCh38
rs913967906 Chr.16: 77199175-77199175 on Build GRCh38
rs535625150 Chr.16: 77199587-77199587 on Build GRCh38
rs756075235 Chr.16: 77199471-77199472 on Build GRCh38
rs772245220 Chr.16: 77199496-77199496 on Build GRCh38
rs754198233 Chr.16: 77199422-77199422 on Build GRCh38
rs893942225 Chr.16: 77199490-77199490 on Build GRCh38
rs145600976 Chr.16: 77199226-77199226 on Build GRCh38
rs886846334 Chr.16: 77199251-77199251 on Build GRCh38
rs76154891 Chr.16: 77199617-77199617 on Build GRCh38
rs943973714 Chr.16: 77199214-77199214 on Build GRCh38
rs890933302 Chr.16: 77199202-77199202 on Build GRCh38
rs749575341 Chr.16: 77199294-77199294 on Build GRCh38
rs958858422 Chr.16: 77199531-77199531 on Build GRCh38
rs183542732 Chr.16: 77199292-77199292 on Build GRCh38
rs1011172908 Chr.16: 77199494-77199494 on Build GRCh38
rs4888607 Chr.16: 77199497-77199497 on Build GRCh38
rs989465981 Chr.16: 77199295-77199295 on Build GRCh38
rs937106290 Chr.16: 77199297-77199297 on Build GRCh38
rs570203837 Chr.16: 77199454-77199454 on Build GRCh38
rs11640897 Chr.16: 77199176-77199176 on Build GRCh38
rs1041809331 Chr.16: 77199657-77199657 on Build GRCh38
rs1020493882 Chr.16: 77199409-77199409 on Build GRCh38
rs945889210 Chr.16: 77199194-77199194 on Build GRCh38
rs113908772 Chr.16: 77199327-77199327 on Build GRCh38
rs367810281 Chr.16: 77199400-77199400 on Build GRCh38
rs952540401 Chr.16: 77199543-77199543 on Build GRCh38
rs77430739 Chr.16: 77199505-77199505 on Build GRCh38
rs11544784 Chr.16: 77199433-77199433 on Build GRCh38
rs936647197 Chr.16: 77199172-77199172 on Build GRCh38
rs1020972362 Chr.16: 77199421-77199421 on Build GRCh38
rs11640619 Chr.16: 77199271-77199271 on Build GRCh38
rs112674196 Chr.16: 77199473-77199473 on Build GRCh38
rs117687745 Chr.16: 77199638-77199638 on Build GRCh38
rs971268336 Chr.16: 77199274-77199274 on Build GRCh38
rs899120741 Chr.16: 77199343-77199343 on Build GRCh38
rs963230357 Chr.16: 77199201-77199201 on Build GRCh38
rs937536329 Chr.16: 77199435-77199435 on Build GRCh38
rs111823947 Chr.16: 77199442-77199442 on Build GRCh38
rs145052441 Chr.16: 77199591-77199591 on Build GRCh38
rs985429325 Chr.16: 77199227-77199227 on Build GRCh38
rs775165602 Chr.16: 77199623-77199623 on Build GRCh38
rs971021394 Chr.16: 77199127-77199127 on Build GRCh38
rs931008892 Chr.16: 77199355-77199355 on Build GRCh38
rs1056736047 Chr.16: 77199138-77199138 on Build GRCh38
rs532227355 Chr.16: 77199475-77199475 on Build GRCh38
rs570238995 Chr.16: 77199288-77199288 on Build GRCh38
rs921068696 Chr.16: 77199443-77199443 on Build GRCh38
rs752430911 Chr.16: 77199539-77199539 on Build GRCh38
rs539529755 Chr.16: 77199533-77199533 on Build GRCh38
rs939729932 Chr.16: 77199578-77199578 on Build GRCh38
rs777479812 Chr.16: 77199562-77199562 on Build GRCh38
rs965171145 Chr.16: 77199516-77199516 on Build GRCh38
rs540626387 Chr.16: 77199362-77199362 on Build GRCh38
rs375185057 Chr.16: 77199278-77199278 on Build GRCh38
rs114764479 Chr.16: 77199432-77199432 on Build GRCh38
rs28676830 Chr.16: 77199338-77199338 on Build GRCh38
rs913506131 Chr.16: 77199283-77199283 on Build GRCh38
rs745591157 Chr.16: 77199464-77199464 on Build GRCh38

View Full Product Details