Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53808356-53808929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP12
Entrez Gene ID
91662
Gene Name
NLR family pyrin domain containing 12
Gene Aliases
CLR19.3, FCAS2, NALP12, PAN6, PYPAF7, RNO, RNO2
UniGene
Hs.631573
Gene Chromosome Location
Chr.19: 53793584-53824403 on Build GRCh38
Transcripts
NM_001277126.1 NM_001277129.1 NM_144687.3 XM_011527479.1 XM_011527480.1 XM_011527482.1 XM_017027460.1 XM_017027461.1 XM_017027462.1 XM_017027463.1 XM_017027464.1 XM_017027465.1 XM_017027466.1 XM_017027467.1

SNP Detail

SNP ID Chromosome Location
rs763495626 Chr.19: 53808799-53808799 on Build GRCh38
rs530211129 Chr.19: 53808400-53808400 on Build GRCh38
rs199475866 Chr.19: 53808849-53808849 on Build GRCh38
rs568396938 Chr.19: 53808670-53808670 on Build GRCh38
rs972376544 Chr.19: 53808725-53808725 on Build GRCh38
rs1029151714 Chr.19: 53808885-53808885 on Build GRCh38
rs1011308346 Chr.19: 53808532-53808532 on Build GRCh38
rs891895979 Chr.19: 53808859-53808859 on Build GRCh38
rs138518826 Chr.19: 53808882-53808882 on Build GRCh38
rs773859971 Chr.19: 53808745-53808745 on Build GRCh38
rs1054442975 Chr.19: 53808546-53808547 on Build GRCh38
rs542408511 Chr.19: 53808879-53808879 on Build GRCh38
rs75857662 Chr.19: 53808693-53808693 on Build GRCh38
rs544469502 Chr.19: 53808379-53808379 on Build GRCh38
rs961944729 Chr.19: 53808770-53808770 on Build GRCh38
rs562864662 Chr.19: 53808397-53808397 on Build GRCh38
rs932748715 Chr.19: 53808481-53808481 on Build GRCh38
rs539120862 Chr.19: 53808715-53808715 on Build GRCh38
rs536677214 Chr.19: 53808791-53808791 on Build GRCh38
rs984423558 Chr.19: 53808422-53808422 on Build GRCh38
rs953805200 Chr.19: 53808676-53808677 on Build GRCh38
rs744759 Chr.19: 53808876-53808876 on Build GRCh38
rs562973129 Chr.19: 53808593-53808593 on Build GRCh38
rs552646453 Chr.19: 53808571-53808571 on Build GRCh38
rs1042794341 Chr.19: 53808887-53808887 on Build GRCh38
rs966284334 Chr.19: 53808606-53808606 on Build GRCh38
rs575605156 Chr.19: 53808781-53808781 on Build GRCh38
rs368914360 Chr.19: 53808635-53808635 on Build GRCh38
rs959154392 Chr.19: 53808708-53808708 on Build GRCh38
rs141702385 Chr.19: 53808499-53808499 on Build GRCh38
rs932812325 Chr.19: 53808802-53808802 on Build GRCh38
rs902445094 Chr.19: 53808658-53808658 on Build GRCh38
rs1010252876 Chr.19: 53808585-53808585 on Build GRCh38
rs1003431799 Chr.19: 53808661-53808661 on Build GRCh38
rs770242698 Chr.19: 53808743-53808743 on Build GRCh38
rs1044215926 Chr.19: 53808487-53808487 on Build GRCh38
rs879331211 Chr.19: 53808740-53808740 on Build GRCh38
rs751153604 Chr.19: 53808854-53808855 on Build GRCh38
rs891817223 Chr.19: 53808565-53808565 on Build GRCh38
rs889163 Chr.19: 53808851-53808851 on Build GRCh38
rs907514022 Chr.19: 53808754-53808754 on Build GRCh38
rs550075661 Chr.19: 53808624-53808624 on Build GRCh38
rs984306837 Chr.19: 53808685-53808686 on Build GRCh38
rs748572377 Chr.19: 53808733-53808733 on Build GRCh38
rs542093888 Chr.19: 53808428-53808428 on Build GRCh38
rs939940465 Chr.19: 53808384-53808384 on Build GRCh38
rs531757433 Chr.19: 53808618-53808618 on Build GRCh38
rs945990367 Chr.19: 53808866-53808866 on Build GRCh38
rs199475867 Chr.19: 53808703-53808703 on Build GRCh38
rs1042329370 Chr.19: 53808640-53808640 on Build GRCh38
rs1029617697 Chr.19: 53808655-53808655 on Build GRCh38
rs147090465 Chr.19: 53808747-53808747 on Build GRCh38
rs564436632 Chr.19: 53808617-53808617 on Build GRCh38
rs922745837 Chr.19: 53808464-53808464 on Build GRCh38
rs535628387 Chr.19: 53808672-53808672 on Build GRCh38
rs184050626 Chr.19: 53808701-53808701 on Build GRCh38
rs940227649 Chr.19: 53808450-53808450 on Build GRCh38
rs936084223 Chr.19: 53808822-53808822 on Build GRCh38
rs917184446 Chr.19: 53808453-53808453 on Build GRCh38
rs541075316 Chr.19: 53808793-53808793 on Build GRCh38
rs1041629405 Chr.19: 53808458-53808458 on Build GRCh38
rs192266445 Chr.19: 53808498-53808498 on Build GRCh38
rs766892668 Chr.19: 53808811-53808811 on Build GRCh38
rs374794370 Chr.19: 53808619-53808619 on Build GRCh38
rs1036880905 Chr.19: 53808404-53808404 on Build GRCh38

View Full Product Details