Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 125966274-125966794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PBX3
Entrez Gene ID
5090
Gene Name
PBX homeobox 3
Gene Aliases
-
UniGene
Hs.428027
Gene Chromosome Location
Chr.9: 125747338-125967377 on Build GRCh38
Transcripts
NM_001134778.1 NM_006195.5 NR_024122.1 NR_024123.1 XM_006717130.1 XM_006717131.1 XM_006717132.1 XM_006717133.1 XM_011518755.1 XM_017014786.1

SNP Detail

SNP ID Chromosome Location
rs1046268439 Chr.9: 125966515-125966515 on Build GRCh38
rs940719669 Chr.9: 125966735-125966735 on Build GRCh38
rs1007762512 Chr.9: 125966303-125966303 on Build GRCh38
rs915731939 Chr.9: 125966496-125966496 on Build GRCh38
rs758389895 Chr.9: 125966329-125966329 on Build GRCh38
rs73667816 Chr.9: 125966618-125966618 on Build GRCh38
rs949490569 Chr.9: 125966402-125966402 on Build GRCh38
rs781091407 Chr.9: 125966602-125966602 on Build GRCh38
rs1037970579 Chr.9: 125966455-125966455 on Build GRCh38
rs971677143 Chr.9: 125966693-125966693 on Build GRCh38
rs978014427 Chr.9: 125966316-125966316 on Build GRCh38
rs13296162 Chr.9: 125966434-125966434 on Build GRCh38
rs754736475 Chr.9: 125966565-125966565 on Build GRCh38
rs151283697 Chr.9: 125966607-125966607 on Build GRCh38
rs996347092 Chr.9: 125966488-125966488 on Build GRCh38
rs981281364 Chr.9: 125966698-125966698 on Build GRCh38
rs893376119 Chr.9: 125966300-125966300 on Build GRCh38
rs949880137 Chr.9: 125966508-125966508 on Build GRCh38
rs903265029 Chr.9: 125966521-125966521 on Build GRCh38
rs982466177 Chr.9: 125966679-125966679 on Build GRCh38
rs941294686 Chr.9: 125966458-125966458 on Build GRCh38
rs934674045 Chr.9: 125966534-125966534 on Build GRCh38
rs764229917 Chr.9: 125966620-125966620 on Build GRCh38
rs966217105 Chr.9: 125966323-125966323 on Build GRCh38
rs113094250 Chr.9: 125966439-125966439 on Build GRCh38
rs951119829 Chr.9: 125966659-125966659 on Build GRCh38
rs907923824 Chr.9: 125966715-125966715 on Build GRCh38
rs545074104 Chr.9: 125966613-125966613 on Build GRCh38
rs899391790 Chr.9: 125966485-125966485 on Build GRCh38
rs565690880 Chr.9: 125966353-125966353 on Build GRCh38
rs916673020 Chr.9: 125966382-125966382 on Build GRCh38
rs544015901 Chr.9: 125966493-125966493 on Build GRCh38
rs982206008 Chr.9: 125966695-125966695 on Build GRCh38
rs141512258 Chr.9: 125966628-125966628 on Build GRCh38
rs915822990 Chr.9: 125966686-125966686 on Build GRCh38
rs376294286 Chr.9: 125966366-125966366 on Build GRCh38
rs374619196 Chr.9: 125966614-125966614 on Build GRCh38
rs373063178 Chr.9: 125966355-125966355 on Build GRCh38
rs1054446820 Chr.9: 125966535-125966535 on Build GRCh38
rs878999337 Chr.9: 125966740-125966740 on Build GRCh38
rs951088861 Chr.9: 125966418-125966418 on Build GRCh38
rs990576320 Chr.9: 125966374-125966374 on Build GRCh38
rs529921372 Chr.9: 125966734-125966734 on Build GRCh38
rs940970217 Chr.9: 125966453-125966453 on Build GRCh38
rs558540272 Chr.9: 125966347-125966347 on Build GRCh38
rs576096520 Chr.9: 125966612-125966612 on Build GRCh38
rs1016572413 Chr.9: 125966682-125966682 on Build GRCh38
rs1018204567 Chr.9: 125966321-125966321 on Build GRCh38
rs991341869 Chr.9: 125966495-125966495 on Build GRCh38
rs374738745 Chr.9: 125966685-125966685 on Build GRCh38
rs545284875 Chr.9: 125966683-125966683 on Build GRCh38

View Full Product Details