Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 112889412-112889959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC46A2
Entrez Gene ID
57864
Gene Name
solute carrier family 46 member 2
Gene Aliases
Ly110, TSCOT
UniGene
Hs.512668
Gene Chromosome Location
Chr.9: 112878920-112890981 on Build GRCh38
Transcripts
NM_033051.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
752021 c.767T>C p.L256P Chr.9: 112889915-112889915 on Build GRCh38
3170388 c.727G>A p.V243M Chr.9: 112889955-112889955 on Build GRCh38
607314 c.1002C>T p.S334S Chr.9: 112889680-112889680 on Build GRCh38
752022 c.908T>C p.V303A Chr.9: 112889774-112889774 on Build GRCh38
4573029 c.942T>C p.G314G Chr.9: 112889740-112889740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773403462 Chr.9: 112889745-112889745 on Build GRCh38
rs768315734 Chr.9: 112889503-112889503 on Build GRCh38
rs748258582 Chr.9: 112889740-112889740 on Build GRCh38
rs955892861 Chr.9: 112889653-112889653 on Build GRCh38
rs979877412 Chr.9: 112889696-112889696 on Build GRCh38
rs141297560 Chr.9: 112889939-112889939 on Build GRCh38
rs200092270 Chr.9: 112889749-112889749 on Build GRCh38
rs928490542 Chr.9: 112889697-112889697 on Build GRCh38
rs752261062 Chr.9: 112889684-112889684 on Build GRCh38
rs558581867 Chr.9: 112889775-112889775 on Build GRCh38
rs976237620 Chr.9: 112889636-112889636 on Build GRCh38
rs753195812 Chr.9: 112889929-112889929 on Build GRCh38
rs749370697 Chr.9: 112889935-112889935 on Build GRCh38
rs774922710 Chr.9: 112889548-112889548 on Build GRCh38
rs769223313 Chr.9: 112889725-112889725 on Build GRCh38
rs907284823 Chr.9: 112889501-112889501 on Build GRCh38
rs756670957 Chr.9: 112889932-112889932 on Build GRCh38
rs754422036 Chr.9: 112889597-112889597 on Build GRCh38
rs780729364 Chr.9: 112889514-112889514 on Build GRCh38
rs780000210 Chr.9: 112889592-112889592 on Build GRCh38
rs762093438 Chr.9: 112889583-112889583 on Build GRCh38
rs201410747 Chr.9: 112889795-112889795 on Build GRCh38
rs747881413 Chr.9: 112889522-112889522 on Build GRCh38
rs914526902 Chr.9: 112889443-112889443 on Build GRCh38
rs139403102 Chr.9: 112889743-112889743 on Build GRCh38
rs765450547 Chr.9: 112889646-112889646 on Build GRCh38
rs370687401 Chr.9: 112889783-112889783 on Build GRCh38
rs775544516 Chr.9: 112889903-112889903 on Build GRCh38
rs61741119 Chr.9: 112889593-112889593 on Build GRCh38
rs948555594 Chr.9: 112889480-112889480 on Build GRCh38
rs765007038 Chr.9: 112889584-112889584 on Build GRCh38
rs777976291 Chr.9: 112889778-112889778 on Build GRCh38
rs751822632 Chr.9: 112889922-112889922 on Build GRCh38
rs867571666 Chr.9: 112889786-112889786 on Build GRCh38
rs759403730 Chr.9: 112889878-112889878 on Build GRCh38
rs746184523 Chr.9: 112889703-112889703 on Build GRCh38
rs760697229 Chr.9: 112889558-112889558 on Build GRCh38
rs558085502 Chr.9: 112889752-112889752 on Build GRCh38
rs771391307 Chr.9: 112889533-112889533 on Build GRCh38
rs755758076 Chr.9: 112889700-112889700 on Build GRCh38
rs191267096 Chr.9: 112889818-112889818 on Build GRCh38
rs1037530870 Chr.9: 112889706-112889706 on Build GRCh38
rs148086697 Chr.9: 112889624-112889624 on Build GRCh38
rs764973566 Chr.9: 112889921-112889921 on Build GRCh38
rs752211122 Chr.9: 112889776-112889776 on Build GRCh38
rs753416271 Chr.9: 112889915-112889915 on Build GRCh38
rs899105702 Chr.9: 112889834-112889834 on Build GRCh38
rs751419530 Chr.9: 112889667-112889667 on Build GRCh38
rs749497725 Chr.9: 112889861-112889861 on Build GRCh38
rs112054166 Chr.9: 112889913-112889913 on Build GRCh38
rs188435024 Chr.9: 112889606-112889606 on Build GRCh38
rs376496916 Chr.9: 112889670-112889670 on Build GRCh38
rs758806577 Chr.9: 112889705-112889705 on Build GRCh38
rs755008769 Chr.9: 112889668-112889668 on Build GRCh38
rs777772579 Chr.9: 112889933-112889933 on Build GRCh38
rs1048453722 Chr.9: 112889756-112889756 on Build GRCh38
rs780793473 Chr.9: 112889599-112889599 on Build GRCh38
rs753539221 Chr.9: 112889785-112889785 on Build GRCh38
rs117604258 Chr.9: 112889863-112889863 on Build GRCh38
rs7022287 Chr.9: 112889923-112889923 on Build GRCh38
rs773346229 Chr.9: 112889860-112889860 on Build GRCh38
rs369095004 Chr.9: 112889576-112889576 on Build GRCh38
rs758266564 Chr.9: 112889589-112889589 on Build GRCh38
rs756996058 Chr.9: 112889792-112889792 on Build GRCh38
rs752839127 Chr.9: 112889754-112889754 on Build GRCh38
rs145661725 Chr.9: 112889482-112889482 on Build GRCh38
rs146384415 Chr.9: 112889702-112889702 on Build GRCh38
rs772716159 Chr.9: 112889551-112889551 on Build GRCh38
rs990482887 Chr.9: 112889438-112889438 on Build GRCh38
rs766244345 Chr.9: 112889666-112889666 on Build GRCh38
rs368884299 Chr.9: 112889899-112889899 on Build GRCh38
rs772953331 Chr.9: 112889652-112889652 on Build GRCh38
rs775840775 Chr.9: 112889627-112889627 on Build GRCh38
rs754025994 Chr.9: 112889633-112889633 on Build GRCh38
rs372648155 Chr.9: 112889712-112889712 on Build GRCh38
rs144886693 Chr.9: 112889870-112889870 on Build GRCh38
rs207470497 Chr.9: 112889550-112889550 on Build GRCh38
rs1037012102 Chr.9: 112889536-112889536 on Build GRCh38
rs755100218 Chr.9: 112889655-112889655 on Build GRCh38
rs761309006 Chr.9: 112889919-112889919 on Build GRCh38
rs769832225 Chr.9: 112889831-112889831 on Build GRCh38
rs747405802 Chr.9: 112889798-112889798 on Build GRCh38
rs761287848 Chr.9: 112889628-112889628 on Build GRCh38
rs200558726 Chr.9: 112889728-112889728 on Build GRCh38
rs549699756 Chr.9: 112889600-112889600 on Build GRCh38
rs377646939 Chr.9: 112889888-112889888 on Build GRCh38
rs746235984 Chr.9: 112889619-112889619 on Build GRCh38
rs375417558 Chr.9: 112889794-112889794 on Build GRCh38
rs16917454 Chr.9: 112889585-112889585 on Build GRCh38
rs199805390 Chr.9: 112889936-112889936 on Build GRCh38
rs747461684 Chr.9: 112889716-112889716 on Build GRCh38
rs763201086 Chr.9: 112889746-112889746 on Build GRCh38
rs914473532 Chr.9: 112889659-112889659 on Build GRCh38
rs779359370 Chr.9: 112889617-112889617 on Build GRCh38
rs7021942 Chr.9: 112889905-112889905 on Build GRCh38
rs142845353 Chr.9: 112889557-112889557 on Build GRCh38
rs527518235 Chr.9: 112889481-112889481 on Build GRCh38
rs755400329 Chr.9: 112889799-112889799 on Build GRCh38
rs781734759 Chr.9: 112889812-112889812 on Build GRCh38
rs768618452 Chr.9: 112889629-112889629 on Build GRCh38
rs764287255 Chr.9: 112889572-112889572 on Build GRCh38
rs938778700 Chr.9: 112889525-112889525 on Build GRCh38
rs761965885 Chr.9: 112889645-112889645 on Build GRCh38

View Full Product Details