Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70301680-70302197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF4A
Entrez Gene ID
24137
Gene Name
kinesin family member 4A
Gene Aliases
KIF4, KIF4G1, MRX100
UniGene
Hs.648326
Gene Chromosome Location
Chr.X: 70290029-70420924 on Build GRCh38
Transcripts
NM_012310.4

SNP Detail

SNP ID Chromosome Location
rs765460858 Chr.X: 70302120-70302121 on Build GRCh38
rs901159564 Chr.X: 70301992-70301992 on Build GRCh38
rs1044979780 Chr.X: 70301786-70301786 on Build GRCh38
rs747990121 Chr.X: 70302102-70302102 on Build GRCh38
rs767167397 Chr.X: 70301740-70301740 on Build GRCh38
rs1049622734 Chr.X: 70301838-70301838 on Build GRCh38
rs776683982 Chr.X: 70301875-70301875 on Build GRCh38
rs1041149699 Chr.X: 70301934-70301934 on Build GRCh38
rs559410302 Chr.X: 70301872-70301872 on Build GRCh38
rs956700667 Chr.X: 70301705-70301705 on Build GRCh38
rs1199449 Chr.X: 70301767-70301767 on Build GRCh38
rs373592690 Chr.X: 70301779-70301779 on Build GRCh38
rs778651302 Chr.X: 70302073-70302073 on Build GRCh38
rs987807484 Chr.X: 70301737-70301737 on Build GRCh38
rs368141479 Chr.X: 70302043-70302043 on Build GRCh38
rs1048736615 Chr.X: 70301854-70301854 on Build GRCh38
rs761818735 Chr.X: 70302077-70302077 on Build GRCh38
rs764648571 Chr.X: 70302053-70302053 on Build GRCh38
rs759850769 Chr.X: 70301877-70301877 on Build GRCh38
rs754941471 Chr.X: 70302016-70302016 on Build GRCh38
rs887195106 Chr.X: 70301858-70301858 on Build GRCh38
rs760620348 Chr.X: 70301861-70301861 on Build GRCh38
rs751867510 Chr.X: 70302075-70302075 on Build GRCh38
rs762259797 Chr.X: 70302081-70302082 on Build GRCh38
rs765682595 Chr.X: 70301906-70301906 on Build GRCh38
rs753179678 Chr.X: 70302031-70302031 on Build GRCh38
rs1038448207 Chr.X: 70301844-70301844 on Build GRCh38
rs1004727284 Chr.X: 70301901-70301901 on Build GRCh38
rs760577792 Chr.X: 70301718-70301726 on Build GRCh38
rs755648523 Chr.X: 70301803-70301803 on Build GRCh38
rs779521430 Chr.X: 70301810-70301810 on Build GRCh38
rs1028500029 Chr.X: 70302074-70302074 on Build GRCh38

View Full Product Details