Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 80025395-80025979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX22
Entrez Gene ID
50945
Gene Name
T-box 22
Gene Aliases
ABERS, CLPA, CPX, TBXX, dJ795G23.1
UniGene
Hs.374253
Gene Chromosome Location
Chr.X: 80014730-80035828 on Build GRCh38
Transcripts
NM_001109878.1 NM_001109879.1 NM_001303475.1 NM_016954.2 XM_005262136.2 XM_006724657.3 XM_011530972.1

SNP Detail

SNP ID Chromosome Location
rs776905471 Chr.X: 80025736-80025736 on Build GRCh38
rs386825919 Chr.X: 80025896-80025898 on Build GRCh38
rs776093447 Chr.X: 80025752-80025752 on Build GRCh38
rs146024976 Chr.X: 80025693-80025693 on Build GRCh38
rs193027778 Chr.X: 80025430-80025430 on Build GRCh38
rs546083415 Chr.X: 80025807-80025807 on Build GRCh38
rs991437182 Chr.X: 80025868-80025868 on Build GRCh38
rs754958166 Chr.X: 80025687-80025687 on Build GRCh38
rs779543847 Chr.X: 80025683-80025683 on Build GRCh38
rs185126190 Chr.X: 80025952-80025952 on Build GRCh38
rs755560775 Chr.X: 80025681-80025681 on Build GRCh38
rs1052008333 Chr.X: 80025512-80025512 on Build GRCh38
rs761492404 Chr.X: 80025654-80025654 on Build GRCh38
rs755501455 Chr.X: 80025670-80025670 on Build GRCh38
rs753047555 Chr.X: 80025652-80025652 on Build GRCh38
rs771150368 Chr.X: 80025584-80025584 on Build GRCh38
rs763177430 Chr.X: 80025764-80025764 on Build GRCh38
rs896034937 Chr.X: 80025524-80025524 on Build GRCh38
rs195294 Chr.X: 80025898-80025898 on Build GRCh38
rs753232034 Chr.X: 80025699-80025699 on Build GRCh38
rs760190777 Chr.X: 80025613-80025613 on Build GRCh38
rs747262805 Chr.X: 80025716-80025716 on Build GRCh38
rs1001633015 Chr.X: 80025477-80025477 on Build GRCh38
rs770461385 Chr.X: 80025739-80025739 on Build GRCh38
rs754217169 Chr.X: 80025445-80025445 on Build GRCh38
rs911732619 Chr.X: 80025766-80025766 on Build GRCh38
rs370955267 Chr.X: 80025781-80025781 on Build GRCh38
rs1035833357 Chr.X: 80025496-80025496 on Build GRCh38
rs752696388 Chr.X: 80025826-80025826 on Build GRCh38
rs199593335 Chr.X: 80025810-80025810 on Build GRCh38
rs374171441 Chr.X: 80025684-80025684 on Build GRCh38
rs767034098 Chr.X: 80025656-80025656 on Build GRCh38
rs760626449 Chr.X: 80025559-80025559 on Build GRCh38
rs781287118 Chr.X: 80025593-80025594 on Build GRCh38
rs1035554884 Chr.X: 80025848-80025848 on Build GRCh38
rs751807223 Chr.X: 80025937-80025937 on Build GRCh38
rs1005546228 Chr.X: 80025589-80025589 on Build GRCh38
rs34244923 Chr.X: 80025703-80025703 on Build GRCh38
rs200542790 Chr.X: 80025737-80025737 on Build GRCh38
rs796291421 Chr.X: 80025565-80025565 on Build GRCh38
rs906022144 Chr.X: 80025448-80025448 on Build GRCh38
rs765036287 Chr.X: 80025825-80025825 on Build GRCh38
rs773515696 Chr.X: 80025553-80025553 on Build GRCh38
rs764530704 Chr.X: 80025772-80025772 on Build GRCh38
rs993952750 Chr.X: 80025715-80025715 on Build GRCh38
rs560252054 Chr.X: 80025928-80025928 on Build GRCh38
rs754515624 Chr.X: 80025821-80025821 on Build GRCh38
rs958593934 Chr.X: 80025865-80025865 on Build GRCh38
rs772672988 Chr.X: 80025773-80025773 on Build GRCh38
rs761336597 Chr.X: 80025957-80025957 on Build GRCh38
rs200060292 Chr.X: 80025598-80025598 on Build GRCh38
rs963408036 Chr.X: 80025658-80025658 on Build GRCh38
rs765734398 Chr.X: 80025698-80025698 on Build GRCh38
rs765680258 Chr.X: 80025632-80025632 on Build GRCh38
rs376259951 Chr.X: 80025777-80025777 on Build GRCh38
rs778648477 Chr.X: 80025691-80025691 on Build GRCh38
rs201139211 Chr.X: 80025883-80025884 on Build GRCh38
rs914189434 Chr.X: 80025882-80025882 on Build GRCh38
rs779774947 Chr.X: 80025844-80025844 on Build GRCh38
rs143779422 Chr.X: 80025896-80025896 on Build GRCh38

View Full Product Details