Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112647326-112647886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARP7
Entrez Gene ID
51574
Gene Name
La ribonucleoprotein domain family member 7
Gene Aliases
ALAZS, HDCMA18P, PIP7S
UniGene
Hs.713663
Gene Chromosome Location
Chr.4: 112636964-112657592 on Build GRCh38
Transcripts
NM_001267039.1 NM_015454.2 NM_016648.3 NR_049768.1 NR_029860.1

Gene Symbol
MIR367
Entrez Gene ID
442912
Gene Name
microRNA 367
Gene Aliases
MIRN367, hsa-mir-367
UniGene
Gene Chromosome Location
Chr.4: 112647874-112647941 on Build GRCh38
Transcripts
NM_001267039.1 NM_015454.2 NM_016648.3 NR_049768.1 NR_029860.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1256334 c.794C>A p.S265Y Chr.4: 112647346-112647346 on Build GRCh38
5174513 c.1050_1051delCT p.L353fs*7 Chr.4: 112647742-112647743 on Build GRCh38
1319389 c.1013C>T p.T338I Chr.4: 112647705-112647705 on Build GRCh38
3824983 c.1008C>T p.I336I Chr.4: 112647700-112647700 on Build GRCh38
5474669 c.813G>T p.Q271H Chr.4: 112647365-112647365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761645480 Chr.4: 112647696-112647696 on Build GRCh38
rs750405942 Chr.4: 112647704-112647704 on Build GRCh38
rs1057519017 Chr.4: 112647722-112647723 on Build GRCh38
rs779380562 Chr.4: 112647742-112647743 on Build GRCh38
rs371035787 Chr.4: 112647478-112647478 on Build GRCh38
rs370622429 Chr.4: 112647586-112647587 on Build GRCh38
rs375829533 Chr.4: 112647366-112647366 on Build GRCh38
rs778892987 Chr.4: 112647581-112647581 on Build GRCh38
rs752382139 Chr.4: 112647552-112647552 on Build GRCh38
rs931666699 Chr.4: 112647809-112647809 on Build GRCh38
rs756465661 Chr.4: 112647840-112647841 on Build GRCh38
rs201250325 Chr.4: 112647387-112647387 on Build GRCh38
rs764744775 Chr.4: 112647471-112647471 on Build GRCh38
rs372159818 Chr.4: 112647583-112647583 on Build GRCh38
rs764693485 Chr.4: 112647380-112647380 on Build GRCh38
rs757205617 Chr.4: 112647575-112647575 on Build GRCh38
rs147971163 Chr.4: 112647665-112647665 on Build GRCh38
rs771384465 Chr.4: 112647588-112647603 on Build GRCh38
rs757874580 Chr.4: 112647480-112647499 on Build GRCh38
rs578224265 Chr.4: 112647855-112647856 on Build GRCh38
rs772451147 Chr.4: 112647780-112647781 on Build GRCh38
rs201535779 Chr.4: 112647663-112647663 on Build GRCh38
rs780240533 Chr.4: 112647596-112647596 on Build GRCh38
rs200443782 Chr.4: 112647563-112647563 on Build GRCh38
rs749872437 Chr.4: 112647696-112647732 on Build GRCh38
rs990748843 Chr.4: 112647470-112647470 on Build GRCh38
rs760626307 Chr.4: 112647598-112647601 on Build GRCh38
rs765102180 Chr.4: 112647833-112647833 on Build GRCh38
rs763050541 Chr.4: 112647839-112647839 on Build GRCh38
rs765820560 Chr.4: 112647481-112647481 on Build GRCh38
rs373634124 Chr.4: 112647401-112647401 on Build GRCh38
rs749625960 Chr.4: 112647588-112647602 on Build GRCh38
rs767678965 Chr.4: 112647590-112647593 on Build GRCh38
rs750126754 Chr.4: 112647837-112647837 on Build GRCh38
rs71597609 Chr.4: 112647587-112647588 on Build GRCh38
rs780287133 Chr.4: 112647715-112647715 on Build GRCh38
rs765125762 Chr.4: 112647702-112647702 on Build GRCh38
rs528737801 Chr.4: 112647733-112647733 on Build GRCh38
rs746034480 Chr.4: 112647750-112647760 on Build GRCh38
rs778127703 Chr.4: 112647674-112647675 on Build GRCh38
rs751686070 Chr.4: 112647717-112647717 on Build GRCh38
rs763929099 Chr.4: 112647380-112647381 on Build GRCh38
rs755081512 Chr.4: 112647852-112647852 on Build GRCh38
rs975850028 Chr.4: 112647771-112647771 on Build GRCh38
rs541057628 Chr.4: 112647460-112647460 on Build GRCh38
rs777079200 Chr.4: 112647526-112647526 on Build GRCh38
rs768849339 Chr.4: 112647802-112647804 on Build GRCh38
rs70958470 Chr.4: 112647616-112647617 on Build GRCh38
rs755310911 Chr.4: 112647721-112647721 on Build GRCh38
rs781652858 Chr.4: 112647611-112647611 on Build GRCh38
rs772265784 Chr.4: 112647436-112647436 on Build GRCh38
rs62317770 Chr.4: 112647388-112647388 on Build GRCh38
rs775781419 Chr.4: 112647448-112647448 on Build GRCh38
rs778420137 Chr.4: 112647386-112647387 on Build GRCh38
rs763887078 Chr.4: 112647566-112647566 on Build GRCh38
rs773792953 Chr.4: 112647539-112647539 on Build GRCh38
rs761860253 Chr.4: 112647823-112647833 on Build GRCh38
rs767188889 Chr.4: 112647493-112647493 on Build GRCh38
rs754602989 Chr.4: 112647389-112647389 on Build GRCh38
rs750249723 Chr.4: 112647842-112647843 on Build GRCh38
rs767792487 Chr.4: 112647369-112647369 on Build GRCh38
rs777508785 Chr.4: 112647503-112647503 on Build GRCh38
rs773773118 Chr.4: 112647362-112647362 on Build GRCh38
rs747249439 Chr.4: 112647597-112647597 on Build GRCh38
rs921664262 Chr.4: 112647772-112647772 on Build GRCh38
rs770312658 Chr.4: 112647569-112647569 on Build GRCh38
rs201212143 Chr.4: 112647378-112647378 on Build GRCh38
rs757958464 Chr.4: 112647385-112647385 on Build GRCh38
rs368186137 Chr.4: 112647760-112647760 on Build GRCh38
rs766407669 Chr.4: 112647843-112647843 on Build GRCh38
rs201727777 Chr.4: 112647402-112647402 on Build GRCh38
rs200072154 Chr.4: 112647530-112647530 on Build GRCh38
rs368723744 Chr.4: 112647853-112647853 on Build GRCh38
rs753043822 Chr.4: 112647371-112647371 on Build GRCh38
rs1023594873 Chr.4: 112647406-112647406 on Build GRCh38
rs532966854 Chr.4: 112647613-112647613 on Build GRCh38
rs770950781 Chr.4: 112647423-112647423 on Build GRCh38
rs771357243 Chr.4: 112647751-112647751 on Build GRCh38
rs200831675 Chr.4: 112647693-112647693 on Build GRCh38
rs768933749 Chr.4: 112647600-112647600 on Build GRCh38
rs969327433 Chr.4: 112647445-112647445 on Build GRCh38
rs748533852 Chr.4: 112647617-112647617 on Build GRCh38
rs779500751 Chr.4: 112647630-112647631 on Build GRCh38
rs759067833 Chr.4: 112647540-112647540 on Build GRCh38
rs900139242 Chr.4: 112647349-112647349 on Build GRCh38
rs754655165 Chr.4: 112647491-112647491 on Build GRCh38
rs751017218 Chr.4: 112647486-112647486 on Build GRCh38
rs750024669 Chr.4: 112647605-112647608 on Build GRCh38
rs764144078 Chr.4: 112647598-112647599 on Build GRCh38
rs752990563 Chr.4: 112647863-112647863 on Build GRCh38
rs748605983 Chr.4: 112647581-112647584 on Build GRCh38
rs907718553 Chr.4: 112647711-112647711 on Build GRCh38
rs772240434 Chr.4: 112647518-112647518 on Build GRCh38
rs377090014 Chr.4: 112647545-112647545 on Build GRCh38
rs756166445 Chr.4: 112647371-112647376 on Build GRCh38
rs759582573 Chr.4: 112647522-112647522 on Build GRCh38
rs374940366 Chr.4: 112647846-112647846 on Build GRCh38
rs146962383 Chr.4: 112647664-112647664 on Build GRCh38
rs965500951 Chr.4: 112647768-112647768 on Build GRCh38
rs745604512 Chr.4: 112647391-112647392 on Build GRCh38
rs768343315 Chr.4: 112647807-112647807 on Build GRCh38
rs753558578 Chr.4: 112647572-112647572 on Build GRCh38
rs774437783 Chr.4: 112647424-112647424 on Build GRCh38
rs776404304 Chr.4: 112647812-112647821 on Build GRCh38
rs959431444 Chr.4: 112647671-112647672 on Build GRCh38
rs771165560 Chr.4: 112647733-112647734 on Build GRCh38
rs754365322 Chr.4: 112647473-112647473 on Build GRCh38
rs770316916 Chr.4: 112647581-112647598 on Build GRCh38
rs776314449 Chr.4: 112647829-112647829 on Build GRCh38
rs375697987 Chr.4: 112647502-112647502 on Build GRCh38
rs769173145 Chr.4: 112647523-112647523 on Build GRCh38
rs775657157 Chr.4: 112647782-112647785 on Build GRCh38
rs775730829 Chr.4: 112647590-112647591 on Build GRCh38
rs900175358 Chr.4: 112647535-112647535 on Build GRCh38
rs761264490 Chr.4: 112647374-112647374 on Build GRCh38
rs866288154 Chr.4: 112647477-112647477 on Build GRCh38
rs773740501 Chr.4: 112647621-112647621 on Build GRCh38
rs749819793 Chr.4: 112647746-112647746 on Build GRCh38
rs770227975 Chr.4: 112647737-112647737 on Build GRCh38
rs937970407 Chr.4: 112647442-112647442 on Build GRCh38
rs368343990 Chr.4: 112647515-112647515 on Build GRCh38
rs189806015 Chr.4: 112647705-112647705 on Build GRCh38
rs758068797 Chr.4: 112647608-112647609 on Build GRCh38
rs566464249 Chr.4: 112647784-112647785 on Build GRCh38
rs1033036917 Chr.4: 112647725-112647725 on Build GRCh38
rs777955582 Chr.4: 112647742-112647742 on Build GRCh38
rs747933441 Chr.4: 112647391-112647391 on Build GRCh38
rs988752351 Chr.4: 112647757-112647757 on Build GRCh38
rs751072973 Chr.4: 112647672-112647674 on Build GRCh38
rs765771089 Chr.4: 112647534-112647534 on Build GRCh38
rs765122296 Chr.4: 112647603-112647617 on Build GRCh38
rs917902754 Chr.4: 112647694-112647694 on Build GRCh38
rs757156044 Chr.4: 112647512-112647512 on Build GRCh38
rs763729793 Chr.4: 112647692-112647692 on Build GRCh38
rs761511399 Chr.4: 112647602-112647603 on Build GRCh38
rs529243678 Chr.4: 112647367-112647367 on Build GRCh38
rs753874572 Chr.4: 112647602-112647602 on Build GRCh38
rs756554653 Chr.4: 112647675-112647678 on Build GRCh38
rs749413494 Chr.4: 112647408-112647408 on Build GRCh38
rs763333341 Chr.4: 112647640-112647640 on Build GRCh38
rs745982052 Chr.4: 112647582-112647582 on Build GRCh38
rs775043949 Chr.4: 112647756-112647756 on Build GRCh38
rs997074760 Chr.4: 112647350-112647350 on Build GRCh38
rs577197566 Chr.4: 112647426-112647426 on Build GRCh38
rs140741616 Chr.4: 112647832-112647832 on Build GRCh38
rs979666067 Chr.4: 112647472-112647472 on Build GRCh38
rs770501465 Chr.4: 112647509-112647511 on Build GRCh38
rs751739317 Chr.4: 112647844-112647844 on Build GRCh38
rs781544374 Chr.4: 112647723-112647723 on Build GRCh38

View Full Product Details