Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 146592833-146593358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP2R2B
Entrez Gene ID
5521
Gene Name
protein phosphatase 2 regulatory subunit Bbeta
Gene Aliases
B55BETA, PP2AB55BETA, PP2ABBETA, PP2APR55B, PP2APR55BETA, PR2AB55BETA, PR2ABBETA, PR2APR55BETA, PR52B, PR55-BETA, PR55BETA, SCA12
UniGene
Hs.627618
Gene Chromosome Location
Chr.5: 146589504-147081520 on Build GRCh38
Transcripts
NM_001271899.1 NM_001271900.1 NM_001271948.1 NM_181674.2 NM_181675.3 NM_181676.2 NM_181677.2 NM_181678.2 NR_073526.1 NR_073527.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5901401 c.1018G>A p.E340K Chr.5: 146593023-146593023 on Build GRCh38
1434941 c.1036_1037delTG p.W346fs*26 Chr.5: 146592986-146592987 on Build GRCh38
3947111 c.1061G>T p.G354V Chr.5: 146592980-146592980 on Build GRCh38
3947109 c.1010G>T p.G337V Chr.5: 146592980-146592980 on Build GRCh38
1434940 c.1045_1046delTG p.W349fs*26 Chr.5: 146592986-146592987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763364357 Chr.5: 146593110-146593110 on Build GRCh38
rs528429372 Chr.5: 146592873-146592873 on Build GRCh38
rs766902301 Chr.5: 146593113-146593113 on Build GRCh38
rs750102954 Chr.5: 146593105-146593105 on Build GRCh38
rs781377067 Chr.5: 146593030-146593030 on Build GRCh38
rs753409434 Chr.5: 146592942-146592942 on Build GRCh38
rs770113098 Chr.5: 146593054-146593054 on Build GRCh38
rs761355654 Chr.5: 146593092-146593092 on Build GRCh38
rs750854131 Chr.5: 146592977-146592977 on Build GRCh38
rs368510943 Chr.5: 146593065-146593065 on Build GRCh38
rs748278217 Chr.5: 146592933-146592934 on Build GRCh38
rs764882923 Chr.5: 146593102-146593102 on Build GRCh38
rs1019612179 Chr.5: 146593047-146593047 on Build GRCh38
rs113207618 Chr.5: 146593247-146593247 on Build GRCh38
rs996672271 Chr.5: 146593025-146593025 on Build GRCh38
rs886675929 Chr.5: 146592932-146592932 on Build GRCh38
rs1004238937 Chr.5: 146593024-146593024 on Build GRCh38
rs924389985 Chr.5: 146593129-146593129 on Build GRCh38
rs200347992 Chr.5: 146592872-146592872 on Build GRCh38
rs564237312 Chr.5: 146593122-146593122 on Build GRCh38
rs376231376 Chr.5: 146593080-146593080 on Build GRCh38
rs149430113 Chr.5: 146592989-146592989 on Build GRCh38
rs756170659 Chr.5: 146593262-146593262 on Build GRCh38
rs374253101 Chr.5: 146592943-146592943 on Build GRCh38
rs372581670 Chr.5: 146593076-146593076 on Build GRCh38
rs745597400 Chr.5: 146593074-146593074 on Build GRCh38
rs930974355 Chr.5: 146592916-146592916 on Build GRCh38
rs140554265 Chr.5: 146593266-146593266 on Build GRCh38
rs965659495 Chr.5: 146593056-146593056 on Build GRCh38
rs1056760048 Chr.5: 146593177-146593177 on Build GRCh38
rs199518697 Chr.5: 146593071-146593071 on Build GRCh38
rs576664594 Chr.5: 146593222-146593222 on Build GRCh38
rs189529065 Chr.5: 146593324-146593324 on Build GRCh38
rs80005726 Chr.5: 146593228-146593228 on Build GRCh38
rs919685453 Chr.5: 146593290-146593290 on Build GRCh38
rs934399433 Chr.5: 146593168-146593168 on Build GRCh38
rs77148477 Chr.5: 146592933-146592933 on Build GRCh38
rs899047713 Chr.5: 146593256-146593256 on Build GRCh38
rs1044659139 Chr.5: 146592859-146592859 on Build GRCh38
rs747072131 Chr.5: 146593298-146593298 on Build GRCh38
rs748318197 Chr.5: 146593046-146593046 on Build GRCh38
rs1049340732 Chr.5: 146592920-146592920 on Build GRCh38
rs776267459 Chr.5: 146593090-146593090 on Build GRCh38
rs765511385 Chr.5: 146592961-146592961 on Build GRCh38
rs116519070 Chr.5: 146593183-146593183 on Build GRCh38
rs566482178 Chr.5: 146593226-146593226 on Build GRCh38
rs758845118 Chr.5: 146592987-146592987 on Build GRCh38
rs546868441 Chr.5: 146593214-146593214 on Build GRCh38
rs550778853 Chr.5: 146593084-146593084 on Build GRCh38
rs772315664 Chr.5: 146593062-146593062 on Build GRCh38
rs752137263 Chr.5: 146593012-146593012 on Build GRCh38
rs546721589 Chr.5: 146593189-146593189 on Build GRCh38
rs371540157 Chr.5: 146593081-146593081 on Build GRCh38
rs751673352 Chr.5: 146592867-146592867 on Build GRCh38
rs755089832 Chr.5: 146593021-146593021 on Build GRCh38
rs760136602 Chr.5: 146592928-146592928 on Build GRCh38

View Full Product Details