Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177595142-177595689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMED9
Entrez Gene ID
54732
Gene Name
transmembrane p24 trafficking protein 9
Gene Aliases
GMP25, HSGP25L2G, p24a2, p25
UniGene
Hs.279929
Gene Chromosome Location
Chr.5: 177592212-177596107 on Build GRCh38
Transcripts
NM_017510.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1696807 c.664A>G p.M222V Chr.5: 177595372-177595372 on Build GRCh38
1672159 c.668G>T p.R223L Chr.5: 177595376-177595376 on Build GRCh38
5995976 c.573C>A p.R191R Chr.5: 177595281-177595281 on Build GRCh38
1436365 c.636C>T p.L212L Chr.5: 177595344-177595344 on Build GRCh38
6513678 c.609G>A p.V203V Chr.5: 177595317-177595317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546091419 Chr.5: 177595485-177595485 on Build GRCh38
rs1011355438 Chr.5: 177595518-177595518 on Build GRCh38
rs773806110 Chr.5: 177595312-177595312 on Build GRCh38
rs200819997 Chr.5: 177595363-177595363 on Build GRCh38
rs781524933 Chr.5: 177595610-177595610 on Build GRCh38
rs547207372 Chr.5: 177595216-177595216 on Build GRCh38
rs781540423 Chr.5: 177595267-177595267 on Build GRCh38
rs139864777 Chr.5: 177595212-177595212 on Build GRCh38
rs749135860 Chr.5: 177595383-177595383 on Build GRCh38
rs747432853 Chr.5: 177595535-177595535 on Build GRCh38
rs962219070 Chr.5: 177595631-177595631 on Build GRCh38
rs866118614 Chr.5: 177595328-177595328 on Build GRCh38
rs201432047 Chr.5: 177595263-177595263 on Build GRCh38
rs752291089 Chr.5: 177595320-177595320 on Build GRCh38
rs912390027 Chr.5: 177595529-177595530 on Build GRCh38
rs751079009 Chr.5: 177595219-177595219 on Build GRCh38
rs760435425 Chr.5: 177595322-177595322 on Build GRCh38
rs564744757 Chr.5: 177595556-177595556 on Build GRCh38
rs750805126 Chr.5: 177595323-177595323 on Build GRCh38
rs958315621 Chr.5: 177595224-177595224 on Build GRCh38
rs371866140 Chr.5: 177595376-177595376 on Build GRCh38
rs781487309 Chr.5: 177595359-177595359 on Build GRCh38
rs1010905692 Chr.5: 177595245-177595245 on Build GRCh38
rs759028793 Chr.5: 177595314-177595314 on Build GRCh38
rs762737047 Chr.5: 177595294-177595294 on Build GRCh38
rs368474774 Chr.5: 177595271-177595271 on Build GRCh38
rs770764481 Chr.5: 177595386-177595386 on Build GRCh38
rs199622092 Chr.5: 177595280-177595280 on Build GRCh38
rs928154693 Chr.5: 177595335-177595335 on Build GRCh38
rs766890084 Chr.5: 177595465-177595465 on Build GRCh38
rs370667269 Chr.5: 177595350-177595350 on Build GRCh38
rs921210022 Chr.5: 177595474-177595474 on Build GRCh38
rs1023606323 Chr.5: 177595552-177595552 on Build GRCh38
rs760181009 Chr.5: 177595441-177595441 on Build GRCh38
rs773398674 Chr.5: 177595453-177595453 on Build GRCh38
rs995529509 Chr.5: 177595665-177595665 on Build GRCh38
rs1042087377 Chr.5: 177595235-177595235 on Build GRCh38
rs999183449 Chr.5: 177595201-177595201 on Build GRCh38
rs373990474 Chr.5: 177595264-177595264 on Build GRCh38
rs147493041 Chr.5: 177595183-177595183 on Build GRCh38
rs368292381 Chr.5: 177595625-177595625 on Build GRCh38
rs893034886 Chr.5: 177595481-177595481 on Build GRCh38
rs376717230 Chr.5: 177595443-177595443 on Build GRCh38
rs11545862 Chr.5: 177595372-177595372 on Build GRCh38
rs953924191 Chr.5: 177595479-177595479 on Build GRCh38
rs753590322 Chr.5: 177595270-177595270 on Build GRCh38
rs4380682 Chr.5: 177595651-177595651 on Build GRCh38
rs894771691 Chr.5: 177595616-177595616 on Build GRCh38
rs202086557 Chr.5: 177595329-177595329 on Build GRCh38
rs186881028 Chr.5: 177595247-177595247 on Build GRCh38
rs191767305 Chr.5: 177595650-177595650 on Build GRCh38
rs200110645 Chr.5: 177595460-177595460 on Build GRCh38
rs11545868 Chr.5: 177595427-177595427 on Build GRCh38
rs200591942 Chr.5: 177595317-177595317 on Build GRCh38
rs1054820378 Chr.5: 177595473-177595473 on Build GRCh38
rs758957758 Chr.5: 177595250-177595250 on Build GRCh38
rs1050901482 Chr.5: 177595214-177595216 on Build GRCh38
rs1018775085 Chr.5: 177595179-177595179 on Build GRCh38
rs139647055 Chr.5: 177595351-177595351 on Build GRCh38
rs771557662 Chr.5: 177595435-177595435 on Build GRCh38
rs762820519 Chr.5: 177595591-177595591 on Build GRCh38
rs924987872 Chr.5: 177595583-177595583 on Build GRCh38
rs367659530 Chr.5: 177595428-177595428 on Build GRCh38
rs145382488 Chr.5: 177595632-177595632 on Build GRCh38
rs569623224 Chr.5: 177595311-177595311 on Build GRCh38
rs375554888 Chr.5: 177595360-177595360 on Build GRCh38
rs539276221 Chr.5: 177595232-177595232 on Build GRCh38
rs752398480 Chr.5: 177595236-177595236 on Build GRCh38
rs903481351 Chr.5: 177595238-177595238 on Build GRCh38
rs372205411 Chr.5: 177595438-177595438 on Build GRCh38
rs776052110 Chr.5: 177595447-177595447 on Build GRCh38
rs773114554 Chr.5: 177595284-177595284 on Build GRCh38
rs182914993 Chr.5: 177595222-177595222 on Build GRCh38
rs145395803 Chr.5: 177595480-177595480 on Build GRCh38
rs1003585291 Chr.5: 177595611-177595611 on Build GRCh38
rs1013115778 Chr.5: 177595624-177595624 on Build GRCh38
rs1040110625 Chr.5: 177595439-177595439 on Build GRCh38

View Full Product Details