Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 88218478-88218980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102723885
Entrez Gene ID
102723885
Gene Name
uncharacterized LOC102723885
Gene Aliases
-
UniGene
Hs.489040
Gene Chromosome Location
Chr.7: 88216660-88229768 on Build GRCh38
Transcripts
NR_120517.1 NM_001256891.1 NM_001256892.1 NM_003130.3 NM_198901.1 XM_011516528.1

Gene Symbol
SRI
Entrez Gene ID
6717
Gene Name
sorcin
Gene Aliases
CP-22, CP22, SCN, V19
UniGene
Hs.489040
Gene Chromosome Location
Chr.7: 88205115-88226993 on Build GRCh38
Transcripts
NR_120517.1 NM_001256891.1 NM_001256892.1 NM_003130.3 NM_198901.1 XM_011516528.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3698579 c.66T>C p.G22G Chr.7: 88218883-88218883 on Build GRCh38
5438768 c.7-3C>T p.? Chr.7: 88218945-88218945 on Build GRCh38
6224992 c.13G>A p.G5R Chr.7: 88218936-88218936 on Build GRCh38
3698578 c.111T>C p.G37G Chr.7: 88218883-88218883 on Build GRCh38
5438767 c.52-3C>T p.? Chr.7: 88218945-88218945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997023560 Chr.7: 88218656-88218656 on Build GRCh38
rs898714046 Chr.7: 88218923-88218923 on Build GRCh38
rs761512527 Chr.7: 88218892-88218892 on Build GRCh38
rs764700340 Chr.7: 88218893-88218893 on Build GRCh38
rs769678991 Chr.7: 88218649-88218649 on Build GRCh38
rs955618863 Chr.7: 88218586-88218586 on Build GRCh38
rs747539890 Chr.7: 88218842-88218842 on Build GRCh38
rs941724647 Chr.7: 88218864-88218864 on Build GRCh38
rs557472577 Chr.7: 88218624-88218624 on Build GRCh38
rs983187556 Chr.7: 88218736-88218736 on Build GRCh38
rs755405864 Chr.7: 88218926-88218926 on Build GRCh38
rs569592532 Chr.7: 88218752-88218752 on Build GRCh38
rs925221798 Chr.7: 88218761-88218761 on Build GRCh38
rs763829519 Chr.7: 88218557-88218557 on Build GRCh38
rs573724652 Chr.7: 88218933-88218933 on Build GRCh38
rs776492181 Chr.7: 88218883-88218883 on Build GRCh38
rs755317859 Chr.7: 88218833-88218833 on Build GRCh38
rs749040752 Chr.7: 88218846-88218846 on Build GRCh38
rs974179160 Chr.7: 88218783-88218783 on Build GRCh38
rs772085816 Chr.7: 88218856-88218856 on Build GRCh38
rs201732383 Chr.7: 88218834-88218834 on Build GRCh38
rs749408370 Chr.7: 88218941-88218941 on Build GRCh38
rs148940168 Chr.7: 88218509-88218509 on Build GRCh38
rs777820078 Chr.7: 88218940-88218940 on Build GRCh38
rs766889555 Chr.7: 88218920-88218920 on Build GRCh38
rs1011353463 Chr.7: 88218547-88218547 on Build GRCh38
rs964072058 Chr.7: 88218745-88218745 on Build GRCh38
rs748299267 Chr.7: 88218835-88218835 on Build GRCh38
rs187125632 Chr.7: 88218666-88218666 on Build GRCh38
rs760560497 Chr.7: 88218868-88218868 on Build GRCh38
rs776543620 Chr.7: 88218953-88218953 on Build GRCh38
rs771076648 Chr.7: 88218944-88218944 on Build GRCh38
rs538845459 Chr.7: 88218716-88218716 on Build GRCh38
rs962890868 Chr.7: 88218528-88218528 on Build GRCh38
rs756913104 Chr.7: 88218657-88218657 on Build GRCh38
rs200699637 Chr.7: 88218910-88218910 on Build GRCh38
rs553261941 Chr.7: 88218542-88218542 on Build GRCh38
rs1001734382 Chr.7: 88218513-88218513 on Build GRCh38
rs747417833 Chr.7: 88218828-88218828 on Build GRCh38
rs1044827723 Chr.7: 88218829-88218829 on Build GRCh38
rs994772541 Chr.7: 88218573-88218573 on Build GRCh38
rs778469616 Chr.7: 88218705-88218705 on Build GRCh38
rs763914799 Chr.7: 88218869-88218869 on Build GRCh38
rs1017180582 Chr.7: 88218523-88218523 on Build GRCh38
rs201990001 Chr.7: 88218730-88218731 on Build GRCh38
rs43098 Chr.7: 88218922-88218922 on Build GRCh38
rs935276894 Chr.7: 88218796-88218796 on Build GRCh38
rs753501889 Chr.7: 88218651-88218651 on Build GRCh38
rs1025823777 Chr.7: 88218583-88218583 on Build GRCh38
rs769774502 Chr.7: 88218837-88218837 on Build GRCh38
rs773291344 Chr.7: 88218839-88218839 on Build GRCh38
rs758891459 Chr.7: 88218817-88218817 on Build GRCh38
rs746938789 Chr.7: 88218951-88218951 on Build GRCh38
rs775544968 Chr.7: 88218859-88218859 on Build GRCh38
rs377694328 Chr.7: 88218819-88218819 on Build GRCh38
rs781526976 Chr.7: 88218928-88218928 on Build GRCh38
rs1038574723 Chr.7: 88218914-88218914 on Build GRCh38
rs752914449 Chr.7: 88218929-88218929 on Build GRCh38
rs745822782 Chr.7: 88218843-88218843 on Build GRCh38
rs555337723 Chr.7: 88218912-88218912 on Build GRCh38
rs373435348 Chr.7: 88218808-88218808 on Build GRCh38
rs768629236 Chr.7: 88218952-88218952 on Build GRCh38
rs934629233 Chr.7: 88218874-88218874 on Build GRCh38
rs979339496 Chr.7: 88218751-88218751 on Build GRCh38
rs1029875090 Chr.7: 88218675-88218675 on Build GRCh38
rs780144309 Chr.7: 88218945-88218945 on Build GRCh38
rs1022399572 Chr.7: 88218551-88218551 on Build GRCh38
rs775255595 Chr.7: 88218772-88218772 on Build GRCh38

View Full Product Details