Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140733725-140734273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAF
Entrez Gene ID
673
Gene Name
B-Raf proto-oncogene, serine/threonine kinase
Gene Aliases
B-RAF1, B-raf, BRAF1, NS7, RAFB1
UniGene
Hs.550061
Gene Chromosome Location
Chr.7: 140719331-140924764 on Build GRCh38
Transcripts
NM_004333.4 XM_005250045.2 XM_017012558.1 XM_017012559.1

SNP Detail

SNP ID Chromosome Location
rs1041663521 Chr.7: 140733913-140733913 on Build GRCh38
rs34950866 Chr.7: 140733886-140733886 on Build GRCh38
rs544007477 Chr.7: 140733815-140733815 on Build GRCh38
rs760205305 Chr.7: 140733778-140733778 on Build GRCh38
rs114105685 Chr.7: 140734210-140734210 on Build GRCh38
rs1003159527 Chr.7: 140733922-140733922 on Build GRCh38
rs140628733 Chr.7: 140733834-140733834 on Build GRCh38
rs71645995 Chr.7: 140733771-140733771 on Build GRCh38
rs1006319747 Chr.7: 140733938-140733938 on Build GRCh38
rs910984172 Chr.7: 140733836-140733836 on Build GRCh38
rs964109573 Chr.7: 140734110-140734110 on Build GRCh38
rs57038951 Chr.7: 140733969-140733969 on Build GRCh38
rs192498662 Chr.7: 140734222-140734222 on Build GRCh38
rs892824127 Chr.7: 140733891-140733891 on Build GRCh38
rs574200772 Chr.7: 140733886-140733886 on Build GRCh38
rs1055866218 Chr.7: 140733929-140733929 on Build GRCh38
rs942365984 Chr.7: 140733885-140733886 on Build GRCh38
rs954880089 Chr.7: 140734038-140734038 on Build GRCh38
rs923404556 Chr.7: 140733813-140733813 on Build GRCh38
rs989917529 Chr.7: 140734111-140734113 on Build GRCh38
rs1030577644 Chr.7: 140733978-140733978 on Build GRCh38
rs559758711 Chr.7: 140733973-140733973 on Build GRCh38
rs962242356 Chr.7: 140733802-140733802 on Build GRCh38
rs1041638677 Chr.7: 140734207-140734207 on Build GRCh38
rs527278391 Chr.7: 140734073-140734073 on Build GRCh38
rs775361617 Chr.7: 140733777-140733777 on Build GRCh38
rs945906927 Chr.7: 140733906-140733906 on Build GRCh38
rs962023576 Chr.7: 140733972-140733972 on Build GRCh38
rs945723819 Chr.7: 140734197-140734197 on Build GRCh38
rs907027442 Chr.7: 140733921-140733921 on Build GRCh38
rs910732865 Chr.7: 140734095-140734095 on Build GRCh38
rs972309471 Chr.7: 140733805-140733805 on Build GRCh38
rs759364815 Chr.7: 140733754-140733754 on Build GRCh38
rs986562698 Chr.7: 140733825-140733825 on Build GRCh38
rs914212811 Chr.7: 140734127-140734127 on Build GRCh38
rs894581269 Chr.7: 140733930-140733930 on Build GRCh38
rs765958384 Chr.7: 140734080-140734085 on Build GRCh38
rs147952030 Chr.7: 140733766-140733766 on Build GRCh38
rs1016504509 Chr.7: 140733785-140733785 on Build GRCh38
rs1002925388 Chr.7: 140733748-140733748 on Build GRCh38

View Full Product Details