Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151565082-151565651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKAG2
Entrez Gene ID
51422
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 2
Gene Aliases
AAKG, AAKG2, CMH6, H91620p, WPWS
UniGene
Hs.647072
Gene Chromosome Location
Chr.7: 151556114-151877231 on Build GRCh38
Transcripts
NM_001040633.1 NM_001304527.1 NM_001304531.1 NM_016203.3 NM_024429.1 XM_005250002.3 XM_005250004.3 XM_005250006.4 XM_006716021.2 XM_011516282.1 XM_011516283.1 XM_011516284.1 XM_011516285.1 XM_011516286.1 XM_011516287.1 XM_017012268.1 XM_017012269.1 XM_017012270.1 XM_017012271.1 XM_017012272.1 XM_017012273.1 XM_017012274.1 XM_017012275.1 XM_017012276.1 XM_017012277.1 XM_017012278.1 XM_017012279.1 XM_017012280.1 XM_017012281.1 XM_017012282.1

SNP Detail

SNP ID Chromosome Location
rs915150288 Chr.7: 151565184-151565184 on Build GRCh38
rs1030978455 Chr.7: 151565448-151565448 on Build GRCh38
rs749730868 Chr.7: 151565330-151565330 on Build GRCh38
rs550467524 Chr.7: 151565480-151565480 on Build GRCh38
rs377381959 Chr.7: 151565239-151565239 on Build GRCh38
rs542618894 Chr.7: 151565109-151565109 on Build GRCh38
rs2302532 Chr.7: 151565219-151565219 on Build GRCh38
rs907977553 Chr.7: 151565257-151565257 on Build GRCh38
rs1010727893 Chr.7: 151565583-151565583 on Build GRCh38
rs994519288 Chr.7: 151565210-151565210 on Build GRCh38
rs1037856846 Chr.7: 151565285-151565285 on Build GRCh38
rs751001570 Chr.7: 151565319-151565319 on Build GRCh38
rs559321987 Chr.7: 151565117-151565117 on Build GRCh38
rs778668093 Chr.7: 151565351-151565351 on Build GRCh38
rs759412101 Chr.7: 151565427-151565427 on Build GRCh38
rs73475418 Chr.7: 151565590-151565590 on Build GRCh38
rs776542378 Chr.7: 151565423-151565423 on Build GRCh38
rs746531067 Chr.7: 151565185-151565185 on Build GRCh38
rs996865649 Chr.7: 151565305-151565305 on Build GRCh38
rs879171393 Chr.7: 151565575-151565575 on Build GRCh38
rs780581700 Chr.7: 151565329-151565329 on Build GRCh38
rs989327938 Chr.7: 151565155-151565155 on Build GRCh38
rs747734859 Chr.7: 151565400-151565400 on Build GRCh38
rs746960088 Chr.7: 151565413-151565413 on Build GRCh38
rs927840313 Chr.7: 151565577-151565577 on Build GRCh38
rs745558358 Chr.7: 151565605-151565605 on Build GRCh38
rs73156279 Chr.7: 151565299-151565299 on Build GRCh38
rs906432930 Chr.7: 151565162-151565162 on Build GRCh38
rs772641721 Chr.7: 151565408-151565408 on Build GRCh38
rs956505337 Chr.7: 151565149-151565149 on Build GRCh38
rs762004401 Chr.7: 151565631-151565631 on Build GRCh38
rs948194892 Chr.7: 151565565-151565565 on Build GRCh38
rs771644041 Chr.7: 151565404-151565404 on Build GRCh38
rs767652298 Chr.7: 151565307-151565307 on Build GRCh38
rs752058604 Chr.7: 151565497-151565497 on Build GRCh38
rs772778891 Chr.7: 151565114-151565114 on Build GRCh38
rs143529602 Chr.7: 151565207-151565207 on Build GRCh38
rs1035966245 Chr.7: 151565186-151565186 on Build GRCh38
rs888585470 Chr.7: 151565277-151565277 on Build GRCh38
rs567559152 Chr.7: 151565533-151565533 on Build GRCh38
rs781576711 Chr.7: 151565149-151565149 on Build GRCh38
rs940849601 Chr.7: 151565272-151565272 on Build GRCh38
rs770911441 Chr.7: 151565418-151565418 on Build GRCh38
rs939319865 Chr.7: 151565181-151565181 on Build GRCh38
rs751773986 Chr.7: 151565619-151565619 on Build GRCh38
rs956709653 Chr.7: 151565492-151565492 on Build GRCh38
rs770372030 Chr.7: 151565201-151565201 on Build GRCh38
rs763329900 Chr.7: 151565495-151565495 on Build GRCh38
rs764415557 Chr.7: 151565574-151565574 on Build GRCh38
rs1057119396 Chr.7: 151565232-151565232 on Build GRCh38
rs759483449 Chr.7: 151565296-151565296 on Build GRCh38
rs966011521 Chr.7: 151565429-151565429 on Build GRCh38
rs530752815 Chr.7: 151565442-151565442 on Build GRCh38
rs1030728886 Chr.7: 151565522-151565522 on Build GRCh38
rs756626353 Chr.7: 151565323-151565323 on Build GRCh38
rs755295667 Chr.7: 151565336-151565336 on Build GRCh38
rs939255313 Chr.7: 151565581-151565581 on Build GRCh38

View Full Product Details