Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140334573-140335143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC37A3
Entrez Gene ID
84255
Gene Name
solute carrier family 37 member 3
Gene Aliases
-
UniGene
Hs.446021
Gene Chromosome Location
Chr.7: 140333752-140404352 on Build GRCh38
Transcripts
NM_001287498.1 NM_032295.3 NM_207113.2 XM_011516626.2 XM_011516627.2 XM_017012712.1 XM_017012713.1 XM_017012714.1 XM_017012715.1 XM_017012716.1 XM_017012717.1

SNP Detail

SNP ID Chromosome Location
rs1139906 Chr.7: 140334942-140334942 on Build GRCh38
rs570724649 Chr.7: 140334651-140334651 on Build GRCh38
rs557745410 Chr.7: 140335065-140335065 on Build GRCh38
rs1004301765 Chr.7: 140334780-140334780 on Build GRCh38
rs777557567 Chr.7: 140334678-140334678 on Build GRCh38
rs945165891 Chr.7: 140334622-140334622 on Build GRCh38
rs957737123 Chr.7: 140334608-140334608 on Build GRCh38
rs1028554096 Chr.7: 140334833-140334833 on Build GRCh38
rs751415172 Chr.7: 140334610-140334610 on Build GRCh38
rs539425434 Chr.7: 140335051-140335051 on Build GRCh38
rs985835860 Chr.7: 140334862-140334862 on Build GRCh38
rs998409909 Chr.7: 140334786-140334786 on Build GRCh38
rs894462347 Chr.7: 140334634-140334634 on Build GRCh38
rs765307298 Chr.7: 140334609-140334609 on Build GRCh38
rs192560001 Chr.7: 140334628-140334628 on Build GRCh38
rs772608589 Chr.7: 140335030-140335030 on Build GRCh38
rs887166862 Chr.7: 140334700-140334700 on Build GRCh38
rs1015830706 Chr.7: 140334704-140334704 on Build GRCh38
rs920482543 Chr.7: 140335029-140335029 on Build GRCh38
rs115617434 Chr.7: 140334798-140334798 on Build GRCh38
rs1055086716 Chr.7: 140334633-140334633 on Build GRCh38
rs1048834660 Chr.7: 140335039-140335039 on Build GRCh38
rs1017811132 Chr.7: 140334882-140334882 on Build GRCh38
rs975644450 Chr.7: 140334605-140334605 on Build GRCh38
rs888553388 Chr.7: 140335037-140335037 on Build GRCh38
rs576288954 Chr.7: 140335107-140335107 on Build GRCh38
rs957686708 Chr.7: 140334839-140334839 on Build GRCh38
rs998357406 Chr.7: 140335118-140335118 on Build GRCh38
rs1006174483 Chr.7: 140334688-140334688 on Build GRCh38
rs538143489 Chr.7: 140334691-140334691 on Build GRCh38
rs927916985 Chr.7: 140335018-140335018 on Build GRCh38
rs575332179 Chr.7: 140334897-140334897 on Build GRCh38
rs114389520 Chr.7: 140334606-140334606 on Build GRCh38
rs966830561 Chr.7: 140334921-140334921 on Build GRCh38
rs1055225454 Chr.7: 140334639-140334639 on Build GRCh38
rs747947354 Chr.7: 140334632-140334632 on Build GRCh38
rs982029024 Chr.7: 140335000-140335000 on Build GRCh38
rs535937555 Chr.7: 140334924-140334924 on Build GRCh38
rs187776590 Chr.7: 140334627-140334627 on Build GRCh38
rs985844118 Chr.7: 140334598-140334598 on Build GRCh38
rs552172083 Chr.7: 140334650-140334650 on Build GRCh38
rs867650820 Chr.7: 140334804-140334804 on Build GRCh38
rs546016636 Chr.7: 140334904-140334904 on Build GRCh38
rs990724278 Chr.7: 140335028-140335028 on Build GRCh38
rs10244212 Chr.7: 140334822-140334822 on Build GRCh38
rs937918696 Chr.7: 140334631-140334631 on Build GRCh38
rs897322062 Chr.7: 140335106-140335106 on Build GRCh38
rs182373730 Chr.7: 140334975-140334975 on Build GRCh38
rs746308704 Chr.7: 140334997-140334997 on Build GRCh38
rs965974555 Chr.7: 140334600-140334600 on Build GRCh38
rs940092601 Chr.7: 140335056-140335056 on Build GRCh38
rs748958564 Chr.7: 140334797-140334797 on Build GRCh38
rs1048438702 Chr.7: 140334665-140334665 on Build GRCh38
rs961899980 Chr.7: 140334705-140334705 on Build GRCh38
rs989748364 Chr.7: 140334899-140334899 on Build GRCh38

View Full Product Details