Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 116770203-116770500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTP23
Entrez Gene ID
84294
Gene Name
UTP23, small subunit processome component
Gene Aliases
C8orf53
UniGene
Hs.86970
Gene Chromosome Location
Chr.8: 116766503-116774684 on Build GRCh38
Transcripts
NM_032334.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7188430 c.230A>G p.Y77C Chr.8: 116770233-116770233 on Build GRCh38
7189544 c.261A>C p.Q87H Chr.8: 116770264-116770264 on Build GRCh38
4542819 c.325G>A p.E109K Chr.8: 116770328-116770328 on Build GRCh38
219551 c.340C>T p.H114Y Chr.8: 116770343-116770343 on Build GRCh38
1313659 c.294G>C p.V98V Chr.8: 116770297-116770297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754570586 Chr.8: 116770268-116770268 on Build GRCh38
rs140131727 Chr.8: 116770229-116770229 on Build GRCh38
rs942275013 Chr.8: 116770272-116770272 on Build GRCh38
rs144867315 Chr.8: 116770356-116770356 on Build GRCh38
rs776513212 Chr.8: 116770307-116770307 on Build GRCh38
rs780709886 Chr.8: 116770230-116770231 on Build GRCh38
rs775320751 Chr.8: 116770293-116770294 on Build GRCh38
rs746976717 Chr.8: 116770391-116770391 on Build GRCh38
rs550076763 Chr.8: 116770420-116770420 on Build GRCh38
rs938611528 Chr.8: 116770401-116770401 on Build GRCh38
rs755689961 Chr.8: 116770249-116770249 on Build GRCh38
rs781008574 Chr.8: 116770403-116770403 on Build GRCh38
rs971882426 Chr.8: 116770393-116770393 on Build GRCh38
rs763459131 Chr.8: 116770320-116770320 on Build GRCh38
rs757074524 Chr.8: 116770394-116770394 on Build GRCh38
rs754748619 Chr.8: 116770362-116770362 on Build GRCh38
rs758179122 Chr.8: 116770289-116770289 on Build GRCh38
rs202052785 Chr.8: 116770237-116770237 on Build GRCh38
rs571606147 Chr.8: 116770375-116770375 on Build GRCh38
rs762179496 Chr.8: 116770348-116770348 on Build GRCh38
rs771063567 Chr.8: 116770302-116770302 on Build GRCh38
rs764522163 Chr.8: 116770330-116770330 on Build GRCh38
rs1037931732 Chr.8: 116770300-116770300 on Build GRCh38
rs895135496 Chr.8: 116770460-116770460 on Build GRCh38
rs763405924 Chr.8: 116770232-116770232 on Build GRCh38
rs777379421 Chr.8: 116770387-116770387 on Build GRCh38
rs531517164 Chr.8: 116770381-116770381 on Build GRCh38
rs976779568 Chr.8: 116770315-116770315 on Build GRCh38
rs574991380 Chr.8: 116770243-116770243 on Build GRCh38
rs746877101 Chr.8: 116770301-116770301 on Build GRCh38
rs764930472 Chr.8: 116770370-116770370 on Build GRCh38
rs561079115 Chr.8: 116770311-116770311 on Build GRCh38
rs774956333 Chr.8: 116770336-116770336 on Build GRCh38
rs754484396 Chr.8: 116770367-116770367 on Build GRCh38
rs200552231 Chr.8: 116770252-116770252 on Build GRCh38
rs752312450 Chr.8: 116770377-116770377 on Build GRCh38
rs770205942 Chr.8: 116770316-116770316 on Build GRCh38
rs769482274 Chr.8: 116770253-116770254 on Build GRCh38
rs762720478 Chr.8: 116770385-116770385 on Build GRCh38
rs371024889 Chr.8: 116770269-116770269 on Build GRCh38
rs948061046 Chr.8: 116770467-116770467 on Build GRCh38
rs1025054063 Chr.8: 116770421-116770421 on Build GRCh38
rs753552659 Chr.8: 116770264-116770264 on Build GRCh38
rs906330298 Chr.8: 116770441-116770442 on Build GRCh38
rs904004296 Chr.8: 116770234-116770234 on Build GRCh38
rs745473707 Chr.8: 116770252-116770252 on Build GRCh38
rs1056152218 Chr.8: 116770453-116770453 on Build GRCh38
rs150299132 Chr.8: 116770291-116770291 on Build GRCh38
rs1011941994 Chr.8: 116770371-116770371 on Build GRCh38

View Full Product Details