Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171701415-171701944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAMP4
Entrez Gene ID
8674
Gene Name
vesicle associated membrane protein 4
Gene Aliases
VAMP-4, VAMP24
UniGene
Hs.6651
Gene Chromosome Location
Chr.1: 171700156-171742239 on Build GRCh38
Transcripts
NM_001185127.1 NM_003762.4 NR_033704.1 XM_017002713.1

SNP Detail

SNP ID Chromosome Location
rs1016908401 Chr.1: 171701508-171701508 on Build GRCh38
rs958517251 Chr.1: 171701869-171701869 on Build GRCh38
rs183816201 Chr.1: 171701661-171701661 on Build GRCh38
rs780491799 Chr.1: 171701583-171701583 on Build GRCh38
rs755159279 Chr.1: 171701643-171701643 on Build GRCh38
rs187459798 Chr.1: 171701760-171701760 on Build GRCh38
rs879580437 Chr.1: 171701623-171701623 on Build GRCh38
rs551627924 Chr.1: 171701630-171701630 on Build GRCh38
rs928439591 Chr.1: 171701803-171701803 on Build GRCh38
rs1056583706 Chr.1: 171701839-171701839 on Build GRCh38
rs1012466520 Chr.1: 171701920-171701920 on Build GRCh38
rs371800544 Chr.1: 171701457-171701457 on Build GRCh38
rs758679561 Chr.1: 171701547-171701547 on Build GRCh38
rs945995095 Chr.1: 171701529-171701529 on Build GRCh38
rs140463734 Chr.1: 171701459-171701459 on Build GRCh38
rs188148720 Chr.1: 171701510-171701510 on Build GRCh38
rs1041856564 Chr.1: 171701773-171701773 on Build GRCh38
rs952114742 Chr.1: 171701794-171701794 on Build GRCh38
rs10913514 Chr.1: 171701683-171701683 on Build GRCh38
rs111392683 Chr.1: 171701439-171701439 on Build GRCh38
rs945779943 Chr.1: 171701723-171701723 on Build GRCh38
rs553236038 Chr.1: 171701690-171701690 on Build GRCh38
rs149955119 Chr.1: 171701717-171701717 on Build GRCh38
rs1009092707 Chr.1: 171701646-171701646 on Build GRCh38
rs895378428 Chr.1: 171701909-171701909 on Build GRCh38
rs528903492 Chr.1: 171701467-171701467 on Build GRCh38
rs935702282 Chr.1: 171701620-171701620 on Build GRCh38
rs192937368 Chr.1: 171701511-171701511 on Build GRCh38
rs1033751470 Chr.1: 171701829-171701829 on Build GRCh38
rs535884503 Chr.1: 171701737-171701737 on Build GRCh38
rs904189840 Chr.1: 171701557-171701557 on Build GRCh38
rs939510772 Chr.1: 171701821-171701821 on Build GRCh38
rs980822476 Chr.1: 171701801-171701801 on Build GRCh38
rs781156781 Chr.1: 171701653-171701653 on Build GRCh38
rs917180283 Chr.1: 171701527-171701527 on Build GRCh38
rs765570198 Chr.1: 171701463-171701463 on Build GRCh38
rs937217349 Chr.1: 171701458-171701458 on Build GRCh38
rs116788793 Chr.1: 171701539-171701539 on Build GRCh38
rs191947936 Chr.1: 171701799-171701799 on Build GRCh38

View Full Product Details