Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 181052040-181052638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MR1
Entrez Gene ID
3140
Gene Name
major histocompatibility complex, class I-related
Gene Aliases
HLALS
UniGene
Hs.13500
Gene Chromosome Location
Chr.1: 181033379-181061938 on Build GRCh38
Transcripts
NM_001194999.1 NM_001195000.1 NM_001195035.1 NM_001310213.1 NM_001531.2 XM_006711286.3 XM_011509463.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5604112 c.650T>C p.V217A Chr.1: 181052280-181052280 on Build GRCh38
5567858 c.765T>C p.S255S Chr.1: 181052395-181052395 on Build GRCh38
1257915 c.716A>C p.N239T Chr.1: 181052346-181052346 on Build GRCh38
3863757 c.865C>T p.L289F Chr.1: 181052495-181052495 on Build GRCh38
5514804 c.759T>A p.L253L Chr.1: 181052389-181052389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748528955 Chr.1: 181052378-181052378 on Build GRCh38
rs749329475 Chr.1: 181052087-181052087 on Build GRCh38
rs146370833 Chr.1: 181052252-181052252 on Build GRCh38
rs768216742 Chr.1: 181052503-181052503 on Build GRCh38
rs765479153 Chr.1: 181052546-181052546 on Build GRCh38
rs778320951 Chr.1: 181052477-181052477 on Build GRCh38
rs779470788 Chr.1: 181052250-181052250 on Build GRCh38
rs749876488 Chr.1: 181052243-181052243 on Build GRCh38
rs371195224 Chr.1: 181052494-181052494 on Build GRCh38
rs980573937 Chr.1: 181052501-181052501 on Build GRCh38
rs139730390 Chr.1: 181052304-181052304 on Build GRCh38
rs944036419 Chr.1: 181052566-181052566 on Build GRCh38
rs767674160 Chr.1: 181052231-181052231 on Build GRCh38
rs757516171 Chr.1: 181052228-181052228 on Build GRCh38
rs372662063 Chr.1: 181052364-181052364 on Build GRCh38
rs571824550 Chr.1: 181052440-181052440 on Build GRCh38
rs376351945 Chr.1: 181052518-181052518 on Build GRCh38
rs764014489 Chr.1: 181052220-181052220 on Build GRCh38
rs771923481 Chr.1: 181052431-181052431 on Build GRCh38
rs779529132 Chr.1: 181052480-181052480 on Build GRCh38
rs919767038 Chr.1: 181052504-181052504 on Build GRCh38
rs753842424 Chr.1: 181052349-181052349 on Build GRCh38
rs567682945 Chr.1: 181052274-181052274 on Build GRCh38
rs758777817 Chr.1: 181052394-181052394 on Build GRCh38
rs753983560 Chr.1: 181052449-181052449 on Build GRCh38
rs917773804 Chr.1: 181052078-181052078 on Build GRCh38
rs763601534 Chr.1: 181052555-181052555 on Build GRCh38
rs762784237 Chr.1: 181052215-181052215 on Build GRCh38
rs947909614 Chr.1: 181052082-181052082 on Build GRCh38
rs186208207 Chr.1: 181052184-181052184 on Build GRCh38
rs765852229 Chr.1: 181052453-181052453 on Build GRCh38
rs200517429 Chr.1: 181052223-181052223 on Build GRCh38
rs751845229 Chr.1: 181052479-181052479 on Build GRCh38
rs773809410 Chr.1: 181052154-181052154 on Build GRCh38
rs149433107 Chr.1: 181052237-181052237 on Build GRCh38
rs141620086 Chr.1: 181052336-181052336 on Build GRCh38
rs906173254 Chr.1: 181052592-181052592 on Build GRCh38
rs750706337 Chr.1: 181052339-181052339 on Build GRCh38
rs762984615 Chr.1: 181052294-181052294 on Build GRCh38
rs373246951 Chr.1: 181052232-181052232 on Build GRCh38
rs1021764970 Chr.1: 181052319-181052319 on Build GRCh38
rs1023414230 Chr.1: 181052084-181052084 on Build GRCh38
rs551334869 Chr.1: 181052578-181052578 on Build GRCh38
rs765089402 Chr.1: 181052201-181052201 on Build GRCh38
rs775111855 Chr.1: 181052409-181052409 on Build GRCh38
rs931257373 Chr.1: 181052509-181052509 on Build GRCh38
rs893493909 Chr.1: 181052064-181052064 on Build GRCh38
rs760348743 Chr.1: 181052523-181052523 on Build GRCh38
rs908557724 Chr.1: 181052563-181052563 on Build GRCh38
rs767743878 Chr.1: 181052316-181052316 on Build GRCh38
rs540886985 Chr.1: 181052131-181052131 on Build GRCh38
rs141366222 Chr.1: 181052401-181052401 on Build GRCh38
rs761551215 Chr.1: 181052314-181052314 on Build GRCh38
rs776659456 Chr.1: 181052534-181052534 on Build GRCh38
rs112877390 Chr.1: 181052490-181052490 on Build GRCh38
rs186300848 Chr.1: 181052426-181052426 on Build GRCh38
rs758968727 Chr.1: 181052464-181052464 on Build GRCh38
rs755410502 Chr.1: 181052248-181052248 on Build GRCh38
rs377066166 Chr.1: 181052446-181052446 on Build GRCh38
rs775439388 Chr.1: 181052210-181052210 on Build GRCh38
rs370861189 Chr.1: 181052076-181052076 on Build GRCh38
rs759467635 Chr.1: 181052452-181052452 on Build GRCh38
rs573659546 Chr.1: 181052115-181052115 on Build GRCh38
rs763993652 Chr.1: 181052300-181052300 on Build GRCh38
rs376525686 Chr.1: 181052254-181052254 on Build GRCh38
rs762295481 Chr.1: 181052341-181052341 on Build GRCh38
rs919850565 Chr.1: 181052126-181052126 on Build GRCh38
rs528551338 Chr.1: 181052416-181052416 on Build GRCh38
rs916381949 Chr.1: 181052522-181052522 on Build GRCh38
rs200078718 Chr.1: 181052519-181052519 on Build GRCh38
rs756165030 Chr.1: 181052234-181052234 on Build GRCh38
rs769402916 Chr.1: 181052262-181052262 on Build GRCh38
rs139617770 Chr.1: 181052351-181052351 on Build GRCh38
rs778394883 Chr.1: 181052511-181052511 on Build GRCh38
rs1044849061 Chr.1: 181052083-181052083 on Build GRCh38
rs35223984 Chr.1: 181052347-181052347 on Build GRCh38
rs1012299528 Chr.1: 181052069-181052069 on Build GRCh38
rs142428410 Chr.1: 181052258-181052258 on Build GRCh38
rs956380752 Chr.1: 181052348-181052348 on Build GRCh38
rs776303667 Chr.1: 181052195-181052195 on Build GRCh38
rs569554944 Chr.1: 181052613-181052613 on Build GRCh38
rs376550039 Chr.1: 181052549-181052549 on Build GRCh38
rs774856707 Chr.1: 181052265-181052265 on Build GRCh38
rs761007984 Chr.1: 181052445-181052445 on Build GRCh38
rs759819183 Chr.1: 181052535-181052535 on Build GRCh38
rs757142331 Chr.1: 181052466-181052467 on Build GRCh38
rs748694184 Chr.1: 181052415-181052415 on Build GRCh38
rs1047545593 Chr.1: 181052171-181052171 on Build GRCh38
rs764455752 Chr.1: 181052558-181052558 on Build GRCh38
rs199621631 Chr.1: 181052200-181052200 on Build GRCh38
rs1044702861 Chr.1: 181052521-181052521 on Build GRCh38
rs771190437 Chr.1: 181052526-181052526 on Build GRCh38
rs769297317 Chr.1: 181052406-181052406 on Build GRCh38
rs753312375 Chr.1: 181052460-181052460 on Build GRCh38

View Full Product Details