Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43397737-43398285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC17
Entrez Gene ID
55761
Gene Name
tetratricopeptide repeat domain 17
Gene Aliases
-
UniGene
Hs.191186
Gene Chromosome Location
Chr.11: 43358885-43494933 on Build GRCh38
Transcripts
NM_001307943.1 NM_018259.5 XM_005253016.1 XM_006718263.1 XM_011520218.2 XM_017018010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3943665 c.1045G>T p.E349* Chr.11: 43398100-43398100 on Build GRCh38
3943664 c.1045G>T p.E349* Chr.11: 43398100-43398100 on Build GRCh38
7311822 c.1048G>T p.A350S Chr.11: 43398103-43398103 on Build GRCh38
6414405 c.967C>T p.Q323* Chr.11: 43398022-43398022 on Build GRCh38
6414404 c.967C>T p.Q323* Chr.11: 43398022-43398022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530317322 Chr.11: 43397877-43397877 on Build GRCh38
rs756783802 Chr.11: 43397936-43397937 on Build GRCh38
rs754371478 Chr.11: 43397941-43397952 on Build GRCh38
rs748397835 Chr.11: 43397945-43397952 on Build GRCh38
rs878985274 Chr.11: 43397901-43397902 on Build GRCh38
rs775208559 Chr.11: 43397931-43397952 on Build GRCh38
rs770968516 Chr.11: 43397949-43397952 on Build GRCh38
rs148662112 Chr.11: 43397832-43397832 on Build GRCh38
rs1000766386 Chr.11: 43398140-43398140 on Build GRCh38
rs763979989 Chr.11: 43397952-43397952 on Build GRCh38
rs147854498 Chr.11: 43397992-43397992 on Build GRCh38
rs761605805 Chr.11: 43397952-43397953 on Build GRCh38
rs55701432 Chr.11: 43398113-43398113 on Build GRCh38
rs776082923 Chr.11: 43398049-43398049 on Build GRCh38
rs779419834 Chr.11: 43398015-43398015 on Build GRCh38
rs184621422 Chr.11: 43397873-43397873 on Build GRCh38
rs776124691 Chr.11: 43398172-43398172 on Build GRCh38
rs139431536 Chr.11: 43398041-43398041 on Build GRCh38
rs547034349 Chr.11: 43397902-43397907 on Build GRCh38
rs755598182 Chr.11: 43397940-43397941 on Build GRCh38
rs759467255 Chr.11: 43398127-43398127 on Build GRCh38
rs3961644 Chr.11: 43397942-43397942 on Build GRCh38
rs762961767 Chr.11: 43397985-43397985 on Build GRCh38
rs964016323 Chr.11: 43397883-43397883 on Build GRCh38
rs994004483 Chr.11: 43397852-43397852 on Build GRCh38
rs952661993 Chr.11: 43398216-43398216 on Build GRCh38
rs758774070 Chr.11: 43398023-43398023 on Build GRCh38
rs1025923725 Chr.11: 43397855-43397855 on Build GRCh38
rs368884855 Chr.11: 43398145-43398145 on Build GRCh38
rs994511306 Chr.11: 43398157-43398157 on Build GRCh38
rs774112706 Chr.11: 43397953-43397956 on Build GRCh38
rs956645836 Chr.11: 43398221-43398221 on Build GRCh38
rs1048476886 Chr.11: 43397950-43397950 on Build GRCh38
rs750649968 Chr.11: 43398161-43398161 on Build GRCh38
rs570099225 Chr.11: 43398227-43398227 on Build GRCh38
rs772928831 Chr.11: 43397964-43397964 on Build GRCh38
rs765608987 Chr.11: 43398159-43398159 on Build GRCh38
rs202107663 Chr.11: 43398086-43398086 on Build GRCh38
rs764469976 Chr.11: 43397952-43397952 on Build GRCh38
rs536732292 Chr.11: 43397829-43397829 on Build GRCh38
rs747530431 Chr.11: 43398036-43398036 on Build GRCh38
rs557328474 Chr.11: 43397871-43397871 on Build GRCh38
rs779333391 Chr.11: 43397943-43397952 on Build GRCh38
rs776742484 Chr.11: 43397948-43397949 on Build GRCh38
rs767120941 Chr.11: 43397955-43397956 on Build GRCh38
rs761409343 Chr.11: 43397960-43397960 on Build GRCh38
rs764246036 Chr.11: 43398003-43398003 on Build GRCh38
rs975276854 Chr.11: 43397889-43397889 on Build GRCh38
rs768205327 Chr.11: 43397958-43397958 on Build GRCh38
rs762810474 Chr.11: 43397932-43397932 on Build GRCh38
rs887188242 Chr.11: 43397953-43397953 on Build GRCh38
rs144941992 Chr.11: 43398103-43398103 on Build GRCh38
rs1018311527 Chr.11: 43398239-43398239 on Build GRCh38
rs968852199 Chr.11: 43397763-43397763 on Build GRCh38
rs894823534 Chr.11: 43397823-43397823 on Build GRCh38
rs548450224 Chr.11: 43397935-43397935 on Build GRCh38
rs943938940 Chr.11: 43397881-43397881 on Build GRCh38
rs1019716260 Chr.11: 43398254-43398254 on Build GRCh38
rs757514502 Chr.11: 43398009-43398009 on Build GRCh38
rs775509114 Chr.11: 43397951-43397952 on Build GRCh38
rs1024897172 Chr.11: 43397842-43397842 on Build GRCh38
rs750019123 Chr.11: 43397960-43397961 on Build GRCh38
rs543607649 Chr.11: 43397789-43397790 on Build GRCh38
rs931115277 Chr.11: 43397946-43397946 on Build GRCh38
rs979182475 Chr.11: 43397795-43397795 on Build GRCh38
rs1057438949 Chr.11: 43397965-43397965 on Build GRCh38
rs769379960 Chr.11: 43398061-43398061 on Build GRCh38
rs1011913342 Chr.11: 43398110-43398110 on Build GRCh38
rs758807077 Chr.11: 43397944-43397945 on Build GRCh38
rs765051727 Chr.11: 43397962-43397962 on Build GRCh38
rs764299323 Chr.11: 43398128-43398128 on Build GRCh38
rs1025447327 Chr.11: 43398179-43398179 on Build GRCh38
rs762606190 Chr.11: 43397951-43397956 on Build GRCh38
rs747262639 Chr.11: 43397948-43397948 on Build GRCh38
rs766286299 Chr.11: 43397989-43397989 on Build GRCh38
rs200432170 Chr.11: 43398016-43398016 on Build GRCh38
rs769549261 Chr.11: 43397950-43397954 on Build GRCh38
rs772906929 Chr.11: 43398063-43398063 on Build GRCh38
rs776316875 Chr.11: 43397959-43397959 on Build GRCh38
rs201210580 Chr.11: 43397948-43397948 on Build GRCh38
rs761563664 Chr.11: 43398181-43398181 on Build GRCh38
rs774202026 Chr.11: 43398122-43398122 on Build GRCh38
rs971087302 Chr.11: 43397845-43397845 on Build GRCh38
rs754134391 Chr.11: 43398004-43398004 on Build GRCh38
rs767172080 Chr.11: 43397939-43397952 on Build GRCh38
rs761211858 Chr.11: 43397902-43397902 on Build GRCh38
rs745793845 Chr.11: 43397950-43397950 on Build GRCh38
rs777813956 Chr.11: 43397947-43397952 on Build GRCh38
rs763594194 Chr.11: 43397934-43397938 on Build GRCh38
rs946317864 Chr.11: 43397966-43397966 on Build GRCh38
rs367955826 Chr.11: 43397956-43397956 on Build GRCh38
rs372213741 Chr.11: 43398155-43398155 on Build GRCh38
rs751578243 Chr.11: 43397908-43397929 on Build GRCh38
rs368934799 Chr.11: 43397902-43397923 on Build GRCh38
rs765060952 Chr.11: 43397928-43397929 on Build GRCh38
rs952804372 Chr.11: 43397862-43397862 on Build GRCh38
rs756190098 Chr.11: 43397998-43397998 on Build GRCh38
rs533625749 Chr.11: 43398072-43398072 on Build GRCh38
rs762135168 Chr.11: 43398154-43398154 on Build GRCh38
rs780490224 Chr.11: 43398029-43398029 on Build GRCh38
rs573527196 Chr.11: 43397957-43397957 on Build GRCh38
rs751247046 Chr.11: 43397937-43397952 on Build GRCh38

View Full Product Details