Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 87302004-87302601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM135
Entrez Gene ID
65084
Gene Name
transmembrane protein 135
Gene Aliases
PMP52
UniGene
Hs.188591
Gene Chromosome Location
Chr.11: 87037844-87328834 on Build GRCh38
Transcripts
NM_001168724.1 NM_022918.3 NR_033149.1 XM_017018140.1 XM_017018141.1 XM_017018142.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
932780 c.557T>G p.I186S Chr.11: 87302301-87302301 on Build GRCh38
5617660 c.583C>T p.H195Y Chr.11: 87302327-87302327 on Build GRCh38
4758471 c.648A>C p.K216N Chr.11: 87302392-87302392 on Build GRCh38
6352943 c.652G>A p.G218R Chr.11: 87302396-87302396 on Build GRCh38
2221927 c.670G>T p.G224C Chr.11: 87302414-87302414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951588755 Chr.11: 87302058-87302058 on Build GRCh38
rs761525630 Chr.11: 87302516-87302516 on Build GRCh38
rs751537658 Chr.11: 87302350-87302350 on Build GRCh38
rs955174356 Chr.11: 87302462-87302462 on Build GRCh38
rs539603820 Chr.11: 87302554-87302554 on Build GRCh38
rs984399147 Chr.11: 87302084-87302084 on Build GRCh38
rs558650816 Chr.11: 87302075-87302075 on Build GRCh38
rs763507950 Chr.11: 87302277-87302278 on Build GRCh38
rs370253736 Chr.11: 87302355-87302355 on Build GRCh38
rs988214289 Chr.11: 87302572-87302572 on Build GRCh38
rs752790111 Chr.11: 87302345-87302345 on Build GRCh38
rs765349069 Chr.11: 87302342-87302342 on Build GRCh38
rs151122474 Chr.11: 87302414-87302414 on Build GRCh38
rs754403580 Chr.11: 87302491-87302491 on Build GRCh38
rs17854687 Chr.11: 87302322-87302322 on Build GRCh38
rs920603572 Chr.11: 87302539-87302539 on Build GRCh38
rs35073361 Chr.11: 87302351-87302351 on Build GRCh38
rs762893120 Chr.11: 87302422-87302422 on Build GRCh38
rs748788236 Chr.11: 87302378-87302378 on Build GRCh38
rs2276103 Chr.11: 87302210-87302210 on Build GRCh38
rs147367412 Chr.11: 87302324-87302324 on Build GRCh38
rs554412457 Chr.11: 87302484-87302485 on Build GRCh38
rs1030639709 Chr.11: 87302209-87302209 on Build GRCh38
rs1003758890 Chr.11: 87302150-87302150 on Build GRCh38
rs761072696 Chr.11: 87302320-87302320 on Build GRCh38
rs761199433 Chr.11: 87302395-87302395 on Build GRCh38
rs867224854 Chr.11: 87302503-87302503 on Build GRCh38
rs187748366 Chr.11: 87302245-87302245 on Build GRCh38
rs368359899 Chr.11: 87302456-87302456 on Build GRCh38
rs377164290 Chr.11: 87302379-87302379 on Build GRCh38
rs183238777 Chr.11: 87302577-87302577 on Build GRCh38
rs148620662 Chr.11: 87302375-87302375 on Build GRCh38
rs372431948 Chr.11: 87302488-87302488 on Build GRCh38
rs901588963 Chr.11: 87302431-87302431 on Build GRCh38
rs745548250 Chr.11: 87302277-87302277 on Build GRCh38
rs745458620 Chr.11: 87302352-87302352 on Build GRCh38
rs557342217 Chr.11: 87302451-87302451 on Build GRCh38
rs969928709 Chr.11: 87302532-87302532 on Build GRCh38
rs572216494 Chr.11: 87302576-87302576 on Build GRCh38
rs948141930 Chr.11: 87302357-87302357 on Build GRCh38
rs779621107 Chr.11: 87302460-87302460 on Build GRCh38
rs906754840 Chr.11: 87302133-87302133 on Build GRCh38
rs531717739 Chr.11: 87302310-87302310 on Build GRCh38
rs757275929 Chr.11: 87302441-87302441 on Build GRCh38
rs981226901 Chr.11: 87302241-87302241 on Build GRCh38
rs759770678 Chr.11: 87302331-87302331 on Build GRCh38
rs766941477 Chr.11: 87302471-87302471 on Build GRCh38
rs775099041 Chr.11: 87302247-87302247 on Build GRCh38
rs558622871 Chr.11: 87302570-87302570 on Build GRCh38
rs143809398 Chr.11: 87302440-87302440 on Build GRCh38
rs1010596620 Chr.11: 87302216-87302216 on Build GRCh38
rs772521617 Chr.11: 87302305-87302305 on Build GRCh38
rs752802886 Chr.11: 87302417-87302417 on Build GRCh38
rs2276102 Chr.11: 87302396-87302396 on Build GRCh38
rs377223796 Chr.11: 87302231-87302232 on Build GRCh38
rs928162352 Chr.11: 87302555-87302555 on Build GRCh38
rs139546975 Chr.11: 87302347-87302347 on Build GRCh38
rs760210448 Chr.11: 87302490-87302492 on Build GRCh38
rs149756646 Chr.11: 87302097-87302097 on Build GRCh38
rs563524442 Chr.11: 87302035-87302035 on Build GRCh38
rs767591408 Chr.11: 87302274-87302274 on Build GRCh38
rs973773158 Chr.11: 87302538-87302538 on Build GRCh38
rs377011738 Chr.11: 87302278-87302278 on Build GRCh38
rs773539460 Chr.11: 87302393-87302393 on Build GRCh38
rs778488082 Chr.11: 87302493-87302493 on Build GRCh38
rs143859974 Chr.11: 87302354-87302354 on Build GRCh38
rs906726263 Chr.11: 87302369-87302369 on Build GRCh38
rs928080343 Chr.11: 87302246-87302246 on Build GRCh38
rs779612083 Chr.11: 87302360-87302360 on Build GRCh38
rs565790919 Chr.11: 87302220-87302220 on Build GRCh38
rs141325445 Chr.11: 87302430-87302430 on Build GRCh38
rs190673002 Chr.11: 87302293-87302293 on Build GRCh38
rs375568783 Chr.11: 87302404-87302404 on Build GRCh38
rs1022453869 Chr.11: 87302232-87302232 on Build GRCh38
rs1045176242 Chr.11: 87302362-87302362 on Build GRCh38
rs148815932 Chr.11: 87302512-87302512 on Build GRCh38
rs777621737 Chr.11: 87302349-87302349 on Build GRCh38

View Full Product Details