Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10125219-10125691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC7A
Entrez Gene ID
64581
Gene Name
C-type lectin domain family 7 member A
Gene Aliases
BGR, CANDF4, CD369, CLECSF12, DECTIN1, SCARE2
UniGene
Hs.143929
Gene Chromosome Location
Chr.12: 10116777-10130269 on Build GRCh38
Transcripts
NM_022570.4 NM_197947.2 NM_197948.2 NM_197949.2 NM_197950.2 NR_125336.1 XM_006719135.3 XM_017019822.1 XM_017019823.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3398257 c.466C>G p.L156V Chr.12: 10125323-10125323 on Build GRCh38
3455181 c.281C>T p.S94F Chr.12: 10125370-10125370 on Build GRCh38
5929383 c.350C>T p.S117F Chr.12: 10125439-10125439 on Build GRCh38
6630249 c.309G>A p.W103* Chr.12: 10125342-10125342 on Build GRCh38
3455180 c.419C>T p.S140F Chr.12: 10125370-10125370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750260600 Chr.12: 10125279-10125279 on Build GRCh38
rs760264788 Chr.12: 10125468-10125468 on Build GRCh38
rs369873648 Chr.12: 10125262-10125262 on Build GRCh38
rs868279605 Chr.12: 10125243-10125243 on Build GRCh38
rs1006238620 Chr.12: 10125491-10125491 on Build GRCh38
rs760554156 Chr.12: 10125388-10125388 on Build GRCh38
rs893962568 Chr.12: 10125438-10125438 on Build GRCh38
rs572174343 Chr.12: 10125520-10125520 on Build GRCh38
rs565247139 Chr.12: 10125471-10125471 on Build GRCh38
rs374225257 Chr.12: 10125466-10125466 on Build GRCh38
rs112313622 Chr.12: 10125590-10125590 on Build GRCh38
rs937799340 Chr.12: 10125649-10125649 on Build GRCh38
rs756466830 Chr.12: 10125353-10125353 on Build GRCh38
rs780102364 Chr.12: 10125290-10125290 on Build GRCh38
rs761503556 Chr.12: 10125368-10125368 on Build GRCh38
rs756111334 Chr.12: 10125285-10125285 on Build GRCh38
rs747442135 Chr.12: 10125326-10125326 on Build GRCh38
rs773988936 Chr.12: 10125450-10125450 on Build GRCh38
rs751209361 Chr.12: 10125257-10125257 on Build GRCh38
rs903629711 Chr.12: 10125379-10125379 on Build GRCh38
rs758623997 Chr.12: 10125313-10125313 on Build GRCh38
rs143893294 Chr.12: 10125375-10125375 on Build GRCh38
rs1030954787 Chr.12: 10125563-10125563 on Build GRCh38
rs191029355 Chr.12: 10125564-10125564 on Build GRCh38
rs59819090 Chr.12: 10125307-10125307 on Build GRCh38
rs950763900 Chr.12: 10125246-10125246 on Build GRCh38
rs913549289 Chr.12: 10125592-10125592 on Build GRCh38
rs996349363 Chr.12: 10125543-10125543 on Build GRCh38
rs753828241 Chr.12: 10125297-10125297 on Build GRCh38
rs746386372 Chr.12: 10125347-10125347 on Build GRCh38
rs979123396 Chr.12: 10125607-10125607 on Build GRCh38
rs140932713 Chr.12: 10125420-10125420 on Build GRCh38
rs1004283102 Chr.12: 10125625-10125625 on Build GRCh38
rs546625102 Chr.12: 10125594-10125594 on Build GRCh38
rs771816303 Chr.12: 10125461-10125461 on Build GRCh38
rs755131422 Chr.12: 10125299-10125299 on Build GRCh38
rs147313614 Chr.12: 10125413-10125413 on Build GRCh38
rs757594070 Chr.12: 10125433-10125433 on Build GRCh38
rs761591534 Chr.12: 10125261-10125261 on Build GRCh38
rs564932655 Chr.12: 10125654-10125654 on Build GRCh38
rs375321656 Chr.12: 10125514-10125514 on Build GRCh38
rs776292856 Chr.12: 10125488-10125488 on Build GRCh38
rs35779431 Chr.12: 10125587-10125588 on Build GRCh38
rs963235239 Chr.12: 10125662-10125662 on Build GRCh38
rs758605469 Chr.12: 10125405-10125405 on Build GRCh38
rs148482816 Chr.12: 10125529-10125529 on Build GRCh38
rs763633038 Chr.12: 10125249-10125249 on Build GRCh38
rs142662684 Chr.12: 10125665-10125665 on Build GRCh38
rs752770917 Chr.12: 10125294-10125295 on Build GRCh38
rs747731438 Chr.12: 10125458-10125458 on Build GRCh38
rs1028827867 Chr.12: 10125666-10125666 on Build GRCh38
rs1013923644 Chr.12: 10125431-10125431 on Build GRCh38
rs138619592 Chr.12: 10125382-10125382 on Build GRCh38
rs369482852 Chr.12: 10125367-10125367 on Build GRCh38
rs933600857 Chr.12: 10125324-10125324 on Build GRCh38
rs765292086 Chr.12: 10125402-10125402 on Build GRCh38
rs752786951 Chr.12: 10125404-10125404 on Build GRCh38
rs781443517 Chr.12: 10125435-10125435 on Build GRCh38
rs768174843 Chr.12: 10125353-10125353 on Build GRCh38
rs747567228 Chr.12: 10125614-10125614 on Build GRCh38
rs748327912 Chr.12: 10125310-10125310 on Build GRCh38
rs149477976 Chr.12: 10125333-10125333 on Build GRCh38
rs766202479 Chr.12: 10125390-10125390 on Build GRCh38
rs964793762 Chr.12: 10125515-10125515 on Build GRCh38
rs570688620 Chr.12: 10125267-10125267 on Build GRCh38
rs954942374 Chr.12: 10125565-10125565 on Build GRCh38
rs762564803 Chr.12: 10125242-10125242 on Build GRCh38
rs144190589 Chr.12: 10125392-10125392 on Build GRCh38
rs753757159 Chr.12: 10125391-10125391 on Build GRCh38
rs778139261 Chr.12: 10125467-10125467 on Build GRCh38
rs112345533 Chr.12: 10125439-10125439 on Build GRCh38
rs777247198 Chr.12: 10125339-10125339 on Build GRCh38
rs966350955 Chr.12: 10125605-10125605 on Build GRCh38
rs913570758 Chr.12: 10125345-10125345 on Build GRCh38
rs1017059662 Chr.12: 10125641-10125641 on Build GRCh38
rs964865505 Chr.12: 10125293-10125293 on Build GRCh38
rs770219030 Chr.12: 10125447-10125447 on Build GRCh38
rs745468969 Chr.12: 10125291-10125291 on Build GRCh38
rs145439653 Chr.12: 10125524-10125524 on Build GRCh38
rs746322311 Chr.12: 10125442-10125442 on Build GRCh38
rs778011232 Chr.12: 10125408-10125408 on Build GRCh38
rs138005591 Chr.12: 10125316-10125316 on Build GRCh38

View Full Product Details