Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128460686-128461284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETS1
Entrez Gene ID
2113
Gene Name
ETS proto-oncogene 1, transcription factor
Gene Aliases
ETS-1, EWSR2, c-ets-1, p54
UniGene
Hs.369438
Gene Chromosome Location
Chr.11: 128458761-128587593 on Build GRCh38
Transcripts
NM_001143820.1 NM_001162422.1 NM_005238.3 XM_011542649.2 XM_011542650.1 XM_011542651.1 XM_011542652.1 XM_017017314.1 XM_017017315.1 XM_017017316.1 XM_017017317.1

SNP Detail

SNP ID Chromosome Location
rs551609993 Chr.11: 128460985-128460985 on Build GRCh38
rs745426110 Chr.11: 128461260-128461260 on Build GRCh38
rs777450296 Chr.11: 128460993-128460993 on Build GRCh38
rs939482555 Chr.11: 128460921-128460921 on Build GRCh38
rs907492955 Chr.11: 128461091-128461091 on Build GRCh38
rs755978392 Chr.11: 128460984-128460984 on Build GRCh38
rs572798580 Chr.11: 128461249-128461249 on Build GRCh38
rs1043809024 Chr.11: 128460977-128460977 on Build GRCh38
rs770493084 Chr.11: 128461220-128461220 on Build GRCh38
rs941813171 Chr.11: 128460981-128460981 on Build GRCh38
rs985834276 Chr.11: 128460787-128460787 on Build GRCh38
rs376429213 Chr.11: 128461148-128461148 on Build GRCh38
rs555321639 Chr.11: 128461033-128461033 on Build GRCh38
rs967581025 Chr.11: 128461177-128461177 on Build GRCh38
rs567572663 Chr.11: 128461157-128461157 on Build GRCh38
rs994355644 Chr.11: 128461009-128461009 on Build GRCh38
rs1030922895 Chr.11: 128460995-128460995 on Build GRCh38
rs976871545 Chr.11: 128460713-128460713 on Build GRCh38
rs200501677 Chr.11: 128460987-128460987 on Build GRCh38
rs559525717 Chr.11: 128460871-128460871 on Build GRCh38
rs1021729051 Chr.11: 128461161-128461161 on Build GRCh38
rs918805243 Chr.11: 128461239-128461239 on Build GRCh38
rs568827381 Chr.11: 128460943-128460943 on Build GRCh38
rs997722360 Chr.11: 128461112-128461112 on Build GRCh38
rs552924673 Chr.11: 128461216-128461216 on Build GRCh38
rs954078348 Chr.11: 128460742-128460742 on Build GRCh38
rs765428249 Chr.11: 128461237-128461237 on Build GRCh38
rs909902028 Chr.11: 128461215-128461215 on Build GRCh38
rs941638086 Chr.11: 128460794-128460794 on Build GRCh38
rs766392041 Chr.11: 128460738-128460738 on Build GRCh38
rs977283012 Chr.11: 128461197-128461197 on Build GRCh38
rs888856601 Chr.11: 128460956-128460956 on Build GRCh38
rs985544280 Chr.11: 128461200-128461200 on Build GRCh38
rs761930196 Chr.11: 128461225-128461225 on Build GRCh38
rs537022278 Chr.11: 128460944-128460944 on Build GRCh38
rs1011642040 Chr.11: 128461154-128461154 on Build GRCh38
rs1037232249 Chr.11: 128460831-128460831 on Build GRCh38
rs550783844 Chr.11: 128461073-128461073 on Build GRCh38
rs536552640 Chr.11: 128461204-128461204 on Build GRCh38
rs1029758313 Chr.11: 128461116-128461116 on Build GRCh38
rs972962634 Chr.11: 128461230-128461230 on Build GRCh38
rs537212976 Chr.11: 128461072-128461072 on Build GRCh38
rs1037431569 Chr.11: 128460981-128460981 on Build GRCh38
rs528581490 Chr.11: 128460942-128460942 on Build GRCh38
rs945917893 Chr.11: 128460733-128460733 on Build GRCh38
rs989972145 Chr.11: 128460721-128460721 on Build GRCh38
rs932940485 Chr.11: 128460858-128460858 on Build GRCh38
rs12288765 Chr.11: 128460792-128460792 on Build GRCh38
rs530039931 Chr.11: 128460732-128460732 on Build GRCh38
rs867377472 Chr.11: 128460784-128460784 on Build GRCh38
rs1003280118 Chr.11: 128461099-128461099 on Build GRCh38

View Full Product Details