Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6536792-6537391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAPDH
Entrez Gene ID
2597
Gene Name
glyceraldehyde-3-phosphate dehydrogenase
Gene Aliases
G3PD, GAPD, HEL-S-162eP
UniGene
Hs.544577
Gene Chromosome Location
Chr.12: 6534405-6538375 on Build GRCh38
Transcripts
NM_001256799.2 NM_001289745.1 NM_001289746.1 NM_002046.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1363707 c.339delG p.G115fs*21 Chr.12: 6537112-6537112 on Build GRCh38
7236353 c.274G>C p.A92P Chr.12: 6536957-6536957 on Build GRCh38
3955065 c.274G>A p.A92T Chr.12: 6536957-6536957 on Build GRCh38
942605 c.266A>G p.D89G Chr.12: 6536949-6536949 on Build GRCh38
397993 c.304T>A p.F102I Chr.12: 6536987-6536987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758590819 Chr.12: 6537296-6537296 on Build GRCh38
rs769281153 Chr.12: 6536864-6536864 on Build GRCh38
rs888937653 Chr.12: 6536922-6536922 on Build GRCh38
rs769671313 Chr.12: 6537359-6537359 on Build GRCh38
rs567602385 Chr.12: 6537018-6537018 on Build GRCh38
rs376053895 Chr.12: 6537212-6537212 on Build GRCh38
rs45579041 Chr.12: 6537067-6537067 on Build GRCh38
rs750217821 Chr.12: 6537020-6537031 on Build GRCh38
rs752569001 Chr.12: 6537088-6537088 on Build GRCh38
rs762501270 Chr.12: 6537368-6537368 on Build GRCh38
rs534300550 Chr.12: 6536968-6536968 on Build GRCh38
rs372259316 Chr.12: 6536877-6536877 on Build GRCh38
rs908960082 Chr.12: 6537288-6537288 on Build GRCh38
rs754522695 Chr.12: 6536852-6536852 on Build GRCh38
rs113341953 Chr.12: 6536942-6536942 on Build GRCh38
rs367712305 Chr.12: 6536898-6536898 on Build GRCh38
rs779153011 Chr.12: 6537295-6537295 on Build GRCh38
rs780109655 Chr.12: 6536848-6536848 on Build GRCh38
rs776162625 Chr.12: 6537199-6537199 on Build GRCh38
rs764928240 Chr.12: 6537207-6537207 on Build GRCh38
rs370355573 Chr.12: 6537221-6537221 on Build GRCh38
rs146516445 Chr.12: 6537150-6537150 on Build GRCh38
rs11266264 Chr.12: 6536929-6536929 on Build GRCh38
rs770105201 Chr.12: 6536969-6536969 on Build GRCh38
rs371192512 Chr.12: 6536966-6536966 on Build GRCh38
rs774700281 Chr.12: 6537166-6537166 on Build GRCh38
rs758108302 Chr.12: 6536904-6536904 on Build GRCh38
rs11549324 Chr.12: 6536965-6536965 on Build GRCh38
rs746840589 Chr.12: 6537312-6537312 on Build GRCh38
rs771766319 Chr.12: 6537173-6537173 on Build GRCh38
rs373809061 Chr.12: 6537230-6537230 on Build GRCh38
rs538867812 Chr.12: 6536912-6536912 on Build GRCh38
rs762862068 Chr.12: 6537022-6537023 on Build GRCh38
rs780898155 Chr.12: 6537333-6537333 on Build GRCh38
rs748092877 Chr.12: 6537353-6537353 on Build GRCh38
rs371116969 Chr.12: 6537235-6537235 on Build GRCh38
rs761293659 Chr.12: 6537202-6537202 on Build GRCh38
rs759382699 Chr.12: 6537076-6537076 on Build GRCh38
rs751972977 Chr.12: 6537136-6537136 on Build GRCh38
rs760555162 Chr.12: 6537178-6537178 on Build GRCh38
rs774807547 Chr.12: 6536869-6536869 on Build GRCh38
rs11549330 Chr.12: 6537366-6537366 on Build GRCh38
rs11549340 Chr.12: 6537117-6537117 on Build GRCh38
rs774327990 Chr.12: 6537072-6537072 on Build GRCh38
rs11549350 Chr.12: 6537103-6537103 on Build GRCh38
rs760591074 Chr.12: 6537112-6537112 on Build GRCh38
rs781595284 Chr.12: 6537297-6537298 on Build GRCh38
rs11266742 Chr.12: 6536992-6536992 on Build GRCh38
rs1007669348 Chr.12: 6536941-6536941 on Build GRCh38
rs767788286 Chr.12: 6537210-6537210 on Build GRCh38
rs188736603 Chr.12: 6537028-6537028 on Build GRCh38
rs74407849 Chr.12: 6537194-6537194 on Build GRCh38
rs747569865 Chr.12: 6536915-6536915 on Build GRCh38
rs776220592 Chr.12: 6536830-6536846 on Build GRCh38
rs769758398 Chr.12: 6537050-6537050 on Build GRCh38
rs762430400 Chr.12: 6537245-6537245 on Build GRCh38
rs576651948 Chr.12: 6537031-6537031 on Build GRCh38
rs972210459 Chr.12: 6537315-6537315 on Build GRCh38
rs747848762 Chr.12: 6536858-6536858 on Build GRCh38
rs768495247 Chr.12: 6537329-6537329 on Build GRCh38
rs765097888 Chr.12: 6536831-6536831 on Build GRCh38
rs755563954 Chr.12: 6537012-6537012 on Build GRCh38
rs756848400 Chr.12: 6537029-6537029 on Build GRCh38
rs11549326 Chr.12: 6537367-6537367 on Build GRCh38
rs375460941 Chr.12: 6537041-6537041 on Build GRCh38
rs764055385 Chr.12: 6537185-6537185 on Build GRCh38
rs755059399 Chr.12: 6537282-6537283 on Build GRCh38
rs762537181 Chr.12: 6537283-6537283 on Build GRCh38
rs773276926 Chr.12: 6537057-6537057 on Build GRCh38
rs763523311 Chr.12: 6537115-6537115 on Build GRCh38
rs375458345 Chr.12: 6536891-6536891 on Build GRCh38
rs750138825 Chr.12: 6536839-6536839 on Build GRCh38
rs781529712 Chr.12: 6537037-6537037 on Build GRCh38
rs941702915 Chr.12: 6536884-6536884 on Build GRCh38
rs77154254 Chr.12: 6537346-6537346 on Build GRCh38
rs755829002 Chr.12: 6536921-6536921 on Build GRCh38
rs376485668 Chr.12: 6536853-6536853 on Build GRCh38
rs766044440 Chr.12: 6537271-6537271 on Build GRCh38
rs369749848 Chr.12: 6537069-6537069 on Build GRCh38
rs780304796 Chr.12: 6537305-6537305 on Build GRCh38
rs750273368 Chr.12: 6536903-6536903 on Build GRCh38
rs11549356 Chr.12: 6536931-6536931 on Build GRCh38
rs11549336 Chr.12: 6537169-6537169 on Build GRCh38
rs372910824 Chr.12: 6537208-6537208 on Build GRCh38
rs149945423 Chr.12: 6536984-6536984 on Build GRCh38
rs369856016 Chr.12: 6536909-6536909 on Build GRCh38
rs117243681 Chr.12: 6537231-6537231 on Build GRCh38
rs767544699 Chr.12: 6537080-6537080 on Build GRCh38
rs368487746 Chr.12: 6536823-6536823 on Build GRCh38
rs1033890711 Chr.12: 6537261-6537261 on Build GRCh38
rs751350310 Chr.12: 6536990-6536990 on Build GRCh38
rs775182146 Chr.12: 6537244-6537244 on Build GRCh38
rs766023125 Chr.12: 6537284-6537284 on Build GRCh38
rs11549353 Chr.12: 6536947-6536947 on Build GRCh38
rs754051138 Chr.12: 6536820-6536820 on Build GRCh38
rs192121392 Chr.12: 6537030-6537030 on Build GRCh38
rs375963780 Chr.12: 6536998-6536998 on Build GRCh38
rs11549338 Chr.12: 6537147-6537147 on Build GRCh38
rs778509730 Chr.12: 6537130-6537130 on Build GRCh38
rs757776820 Chr.12: 6537290-6537290 on Build GRCh38
rs200281741 Chr.12: 6536956-6536956 on Build GRCh38
rs926823751 Chr.12: 6537066-6537066 on Build GRCh38
rs752299792 Chr.12: 6537007-6537007 on Build GRCh38
rs375046413 Chr.12: 6537262-6537262 on Build GRCh38
rs759577147 Chr.12: 6536995-6536995 on Build GRCh38
rs772613331 Chr.12: 6536870-6536870 on Build GRCh38
rs776520100 Chr.12: 6536895-6536895 on Build GRCh38
rs371116598 Chr.12: 6536983-6536983 on Build GRCh38
rs753299793 Chr.12: 6537118-6537118 on Build GRCh38
rs753064371 Chr.12: 6537211-6537211 on Build GRCh38
rs368328203 Chr.12: 6537269-6537269 on Build GRCh38
rs144761391 Chr.12: 6537175-6537175 on Build GRCh38
rs11549322 Chr.12: 6537106-6537106 on Build GRCh38
rs982259127 Chr.12: 6537064-6537064 on Build GRCh38
rs368740142 Chr.12: 6537145-6537145 on Build GRCh38
rs372101846 Chr.12: 6537070-6537070 on Build GRCh38
rs370350073 Chr.12: 6537151-6537151 on Build GRCh38
rs772747939 Chr.12: 6537363-6537363 on Build GRCh38
rs1065691 Chr.12: 6537154-6537154 on Build GRCh38
rs569921809 Chr.12: 6536868-6536868 on Build GRCh38
rs11549333 Chr.12: 6537323-6537323 on Build GRCh38
rs745721511 Chr.12: 6537167-6537167 on Build GRCh38
rs543388326 Chr.12: 6537285-6537285 on Build GRCh38
rs370461292 Chr.12: 6537021-6537021 on Build GRCh38
rs766348481 Chr.12: 6537080-6537083 on Build GRCh38
rs769194156 Chr.12: 6537036-6537036 on Build GRCh38
rs549852225 Chr.12: 6536856-6536856 on Build GRCh38
rs766482229 Chr.12: 6536908-6536908 on Build GRCh38
rs11549329 Chr.12: 6536979-6536979 on Build GRCh38
rs138863602 Chr.12: 6537209-6537209 on Build GRCh38
rs991238799 Chr.12: 6537068-6537068 on Build GRCh38
rs773838199 Chr.12: 6537289-6537289 on Build GRCh38
rs779065074 Chr.12: 6537234-6537234 on Build GRCh38
rs552376556 Chr.12: 6536940-6536940 on Build GRCh38
rs11549349 Chr.12: 6537121-6537121 on Build GRCh38
rs758897659 Chr.12: 6537142-6537142 on Build GRCh38
rs369572357 Chr.12: 6537241-6537241 on Build GRCh38
rs572265558 Chr.12: 6537157-6537157 on Build GRCh38
rs759383529 Chr.12: 6537308-6537308 on Build GRCh38
rs765340513 Chr.12: 6536902-6536902 on Build GRCh38
rs758395461 Chr.12: 6536847-6536847 on Build GRCh38
rs764987029 Chr.12: 6537019-6537020 on Build GRCh38
rs11549374 Chr.12: 6537001-6537001 on Build GRCh38
rs761381114 Chr.12: 6537253-6537253 on Build GRCh38
rs11549337 Chr.12: 6536986-6536986 on Build GRCh38
rs776540125 Chr.12: 6537252-6537252 on Build GRCh38
rs746568927 Chr.12: 6536851-6536851 on Build GRCh38
rs11549363 Chr.12: 6536989-6536989 on Build GRCh38
rs751702388 Chr.12: 6537276-6537277 on Build GRCh38
rs762119403 Chr.12: 6536899-6536899 on Build GRCh38
rs771327842 Chr.12: 6537160-6537160 on Build GRCh38
rs761702708 Chr.12: 6536902-6536903 on Build GRCh38

View Full Product Details