Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51295670-51296046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN2
Entrez Gene ID
51411
Gene Name
bridging integrator 2
Gene Aliases
BRAP-1
UniGene
Hs.14770
Gene Chromosome Location
Chr.12: 51281038-51324680 on Build GRCh38
Transcripts
NM_001290007.1 NM_001290008.1 NM_001290009.1 NM_016293.3 XM_005268957.3 XM_005268958.3 XM_017019422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6488283 c.758C>A p.S253* Chr.12: 51295799-51295799 on Build GRCh38
6072762 c.667G>C p.V223L Chr.12: 51295812-51295812 on Build GRCh38
940475 c.737T>C p.V246A Chr.12: 51295820-51295820 on Build GRCh38
4442715 c.697G>A p.E233K Chr.12: 51295860-51295860 on Build GRCh38
6220322 c.631A>C p.K211Q Chr.12: 51295848-51295848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770133337 Chr.12: 51295814-51295814 on Build GRCh38
rs371296230 Chr.12: 51295872-51295872 on Build GRCh38
rs866914236 Chr.12: 51295766-51295766 on Build GRCh38
rs555906994 Chr.12: 51295785-51295785 on Build GRCh38
rs200511170 Chr.12: 51295924-51295924 on Build GRCh38
rs778577727 Chr.12: 51295748-51295748 on Build GRCh38
rs781087230 Chr.12: 51295810-51295810 on Build GRCh38
rs752515027 Chr.12: 51295889-51295889 on Build GRCh38
rs928053520 Chr.12: 51295933-51295933 on Build GRCh38
rs878928556 Chr.12: 51295700-51295700 on Build GRCh38
rs1046190796 Chr.12: 51295835-51295835 on Build GRCh38
rs758257046 Chr.12: 51295784-51295784 on Build GRCh38
rs780678634 Chr.12: 51295817-51295817 on Build GRCh38
rs374431646 Chr.12: 51295767-51295767 on Build GRCh38
rs543552231 Chr.12: 51296009-51296009 on Build GRCh38
rs757368013 Chr.12: 51295892-51295892 on Build GRCh38
rs1008070017 Chr.12: 51295939-51295939 on Build GRCh38
rs754822349 Chr.12: 51295759-51295759 on Build GRCh38
rs780404584 Chr.12: 51295922-51295922 on Build GRCh38
rs148656859 Chr.12: 51295827-51295827 on Build GRCh38
rs759292596 Chr.12: 51295884-51295884 on Build GRCh38
rs890915293 Chr.12: 51295938-51295938 on Build GRCh38
rs745978130 Chr.12: 51295811-51295811 on Build GRCh38
rs572287300 Chr.12: 51295860-51295860 on Build GRCh38
rs757504090 Chr.12: 51295781-51295781 on Build GRCh38
rs748527283 Chr.12: 51295927-51295927 on Build GRCh38
rs749771729 Chr.12: 51295923-51295923 on Build GRCh38
rs373066757 Chr.12: 51295764-51295764 on Build GRCh38
rs964017189 Chr.12: 51295724-51295724 on Build GRCh38
rs779361581 Chr.12: 51295925-51295925 on Build GRCh38
rs767785621 Chr.12: 51295902-51295902 on Build GRCh38
rs766209007 Chr.12: 51295879-51295879 on Build GRCh38
rs541013236 Chr.12: 51295890-51295890 on Build GRCh38
rs78357540 Chr.12: 51295976-51295976 on Build GRCh38
rs764978534 Chr.12: 51295888-51295888 on Build GRCh38
rs749659460 Chr.12: 51295819-51295819 on Build GRCh38
rs368866801 Chr.12: 51295798-51295798 on Build GRCh38
rs1018414033 Chr.12: 51295998-51295998 on Build GRCh38
rs750466009 Chr.12: 51295908-51295908 on Build GRCh38
rs1041551668 Chr.12: 51295930-51295930 on Build GRCh38
rs369331611 Chr.12: 51295741-51295741 on Build GRCh38
rs908069307 Chr.12: 51295804-51295804 on Build GRCh38
rs760887258 Chr.12: 51295921-51295936 on Build GRCh38
rs760296366 Chr.12: 51295873-51295873 on Build GRCh38
rs771769155 Chr.12: 51295861-51295861 on Build GRCh38
rs899567370 Chr.12: 51295763-51295763 on Build GRCh38
rs776066936 Chr.12: 51295916-51295916 on Build GRCh38
rs577767489 Chr.12: 51295951-51295951 on Build GRCh38
rs954826242 Chr.12: 51295800-51295800 on Build GRCh38
rs1018389021 Chr.12: 51295694-51295694 on Build GRCh38
rs768129226 Chr.12: 51295825-51295825 on Build GRCh38

View Full Product Details