Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 85979787-85980335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGAT4C
Entrez Gene ID
25834
Gene Name
MGAT4 family member C
Gene Aliases
GNTIVH, HGNT-IV-H
UniGene
Hs.589093
Gene Chromosome Location
Chr.12: 85972281-86838999 on Build GRCh38
Transcripts
NM_013244.3 XM_005268776.4 XM_005268779.3 XM_011538150.2 XM_011538151.2 XM_011538156.2 XM_011538157.2 XM_017019143.1 XM_017019144.1 XM_017019145.1 XM_017019146.1 XM_017019147.1 XM_017019148.1 XM_017019149.1 XM_017019150.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7294704 c.445C>A p.R149S Chr.12: 85980281-85980281 on Build GRCh38
6455458 c.766A>C p.K256Q Chr.12: 85979960-85979960 on Build GRCh38
348785 c.595C>T p.R199C Chr.12: 85980131-85980131 on Build GRCh38
1994610 c.400G>A p.E134K Chr.12: 85980326-85980326 on Build GRCh38
4782412 c.931G>T p.E311* Chr.12: 85979882-85979882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757539324 Chr.12: 85980240-85980240 on Build GRCh38
rs897303861 Chr.12: 85979987-85979987 on Build GRCh38
rs566043053 Chr.12: 85980223-85980223 on Build GRCh38
rs914840735 Chr.12: 85980111-85980111 on Build GRCh38
rs753925997 Chr.12: 85979861-85979861 on Build GRCh38
rs770702724 Chr.12: 85980130-85980130 on Build GRCh38
rs753086343 Chr.12: 85980001-85980001 on Build GRCh38
rs866745321 Chr.12: 85980074-85980074 on Build GRCh38
rs765429825 Chr.12: 85980243-85980243 on Build GRCh38
rs372911876 Chr.12: 85980066-85980066 on Build GRCh38
rs140682310 Chr.12: 85979818-85979818 on Build GRCh38
rs779860939 Chr.12: 85980050-85980050 on Build GRCh38
rs1038028949 Chr.12: 85979826-85979826 on Build GRCh38
rs75276700 Chr.12: 85980037-85980037 on Build GRCh38
rs141479653 Chr.12: 85979917-85979917 on Build GRCh38
rs755975066 Chr.12: 85980271-85980271 on Build GRCh38
rs756220458 Chr.12: 85980007-85980007 on Build GRCh38
rs150545721 Chr.12: 85980166-85980166 on Build GRCh38
rs768126876 Chr.12: 85980000-85980000 on Build GRCh38
rs753826277 Chr.12: 85980222-85980222 on Build GRCh38
rs142839367 Chr.12: 85980270-85980270 on Build GRCh38
rs749402312 Chr.12: 85979848-85979848 on Build GRCh38
rs759994326 Chr.12: 85980152-85980152 on Build GRCh38
rs375386863 Chr.12: 85979922-85979922 on Build GRCh38
rs777409272 Chr.12: 85980274-85980274 on Build GRCh38
rs374349115 Chr.12: 85979841-85979841 on Build GRCh38
rs768547952 Chr.12: 85979877-85979877 on Build GRCh38
rs989044611 Chr.12: 85980057-85980057 on Build GRCh38
rs765452856 Chr.12: 85979915-85979915 on Build GRCh38
rs141672136 Chr.12: 85980157-85980157 on Build GRCh38
rs769372443 Chr.12: 85980098-85980098 on Build GRCh38
rs138500929 Chr.12: 85979835-85979835 on Build GRCh38
rs748169936 Chr.12: 85980260-85980260 on Build GRCh38
rs746800765 Chr.12: 85980059-85980059 on Build GRCh38
rs1007149128 Chr.12: 85980245-85980245 on Build GRCh38
rs760188889 Chr.12: 85979978-85979978 on Build GRCh38
rs754111075 Chr.12: 85980043-85980043 on Build GRCh38
rs539250548 Chr.12: 85979943-85979943 on Build GRCh38
rs748796788 Chr.12: 85980280-85980280 on Build GRCh38
rs770595521 Chr.12: 85980132-85980132 on Build GRCh38
rs956407562 Chr.12: 85980049-85980049 on Build GRCh38
rs547661699 Chr.12: 85979974-85979974 on Build GRCh38
rs143449252 Chr.12: 85980060-85980060 on Build GRCh38
rs748989192 Chr.12: 85980116-85980116 on Build GRCh38
rs754801625 Chr.12: 85980252-85980252 on Build GRCh38
rs776093722 Chr.12: 85980159-85980159 on Build GRCh38
rs774149822 Chr.12: 85980300-85980300 on Build GRCh38
rs147998015 Chr.12: 85980082-85980082 on Build GRCh38
rs114903783 Chr.12: 85979819-85979819 on Build GRCh38
rs772953057 Chr.12: 85979940-85979940 on Build GRCh38
rs970146612 Chr.12: 85979949-85979949 on Build GRCh38
rs566418071 Chr.12: 85980281-85980281 on Build GRCh38
rs145889731 Chr.12: 85979825-85979825 on Build GRCh38
rs113387514 Chr.12: 85980216-85980216 on Build GRCh38
rs764040141 Chr.12: 85980009-85980009 on Build GRCh38
rs368524858 Chr.12: 85979958-85979958 on Build GRCh38
rs267603706 Chr.12: 85979919-85979919 on Build GRCh38
rs371453579 Chr.12: 85979916-85979916 on Build GRCh38
rs367612551 Chr.12: 85979914-85979914 on Build GRCh38
rs199811466 Chr.12: 85979998-85979998 on Build GRCh38
rs200155199 Chr.12: 85979887-85979887 on Build GRCh38
rs776069370 Chr.12: 85979900-85979900 on Build GRCh38
rs752818848 Chr.12: 85979833-85979833 on Build GRCh38
rs764279389 Chr.12: 85980198-85980198 on Build GRCh38
rs769121552 Chr.12: 85979926-85979926 on Build GRCh38
rs539968414 Chr.12: 85980305-85980305 on Build GRCh38
rs140499591 Chr.12: 85980244-85980244 on Build GRCh38
rs771884299 Chr.12: 85979849-85979849 on Build GRCh38
rs747999039 Chr.12: 85980088-85980088 on Build GRCh38
rs780805071 Chr.12: 85980255-85980255 on Build GRCh38
rs374072925 Chr.12: 85979870-85979870 on Build GRCh38
rs1018284129 Chr.12: 85980273-85980273 on Build GRCh38
rs778384478 Chr.12: 85980062-85980062 on Build GRCh38
rs746994463 Chr.12: 85979876-85979876 on Build GRCh38
rs968354452 Chr.12: 85980118-85980118 on Build GRCh38
rs749782221 Chr.12: 85980131-85980131 on Build GRCh38
rs536944591 Chr.12: 85980308-85980308 on Build GRCh38
rs766900187 Chr.12: 85979946-85979946 on Build GRCh38
rs772578702 Chr.12: 85980114-85980114 on Build GRCh38
rs757444701 Chr.12: 85980044-85980044 on Build GRCh38

View Full Product Details