Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93360150-93360749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DR1
Entrez Gene ID
1810
Gene Name
down-regulator of transcription 1
Gene Aliases
NC2, NC2-BETA, NC2B
UniGene
Hs.348418
Gene Chromosome Location
Chr.1: 93345921-93362592 on Build GRCh38
Transcripts
NM_001938.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6649216 c.443C>A p.A148D Chr.1: 93360551-93360551 on Build GRCh38
4494156 c.427C>T p.Q143* Chr.1: 93360535-93360535 on Build GRCh38
7266025 c.467C>T p.A156V Chr.1: 93360575-93360575 on Build GRCh38
5495340 c.493G>T p.G165* Chr.1: 93360601-93360601 on Build GRCh38
5502277 c.444C>T p.A148A Chr.1: 93360552-93360552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536164827 Chr.1: 93360197-93360197 on Build GRCh38
rs758545007 Chr.1: 93360672-93360672 on Build GRCh38
rs757082588 Chr.1: 93360616-93360618 on Build GRCh38
rs766616860 Chr.1: 93360600-93360600 on Build GRCh38
rs768881891 Chr.1: 93360574-93360574 on Build GRCh38
rs190835113 Chr.1: 93360680-93360680 on Build GRCh38
rs763993005 Chr.1: 93360447-93360447 on Build GRCh38
rs906309214 Chr.1: 93360199-93360199 on Build GRCh38
rs1011247742 Chr.1: 93360233-93360233 on Build GRCh38
rs754110067 Chr.1: 93360438-93360438 on Build GRCh38
rs765062063 Chr.1: 93360449-93360449 on Build GRCh38
rs908542912 Chr.1: 93360359-93360359 on Build GRCh38
rs1000141571 Chr.1: 93360334-93360334 on Build GRCh38
rs370453509 Chr.1: 93360305-93360306 on Build GRCh38
rs970853834 Chr.1: 93360407-93360407 on Build GRCh38
rs759223047 Chr.1: 93360525-93360525 on Build GRCh38
rs776108066 Chr.1: 93360235-93360235 on Build GRCh38
rs1052549094 Chr.1: 93360227-93360227 on Build GRCh38
rs772435498 Chr.1: 93360507-93360507 on Build GRCh38
rs776093104 Chr.1: 93360516-93360516 on Build GRCh38
rs755230782 Chr.1: 93360483-93360483 on Build GRCh38
rs758170074 Chr.1: 93360461-93360461 on Build GRCh38
rs779376447 Chr.1: 93360485-93360485 on Build GRCh38
rs749580808 Chr.1: 93360477-93360477 on Build GRCh38
rs199518567 Chr.1: 93360467-93360467 on Build GRCh38
rs753810358 Chr.1: 93360467-93360467 on Build GRCh38
rs996642296 Chr.1: 93360292-93360292 on Build GRCh38
rs183185754 Chr.1: 93360689-93360689 on Build GRCh38
rs375320336 Chr.1: 93360599-93360599 on Build GRCh38
rs182894486 Chr.1: 93360511-93360511 on Build GRCh38
rs775157072 Chr.1: 93360546-93360546 on Build GRCh38
rs752399742 Chr.1: 93360460-93360460 on Build GRCh38
rs764897702 Chr.1: 93360539-93360539 on Build GRCh38
rs12060611 Chr.1: 93360232-93360232 on Build GRCh38
rs200292726 Chr.1: 93360555-93360555 on Build GRCh38
rs926858040 Chr.1: 93360190-93360190 on Build GRCh38
rs974318054 Chr.1: 93360435-93360435 on Build GRCh38
rs763881675 Chr.1: 93360556-93360556 on Build GRCh38
rs956707738 Chr.1: 93360367-93360367 on Build GRCh38
rs543244473 Chr.1: 93360408-93360408 on Build GRCh38
rs367910460 Chr.1: 93360657-93360657 on Build GRCh38
rs980931984 Chr.1: 93360434-93360435 on Build GRCh38
rs186067633 Chr.1: 93360671-93360671 on Build GRCh38
rs982782738 Chr.1: 93360346-93360346 on Build GRCh38
rs747487257 Chr.1: 93360512-93360512 on Build GRCh38
rs187237811 Chr.1: 93360180-93360180 on Build GRCh38
rs575819469 Chr.1: 93360306-93360306 on Build GRCh38
rs963844575 Chr.1: 93360300-93360300 on Build GRCh38
rs191054369 Chr.1: 93360261-93360261 on Build GRCh38
rs962952541 Chr.1: 93360364-93360364 on Build GRCh38
rs1050007100 Chr.1: 93360720-93360720 on Build GRCh38
rs763958391 Chr.1: 93360463-93360463 on Build GRCh38
rs779029076 Chr.1: 93360660-93360660 on Build GRCh38
rs955669673 Chr.1: 93360329-93360329 on Build GRCh38
rs996670190 Chr.1: 93360311-93360311 on Build GRCh38
rs3088371 Chr.1: 93360621-93360621 on Build GRCh38
rs751455493 Chr.1: 93360466-93360466 on Build GRCh38
rs916198545 Chr.1: 93360447-93360447 on Build GRCh38
rs747216440 Chr.1: 93360386-93360386 on Build GRCh38
rs371065674 Chr.1: 93360514-93360514 on Build GRCh38
rs199636805 Chr.1: 93360466-93360467 on Build GRCh38
rs1029690441 Chr.1: 93360312-93360312 on Build GRCh38
rs1044827127 Chr.1: 93360186-93360186 on Build GRCh38
rs1003191182 Chr.1: 93360204-93360204 on Build GRCh38
rs748495754 Chr.1: 93360504-93360504 on Build GRCh38

View Full Product Details