Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160739186-160739563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLAMF7
Entrez Gene ID
57823
Gene Name
SLAM family member 7
Gene Aliases
19A, CD319, CRACC, CS1
UniGene
Hs.517265
Gene Chromosome Location
Chr.1: 160739057-160754821 on Build GRCh38
Transcripts
NM_001282588.1 NM_001282589.1 NM_001282590.1 NM_001282591.1 NM_001282592.1 NM_001282593.1 NM_001282594.1 NM_001282595.1 NM_001282596.1 NM_021181.4 XM_011509828.1 XM_011509829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7279395 c.35A>G p.Y12C Chr.1: 160739336-160739336 on Build GRCh38
5624112 c.9T>G p.G3G Chr.1: 160739310-160739310 on Build GRCh38
6059089 c.54A>T p.T18T Chr.1: 160739355-160739355 on Build GRCh38
1194017 c.26C>T p.T9I Chr.1: 160739327-160739327 on Build GRCh38
375523 c.26C>A p.T9N Chr.1: 160739327-160739327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79681994 Chr.1: 160739267-160739267 on Build GRCh38
rs952026743 Chr.1: 160739280-160739280 on Build GRCh38
rs780656572 Chr.1: 160739318-160739318 on Build GRCh38
rs748902302 Chr.1: 160739402-160739402 on Build GRCh38
rs1006403267 Chr.1: 160739495-160739495 on Build GRCh38
rs1008198822 Chr.1: 160739455-160739455 on Build GRCh38
rs142843575 Chr.1: 160739321-160739321 on Build GRCh38
rs896401533 Chr.1: 160739431-160739431 on Build GRCh38
rs1029269992 Chr.1: 160739501-160739501 on Build GRCh38
rs759101192 Chr.1: 160739368-160739368 on Build GRCh38
rs372078303 Chr.1: 160739307-160739308 on Build GRCh38
rs769783823 Chr.1: 160739359-160739359 on Build GRCh38
rs554153726 Chr.1: 160739450-160739450 on Build GRCh38
rs757987743 Chr.1: 160739278-160739278 on Build GRCh38
rs951893534 Chr.1: 160739507-160739507 on Build GRCh38
rs761851071 Chr.1: 160739358-160739358 on Build GRCh38
rs750875024 Chr.1: 160739396-160739396 on Build GRCh38
rs377672387 Chr.1: 160739394-160739394 on Build GRCh38
rs180849166 Chr.1: 160739503-160739503 on Build GRCh38
rs568425830 Chr.1: 160739490-160739490 on Build GRCh38
rs780337097 Chr.1: 160739339-160739339 on Build GRCh38
rs760472213 Chr.1: 160739260-160739260 on Build GRCh38
rs750399761 Chr.1: 160739478-160739478 on Build GRCh38
rs369009453 Chr.1: 160739338-160739338 on Build GRCh38
rs780374283 Chr.1: 160739327-160739327 on Build GRCh38
rs758542980 Chr.1: 160739397-160739397 on Build GRCh38
rs1047581852 Chr.1: 160739464-160739464 on Build GRCh38
rs1019713816 Chr.1: 160739470-160739470 on Build GRCh38
rs534293697 Chr.1: 160739517-160739517 on Build GRCh38
rs375178736 Chr.1: 160739384-160739384 on Build GRCh38
rs753023011 Chr.1: 160739296-160739296 on Build GRCh38
rs566550447 Chr.1: 160739214-160739214 on Build GRCh38
rs1028498362 Chr.1: 160739295-160739295 on Build GRCh38
rs776747789 Chr.1: 160739271-160739271 on Build GRCh38
rs113404744 Chr.1: 160739460-160739460 on Build GRCh38
rs778109093 Chr.1: 160739298-160739298 on Build GRCh38
rs757706994 Chr.1: 160739316-160739316 on Build GRCh38
rs747393576 Chr.1: 160739342-160739342 on Build GRCh38
rs892495759 Chr.1: 160739449-160739449 on Build GRCh38
rs754404591 Chr.1: 160739310-160739310 on Build GRCh38
rs1005913809 Chr.1: 160739231-160739231 on Build GRCh38
rs1055047729 Chr.1: 160739411-160739411 on Build GRCh38
rs1052700875 Chr.1: 160739442-160739442 on Build GRCh38
rs747167147 Chr.1: 160739331-160739331 on Build GRCh38
rs777376514 Chr.1: 160739369-160739369 on Build GRCh38
rs1014833004 Chr.1: 160739542-160739542 on Build GRCh38
rs112761831 Chr.1: 160739233-160739233 on Build GRCh38
rs985149136 Chr.1: 160739537-160739538 on Build GRCh38
rs766012272 Chr.1: 160739289-160739289 on Build GRCh38
rs757792753 Chr.1: 160739389-160739389 on Build GRCh38
rs555046020 Chr.1: 160739489-160739489 on Build GRCh38
rs979516442 Chr.1: 160739381-160739381 on Build GRCh38
rs749890588 Chr.1: 160739275-160739275 on Build GRCh38
rs780426694 Chr.1: 160739333-160739333 on Build GRCh38
rs776553989 Chr.1: 160739339-160739339 on Build GRCh38
rs1008356063 Chr.1: 160739212-160739212 on Build GRCh38
rs779991572 Chr.1: 160739406-160739406 on Build GRCh38
rs538112723 Chr.1: 160739297-160739297 on Build GRCh38
rs1015986864 Chr.1: 160739505-160739505 on Build GRCh38
rs532304247 Chr.1: 160739252-160739252 on Build GRCh38
rs746092187 Chr.1: 160739251-160739251 on Build GRCh38
rs772207834 Chr.1: 160739322-160739322 on Build GRCh38
rs370657629 Chr.1: 160739364-160739364 on Build GRCh38
rs772625868 Chr.1: 160739363-160739363 on Build GRCh38
rs775181827 Chr.1: 160739255-160739255 on Build GRCh38
rs936944926 Chr.1: 160739436-160739436 on Build GRCh38
rs569164097 Chr.1: 160739272-160739272 on Build GRCh38
rs768637770 Chr.1: 160739336-160739336 on Build GRCh38
rs755674796 Chr.1: 160739365-160739365 on Build GRCh38
rs770594560 Chr.1: 160739408-160739408 on Build GRCh38

View Full Product Details