Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39224508-39225107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDGFB
Entrez Gene ID
5155
Gene Name
platelet derived growth factor subunit B
Gene Aliases
IBGC5, PDGF-2, PDGF2, SIS, SSV, c-sis
UniGene
Hs.1976
Gene Chromosome Location
Chr.22: 39223359-39245055 on Build GRCh38
Transcripts
NM_002608.3 NM_033016.3

SNP Detail

SNP ID Chromosome Location
rs567660959 Chr.22: 39224761-39224761 on Build GRCh38
rs886993082 Chr.22: 39225003-39225003 on Build GRCh38
rs545232457 Chr.22: 39224696-39224696 on Build GRCh38
rs774669499 Chr.22: 39224944-39224944 on Build GRCh38
rs181373913 Chr.22: 39224773-39224773 on Build GRCh38
rs145279167 Chr.22: 39224974-39224974 on Build GRCh38
rs565859932 Chr.22: 39224809-39224809 on Build GRCh38
rs952892848 Chr.22: 39224637-39224637 on Build GRCh38
rs577959875 Chr.22: 39225005-39225005 on Build GRCh38
rs549059250 Chr.22: 39224760-39224760 on Build GRCh38
rs991337979 Chr.22: 39225065-39225065 on Build GRCh38
rs185652326 Chr.22: 39224785-39224785 on Build GRCh38
rs897586640 Chr.22: 39224624-39224624 on Build GRCh38
rs56133813 Chr.22: 39224636-39224636 on Build GRCh38
rs764439213 Chr.22: 39225051-39225051 on Build GRCh38
rs949923478 Chr.22: 39224808-39224808 on Build GRCh38
rs183030207 Chr.22: 39224992-39224992 on Build GRCh38
rs1010625385 Chr.22: 39224717-39224717 on Build GRCh38
rs3203596 Chr.22: 39225011-39225011 on Build GRCh38
rs915831979 Chr.22: 39224804-39224804 on Build GRCh38
rs964996806 Chr.22: 39225014-39225014 on Build GRCh38
rs1019287293 Chr.22: 39224834-39224834 on Build GRCh38
rs753875374 Chr.22: 39224545-39224546 on Build GRCh38
rs893417754 Chr.22: 39224680-39224680 on Build GRCh38
rs114630100 Chr.22: 39224975-39224975 on Build GRCh38
rs995721834 Chr.22: 39224979-39224979 on Build GRCh38
rs149665155 Chr.22: 39224662-39224662 on Build GRCh38
rs1041721477 Chr.22: 39224731-39224731 on Build GRCh38
rs957802600 Chr.22: 39225052-39225052 on Build GRCh38
rs745591450 Chr.22: 39225022-39225022 on Build GRCh38
rs761111970 Chr.22: 39225015-39225015 on Build GRCh38
rs982818169 Chr.22: 39224827-39224827 on Build GRCh38
rs1062604 Chr.22: 39224590-39224590 on Build GRCh38
rs190810375 Chr.22: 39224895-39224895 on Build GRCh38
rs559663878 Chr.22: 39224656-39224656 on Build GRCh38
rs1018358988 Chr.22: 39224950-39224950 on Build GRCh38
rs56380606 Chr.22: 39224642-39224642 on Build GRCh38
rs115714565 Chr.22: 39224840-39224840 on Build GRCh38
rs894179167 Chr.22: 39224580-39224580 on Build GRCh38
rs964230701 Chr.22: 39224713-39224713 on Build GRCh38
rs926065653 Chr.22: 39225039-39225039 on Build GRCh38
rs1004126702 Chr.22: 39225004-39225004 on Build GRCh38
rs115727092 Chr.22: 39225076-39225076 on Build GRCh38
rs114967981 Chr.22: 39224623-39224623 on Build GRCh38
rs530957505 Chr.22: 39224748-39224748 on Build GRCh38
rs533328919 Chr.22: 39224671-39224671 on Build GRCh38
rs73424751 Chr.22: 39224786-39224786 on Build GRCh38
rs946326809 Chr.22: 39224897-39224897 on Build GRCh38
rs369900992 Chr.22: 39224615-39224615 on Build GRCh38
rs116011535 Chr.22: 39224923-39224923 on Build GRCh38
rs975131957 Chr.22: 39224856-39224856 on Build GRCh38
rs150532025 Chr.22: 39224588-39224588 on Build GRCh38
rs112869822 Chr.22: 39224878-39224878 on Build GRCh38
rs975077845 Chr.22: 39225031-39225031 on Build GRCh38
rs748483053 Chr.22: 39224858-39224858 on Build GRCh38
rs17000897 Chr.22: 39225068-39225068 on Build GRCh38
rs55708806 Chr.22: 39224743-39224743 on Build GRCh38
rs1042395713 Chr.22: 39224920-39224920 on Build GRCh38
rs139775423 Chr.22: 39224857-39224857 on Build GRCh38
rs1018474018 Chr.22: 39224707-39224707 on Build GRCh38
rs775716108 Chr.22: 39224978-39224978 on Build GRCh38
rs925816768 Chr.22: 39224776-39224776 on Build GRCh38
rs555346802 Chr.22: 39224602-39224602 on Build GRCh38
rs990347665 Chr.22: 39224889-39224889 on Build GRCh38

View Full Product Details