Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 21623705-21624286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2L3
Entrez Gene ID
7332
Gene Name
ubiquitin conjugating enzyme E2 L3
Gene Aliases
E2-F1, L-UBC, UBCH7, UbcM4
UniGene
Hs.108104
Gene Chromosome Location
Chr.22: 21549447-21624034 on Build GRCh38
Transcripts
NM_001256355.1 NM_001256356.1 NM_003347.3 NR_028436.2 NR_046082.1

SNP Detail

SNP ID Chromosome Location
rs764929111 Chr.22: 21623758-21623758 on Build GRCh38
rs974746501 Chr.22: 21624123-21624123 on Build GRCh38
rs530408409 Chr.22: 21624233-21624233 on Build GRCh38
rs1007254317 Chr.22: 21624128-21624128 on Build GRCh38
rs1023659026 Chr.22: 21623951-21623951 on Build GRCh38
rs575711064 Chr.22: 21624164-21624164 on Build GRCh38
rs909240649 Chr.22: 21624041-21624041 on Build GRCh38
rs1052817901 Chr.22: 21624167-21624167 on Build GRCh38
rs565394889 Chr.22: 21624234-21624234 on Build GRCh38
rs5754409 Chr.22: 21624244-21624244 on Build GRCh38
rs932480937 Chr.22: 21623802-21623802 on Build GRCh38
rs371531565 Chr.22: 21624135-21624135 on Build GRCh38
rs916015181 Chr.22: 21624214-21624214 on Build GRCh38
rs983483730 Chr.22: 21623726-21623726 on Build GRCh38
rs1011659623 Chr.22: 21624198-21624198 on Build GRCh38
rs566612876 Chr.22: 21623800-21623800 on Build GRCh38
rs146403135 Chr.22: 21624216-21624216 on Build GRCh38
rs550483894 Chr.22: 21624009-21624009 on Build GRCh38
rs915907115 Chr.22: 21623998-21623998 on Build GRCh38
rs960229577 Chr.22: 21624164-21624165 on Build GRCh38
rs947377928 Chr.22: 21624006-21624006 on Build GRCh38
rs76342132 Chr.22: 21623997-21623997 on Build GRCh38
rs954447762 Chr.22: 21623757-21623757 on Build GRCh38
rs552931045 Chr.22: 21624152-21624152 on Build GRCh38
rs903113110 Chr.22: 21624024-21624024 on Build GRCh38
rs1003458385 Chr.22: 21623992-21623992 on Build GRCh38
rs938731444 Chr.22: 21623991-21623991 on Build GRCh38
rs368287885 Chr.22: 21623750-21623750 on Build GRCh38
rs909209569 Chr.22: 21623749-21623749 on Build GRCh38
rs529614245 Chr.22: 21623768-21623768 on Build GRCh38
rs867000295 Chr.22: 21624266-21624266 on Build GRCh38
rs141583045 Chr.22: 21623962-21623962 on Build GRCh38
rs550960923 Chr.22: 21623727-21623727 on Build GRCh38
rs16985160 Chr.22: 21624158-21624158 on Build GRCh38
rs770663490 Chr.22: 21624106-21624106 on Build GRCh38
rs565492623 Chr.22: 21624160-21624160 on Build GRCh38
rs892793436 Chr.22: 21623889-21623889 on Build GRCh38
rs974694790 Chr.22: 21623901-21623901 on Build GRCh38
rs1011214454 Chr.22: 21623908-21623908 on Build GRCh38
rs372734017 Chr.22: 21623778-21623778 on Build GRCh38
rs1050914983 Chr.22: 21623808-21623808 on Build GRCh38
rs5754407 Chr.22: 21624010-21624010 on Build GRCh38
rs892866493 Chr.22: 21624181-21624181 on Build GRCh38
rs560951287 Chr.22: 21623760-21623760 on Build GRCh38
rs1031325022 Chr.22: 21623858-21623858 on Build GRCh38
rs998558951 Chr.22: 21623853-21623853 on Build GRCh38
rs180774127 Chr.22: 21624129-21624129 on Build GRCh38
rs9621802 Chr.22: 21624086-21624086 on Build GRCh38
rs148369228 Chr.22: 21623838-21623838 on Build GRCh38
rs1003122360 Chr.22: 21624250-21624250 on Build GRCh38
rs575113277 Chr.22: 21623833-21623833 on Build GRCh38
rs117231304 Chr.22: 21624139-21624139 on Build GRCh38
rs1017220238 Chr.22: 21623799-21623799 on Build GRCh38
rs561473041 Chr.22: 21624230-21624230 on Build GRCh38
rs928827466 Chr.22: 21623963-21623963 on Build GRCh38
rs1035883537 Chr.22: 21624254-21624254 on Build GRCh38
rs1054210 Chr.22: 21623980-21623980 on Build GRCh38
rs377302320 Chr.22: 21623950-21623950 on Build GRCh38

View Full Product Details