Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44570647-44571158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZKSCAN7
Entrez Gene ID
55888
Gene Name
zinc finger with KRAB and SCAN domains 7
Gene Aliases
ZFP, ZNF167, ZNF448, ZNF64, ZSCAN39
UniGene
Hs.529512
Gene Chromosome Location
Chr.3: 44555175-44583464 on Build GRCh38
Transcripts
NM_001288590.1 NM_001288591.1 NM_001288592.1 NM_018651.3 NM_025169.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
130364 c.1879A>G p.R627G Chr.3: 44570989-44570989 on Build GRCh38
130363 c.1775C>G p.S592C Chr.3: 44570885-44570885 on Build GRCh38
4117738 c.2039T>C p.V680A Chr.3: 44571149-44571149 on Build GRCh38
4117737 c.1649C>A p.P550H Chr.3: 44570759-44570759 on Build GRCh38
6827340 c.1964G>A p.R655K Chr.3: 44571074-44571074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866272207 Chr.3: 44570741-44570741 on Build GRCh38
rs766396706 Chr.3: 44570787-44570787 on Build GRCh38
rs374620929 Chr.3: 44570837-44570837 on Build GRCh38
rs758257963 Chr.3: 44570893-44570900 on Build GRCh38
rs369490311 Chr.3: 44570717-44570717 on Build GRCh38
rs773100610 Chr.3: 44570958-44570958 on Build GRCh38
rs373560883 Chr.3: 44571101-44571101 on Build GRCh38
rs764398272 Chr.3: 44570892-44570892 on Build GRCh38
rs757722057 Chr.3: 44570902-44570902 on Build GRCh38
rs769457938 Chr.3: 44570712-44570712 on Build GRCh38
rs553442479 Chr.3: 44570820-44570820 on Build GRCh38
rs369673824 Chr.3: 44570982-44570982 on Build GRCh38
rs201164971 Chr.3: 44570833-44570833 on Build GRCh38
rs267599835 Chr.3: 44571079-44571079 on Build GRCh38
rs544033067 Chr.3: 44570971-44570971 on Build GRCh38
rs781541358 Chr.3: 44570705-44570705 on Build GRCh38
rs751799035 Chr.3: 44570692-44570692 on Build GRCh38
rs376393171 Chr.3: 44571014-44571014 on Build GRCh38
rs375856259 Chr.3: 44570758-44570758 on Build GRCh38
rs775446273 Chr.3: 44571086-44571086 on Build GRCh38
rs747103298 Chr.3: 44571089-44571089 on Build GRCh38
rs781065351 Chr.3: 44571059-44571061 on Build GRCh38
rs748452710 Chr.3: 44570711-44570711 on Build GRCh38
rs758376202 Chr.3: 44570687-44570687 on Build GRCh38
rs145479931 Chr.3: 44570759-44570759 on Build GRCh38
rs768404955 Chr.3: 44570940-44570940 on Build GRCh38
rs779697093 Chr.3: 44570905-44570906 on Build GRCh38
rs780858349 Chr.3: 44570941-44570941 on Build GRCh38
rs760536261 Chr.3: 44570763-44570763 on Build GRCh38
rs371308975 Chr.3: 44570726-44570726 on Build GRCh38
rs747931760 Chr.3: 44571032-44571032 on Build GRCh38
rs763563472 Chr.3: 44571123-44571123 on Build GRCh38
rs35320772 Chr.3: 44570876-44570876 on Build GRCh38
rs774471692 Chr.3: 44570859-44570859 on Build GRCh38
rs147008651 Chr.3: 44570813-44570813 on Build GRCh38
rs1020277044 Chr.3: 44570912-44570912 on Build GRCh38
rs145193307 Chr.3: 44570762-44570762 on Build GRCh38
rs539938290 Chr.3: 44571096-44571096 on Build GRCh38
rs200231048 Chr.3: 44570894-44570894 on Build GRCh38
rs754912071 Chr.3: 44571058-44571060 on Build GRCh38
rs373307729 Chr.3: 44570999-44570999 on Build GRCh38
rs765725369 Chr.3: 44571109-44571109 on Build GRCh38
rs753124352 Chr.3: 44570808-44570808 on Build GRCh38
rs761758091 Chr.3: 44570786-44570786 on Build GRCh38
rs760904660 Chr.3: 44570883-44570883 on Build GRCh38
rs573149561 Chr.3: 44570822-44570822 on Build GRCh38
rs149688084 Chr.3: 44570748-44570748 on Build GRCh38
rs751019553 Chr.3: 44571000-44571000 on Build GRCh38
rs983103891 Chr.3: 44571106-44571106 on Build GRCh38
rs747961378 Chr.3: 44570947-44570947 on Build GRCh38
rs756400843 Chr.3: 44570811-44570811 on Build GRCh38
rs750576727 Chr.3: 44570674-44570674 on Build GRCh38
rs182377695 Chr.3: 44570981-44570981 on Build GRCh38
rs776408245 Chr.3: 44570784-44570784 on Build GRCh38
rs267599834 Chr.3: 44571038-44571038 on Build GRCh38
rs766669730 Chr.3: 44570691-44570691 on Build GRCh38
rs755148349 Chr.3: 44570701-44570701 on Build GRCh38
rs148241240 Chr.3: 44570962-44570962 on Build GRCh38
rs974528503 Chr.3: 44571055-44571055 on Build GRCh38
rs1027531148 Chr.3: 44571025-44571025 on Build GRCh38
rs1003605125 Chr.3: 44570974-44570974 on Build GRCh38
rs377644061 Chr.3: 44570917-44570917 on Build GRCh38
rs894052651 Chr.3: 44570861-44570861 on Build GRCh38
rs768831399 Chr.3: 44571099-44571099 on Build GRCh38
rs750026746 Chr.3: 44570829-44570833 on Build GRCh38
rs370821218 Chr.3: 44571057-44571057 on Build GRCh38
rs757954350 Chr.3: 44570916-44570916 on Build GRCh38
rs772162577 Chr.3: 44570970-44570970 on Build GRCh38
rs764323983 Chr.3: 44570977-44570977 on Build GRCh38
rs560147920 Chr.3: 44571104-44571104 on Build GRCh38
rs754217707 Chr.3: 44570979-44570979 on Build GRCh38
rs770788709 Chr.3: 44571080-44571080 on Build GRCh38
rs954460667 Chr.3: 44571067-44571067 on Build GRCh38
rs770327284 Chr.3: 44570745-44570745 on Build GRCh38
rs748871190 Chr.3: 44570742-44570742 on Build GRCh38
rs760811754 Chr.3: 44570976-44570976 on Build GRCh38
rs764160901 Chr.3: 44570765-44570765 on Build GRCh38
rs765888405 Chr.3: 44570990-44570990 on Build GRCh38
rs750986930 Chr.3: 44570994-44570994 on Build GRCh38
rs769789425 Chr.3: 44570948-44570948 on Build GRCh38
rs771910702 Chr.3: 44570825-44570825 on Build GRCh38
rs144800158 Chr.3: 44570794-44570794 on Build GRCh38
rs761656524 Chr.3: 44570844-44570844 on Build GRCh38
rs150218193 Chr.3: 44571121-44571121 on Build GRCh38
rs746703747 Chr.3: 44570852-44570852 on Build GRCh38
rs779554345 Chr.3: 44570904-44570904 on Build GRCh38
rs953456867 Chr.3: 44571048-44571048 on Build GRCh38
rs777496747 Chr.3: 44571056-44571056 on Build GRCh38
rs756701074 Chr.3: 44570714-44570714 on Build GRCh38
rs147536772 Chr.3: 44570671-44570671 on Build GRCh38
rs200003223 Chr.3: 44570812-44570812 on Build GRCh38
rs1032197266 Chr.3: 44571046-44571046 on Build GRCh38
rs527328353 Chr.3: 44570920-44570920 on Build GRCh38
rs746668832 Chr.3: 44570930-44570930 on Build GRCh38
rs1014979929 Chr.3: 44570980-44570980 on Build GRCh38
rs768356790 Chr.3: 44570839-44570839 on Build GRCh38
rs769552916 Chr.3: 44571044-44571044 on Build GRCh38
rs750023286 Chr.3: 44570906-44570906 on Build GRCh38
rs752895358 Chr.3: 44570880-44570880 on Build GRCh38
rs767713061 Chr.3: 44570803-44570803 on Build GRCh38
rs746743985 Chr.3: 44570956-44570956 on Build GRCh38
rs775122975 Chr.3: 44570840-44570840 on Build GRCh38
rs574206888 Chr.3: 44570795-44570795 on Build GRCh38
rs923059537 Chr.3: 44571060-44571060 on Build GRCh38
rs780893198 Chr.3: 44571018-44571018 on Build GRCh38
rs763882793 Chr.3: 44570672-44570672 on Build GRCh38
rs545346916 Chr.3: 44570829-44570829 on Build GRCh38

View Full Product Details