Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112836447-112837046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1B
Entrez Gene ID
3553
Gene Name
interleukin 1 beta
Gene Aliases
IL-1, IL1-BETA, IL1F2
UniGene
Hs.126256
Gene Chromosome Location
Chr.2: 112829758-112836842 on Build GRCh38
Transcripts
NM_000576.2 XM_017003988.1

SNP Detail

SNP ID Chromosome Location
rs1133558 Chr.2: 112836731-112836731 on Build GRCh38
rs145667254 Chr.2: 112836613-112836614 on Build GRCh38
rs886632992 Chr.2: 112836887-112836887 on Build GRCh38
rs910945554 Chr.2: 112836738-112836738 on Build GRCh38
rs973796077 Chr.2: 112836819-112836819 on Build GRCh38
rs775970087 Chr.2: 112836970-112836970 on Build GRCh38
rs189808596 Chr.2: 112836481-112836481 on Build GRCh38
rs56302604 Chr.2: 112836595-112836595 on Build GRCh38
rs932053737 Chr.2: 112836833-112836833 on Build GRCh38
rs992663166 Chr.2: 112836798-112836798 on Build GRCh38
rs999420182 Chr.2: 112837015-112837015 on Build GRCh38
rs74579367 Chr.2: 112836916-112836916 on Build GRCh38
rs749558279 Chr.2: 112837003-112837003 on Build GRCh38
rs1051032471 Chr.2: 112836614-112836619 on Build GRCh38
rs545925461 Chr.2: 112836613-112836613 on Build GRCh38
rs1047893156 Chr.2: 112836479-112836479 on Build GRCh38
rs182185350 Chr.2: 112836614-112836614 on Build GRCh38
rs907898566 Chr.2: 112836799-112836799 on Build GRCh38
rs1143627 Chr.2: 112836810-112836810 on Build GRCh38
rs1143628 Chr.2: 112836701-112836701 on Build GRCh38
rs1009121969 Chr.2: 112836642-112836642 on Build GRCh38
rs770127204 Chr.2: 112836851-112836851 on Build GRCh38
rs1799916 Chr.2: 112836740-112836740 on Build GRCh38
rs866837107 Chr.2: 112836935-112836935 on Build GRCh38
rs1012208559 Chr.2: 112836978-112836978 on Build GRCh38
rs977969008 Chr.2: 112836640-112836640 on Build GRCh38
rs549858786 Chr.2: 112836807-112836807 on Build GRCh38
rs1022285047 Chr.2: 112837002-112837002 on Build GRCh38
rs1035094889 Chr.2: 112836522-112836522 on Build GRCh38
rs4986962 Chr.2: 112836846-112836846 on Build GRCh38
rs892711079 Chr.2: 112836955-112836955 on Build GRCh38
rs561088583 Chr.2: 112836778-112836778 on Build GRCh38
rs115403599 Chr.2: 112836754-112836754 on Build GRCh38
rs551919710 Chr.2: 112837019-112837019 on Build GRCh38
rs973694070 Chr.2: 112836767-112836767 on Build GRCh38
rs1021403628 Chr.2: 112836715-112836715 on Build GRCh38
rs1133555 Chr.2: 112836760-112836760 on Build GRCh38
rs1052660224 Chr.2: 112836927-112836927 on Build GRCh38
rs534454119 Chr.2: 112836661-112836662 on Build GRCh38
rs372973976 Chr.2: 112836614-112836615 on Build GRCh38
rs56142111 Chr.2: 112836585-112836585 on Build GRCh38
rs920731805 Chr.2: 112836820-112836820 on Build GRCh38
rs781566483 Chr.2: 112836741-112836741 on Build GRCh38
rs967904099 Chr.2: 112836637-112836637 on Build GRCh38
rs1012175661 Chr.2: 112836570-112836570 on Build GRCh38
rs56019144 Chr.2: 112836573-112836573 on Build GRCh38
rs895168900 Chr.2: 112836527-112836527 on Build GRCh38
rs939524320 Chr.2: 112836894-112836894 on Build GRCh38
rs955214163 Chr.2: 112836694-112836694 on Build GRCh38
rs932194530 Chr.2: 112836606-112836609 on Build GRCh38
rs577256798 Chr.2: 112836521-112836521 on Build GRCh38
rs1030829996 Chr.2: 112836689-112836689 on Build GRCh38
rs557132984 Chr.2: 112836520-112836520 on Build GRCh38
rs1034588398 Chr.2: 112836769-112836769 on Build GRCh38
rs1019294297 Chr.2: 112836643-112836643 on Build GRCh38
rs3917347 Chr.2: 112836628-112836629 on Build GRCh38
rs529869449 Chr.2: 112836785-112836785 on Build GRCh38

View Full Product Details