Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 199271315-199271904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SATB2
Entrez Gene ID
23314
Gene Name
SATB homeobox 2
Gene Aliases
GLSS
UniGene
Hs.516617
Gene Chromosome Location
Chr.2: 199269500-199471266 on Build GRCh38
Transcripts
NM_001172509.1 NM_001172517.1 NM_015265.3 XM_005246396.2 XM_006712372.2 XM_011510840.2 XM_017003656.1

SNP Detail

SNP ID Chromosome Location
rs957588292 Chr.2: 199271372-199271372 on Build GRCh38
rs559471414 Chr.2: 199271538-199271538 on Build GRCh38
rs910728368 Chr.2: 199271681-199271681 on Build GRCh38
rs181138294 Chr.2: 199271437-199271437 on Build GRCh38
rs941099884 Chr.2: 199271820-199271820 on Build GRCh38
rs35758003 Chr.2: 199271561-199271561 on Build GRCh38
rs568807856 Chr.2: 199271439-199271439 on Build GRCh38
rs35679091 Chr.2: 199271368-199271369 on Build GRCh38
rs1033130436 Chr.2: 199271642-199271642 on Build GRCh38
rs894329615 Chr.2: 199271850-199271850 on Build GRCh38
rs772055529 Chr.2: 199271774-199271774 on Build GRCh38
rs1054713608 Chr.2: 199271683-199271683 on Build GRCh38
rs114405014 Chr.2: 199271580-199271580 on Build GRCh38
rs961620761 Chr.2: 199271371-199271371 on Build GRCh38
rs1040569490 Chr.2: 199271473-199271473 on Build GRCh38
rs949844723 Chr.2: 199271855-199271855 on Build GRCh38
rs949144340 Chr.2: 199271592-199271592 on Build GRCh38
rs397987299 Chr.2: 199271572-199271572 on Build GRCh38
rs761048331 Chr.2: 199271400-199271400 on Build GRCh38
rs551588093 Chr.2: 199271811-199271811 on Build GRCh38
rs1055640386 Chr.2: 199271590-199271593 on Build GRCh38
rs1046840493 Chr.2: 199271554-199271554 on Build GRCh38
rs745498887 Chr.2: 199271717-199271717 on Build GRCh38
rs929693092 Chr.2: 199271741-199271741 on Build GRCh38
rs531993363 Chr.2: 199271354-199271354 on Build GRCh38
rs60420320 Chr.2: 199271483-199271484 on Build GRCh38
rs891230761 Chr.2: 199271809-199271809 on Build GRCh38
rs186107760 Chr.2: 199271596-199271596 on Build GRCh38
rs957447534 Chr.2: 199271677-199271677 on Build GRCh38
rs1008519354 Chr.2: 199271353-199271353 on Build GRCh38
rs1026835905 Chr.2: 199271409-199271409 on Build GRCh38
rs1026866985 Chr.2: 199271876-199271876 on Build GRCh38
rs1055526749 Chr.2: 199271832-199271832 on Build GRCh38
rs924815961 Chr.2: 199271636-199271636 on Build GRCh38
rs932676187 Chr.2: 199271736-199271736 on Build GRCh38
rs77468147 Chr.2: 199271842-199271842 on Build GRCh38
rs910842929 Chr.2: 199271449-199271449 on Build GRCh38
rs1049751107 Chr.2: 199271770-199271770 on Build GRCh38
rs1024617275 Chr.2: 199271626-199271626 on Build GRCh38
rs6435006 Chr.2: 199271860-199271860 on Build GRCh38
rs150702506 Chr.2: 199271411-199271415 on Build GRCh38
rs967426810 Chr.2: 199271631-199271631 on Build GRCh38
rs79165231 Chr.2: 199271573-199271573 on Build GRCh38
rs535960709 Chr.2: 199271569-199271569 on Build GRCh38
rs775655256 Chr.2: 199271484-199271484 on Build GRCh38
rs929613421 Chr.2: 199271494-199271494 on Build GRCh38
rs995261866 Chr.2: 199271874-199271874 on Build GRCh38
rs1047168993 Chr.2: 199271767-199271768 on Build GRCh38
rs778715719 Chr.2: 199271678-199271678 on Build GRCh38
rs888186447 Chr.2: 199271562-199271562 on Build GRCh38
rs1037497108 Chr.2: 199271851-199271851 on Build GRCh38
rs982044703 Chr.2: 199271620-199271620 on Build GRCh38
rs190042308 Chr.2: 199271608-199271608 on Build GRCh38
rs944961989 Chr.2: 199271471-199271472 on Build GRCh38
rs78418954 Chr.2: 199271572-199271572 on Build GRCh38
rs894282114 Chr.2: 199271599-199271599 on Build GRCh38
rs753313213 Chr.2: 199271486-199271486 on Build GRCh38
rs977548653 Chr.2: 199271345-199271345 on Build GRCh38
rs976328391 Chr.2: 199271708-199271708 on Build GRCh38
rs973974701 Chr.2: 199271408-199271408 on Build GRCh38

View Full Product Details