Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69191249-69191746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD2
Entrez Gene ID
55218
Gene Name
exonuclease 3'-5' domain containing 2
Gene Aliases
C14orf114, EXDL2
UniGene
Hs.533878
Gene Chromosome Location
Chr.14: 69191477-69244020 on Build GRCh38
Transcripts
NM_001193360.1 NM_001193361.1 NM_018199.3 NR_034164.1 XM_017021421.1

Gene Symbol
LOC105370550
Entrez Gene ID
105370550
Gene Name
uncharacterized LOC105370550
Gene Aliases
-
UniGene
Hs.667208
Gene Chromosome Location
Chr.14: 69183018-69260567 on Build GRCh38
Transcripts
NM_001193360.1 NM_001193361.1 NM_018199.3 NR_034164.1 XM_017021421.1

SNP Detail

SNP ID Chromosome Location
rs1046305400 Chr.14: 69191727-69191727 on Build GRCh38
rs373309131 Chr.14: 69191705-69191705 on Build GRCh38
rs377004913 Chr.14: 69191394-69191394 on Build GRCh38
rs990468474 Chr.14: 69191479-69191479 on Build GRCh38
rs889796959 Chr.14: 69191715-69191715 on Build GRCh38
rs1041358147 Chr.14: 69191386-69191386 on Build GRCh38
rs920473532 Chr.14: 69191524-69191524 on Build GRCh38
rs974081959 Chr.14: 69191516-69191516 on Build GRCh38
rs893235042 Chr.14: 69191371-69191371 on Build GRCh38
rs144025537 Chr.14: 69191572-69191572 on Build GRCh38
rs894732921 Chr.14: 69191454-69191454 on Build GRCh38
rs931778857 Chr.14: 69191349-69191349 on Build GRCh38
rs558489081 Chr.14: 69191509-69191509 on Build GRCh38
rs546096928 Chr.14: 69191293-69191293 on Build GRCh38
rs1000387723 Chr.14: 69191411-69191411 on Build GRCh38
rs1054128410 Chr.14: 69191689-69191689 on Build GRCh38
rs1004328834 Chr.14: 69191467-69191467 on Build GRCh38
rs1041472802 Chr.14: 69191626-69191626 on Build GRCh38
rs916237039 Chr.14: 69191494-69191494 on Build GRCh38
rs148405864 Chr.14: 69191440-69191440 on Build GRCh38
rs1046226200 Chr.14: 69191301-69191301 on Build GRCh38
rs1008452122 Chr.14: 69191430-69191430 on Build GRCh38
rs565915431 Chr.14: 69191550-69191550 on Build GRCh38
rs572340537 Chr.14: 69191291-69191291 on Build GRCh38
rs145437069 Chr.14: 69191387-69191387 on Build GRCh38
rs902888252 Chr.14: 69191401-69191401 on Build GRCh38
rs865885275 Chr.14: 69191478-69191478 on Build GRCh38
rs191277584 Chr.14: 69191511-69191511 on Build GRCh38
rs1053180894 Chr.14: 69191356-69191356 on Build GRCh38
rs947460514 Chr.14: 69191659-69191659 on Build GRCh38
rs551649514 Chr.14: 69191622-69191622 on Build GRCh38
rs369614069 Chr.14: 69191571-69191571 on Build GRCh38
rs1028749987 Chr.14: 69191390-69191390 on Build GRCh38
rs940574581 Chr.14: 69191336-69191336 on Build GRCh38
rs771473646 Chr.14: 69191551-69191551 on Build GRCh38
rs961510779 Chr.14: 69191448-69191448 on Build GRCh38
rs1011575517 Chr.14: 69191384-69191384 on Build GRCh38
rs944500362 Chr.14: 69191368-69191368 on Build GRCh38
rs1050280623 Chr.14: 69191362-69191362 on Build GRCh38
rs369373648 Chr.14: 69191475-69191475 on Build GRCh38
rs944448205 Chr.14: 69191583-69191583 on Build GRCh38
rs569762724 Chr.14: 69191678-69191678 on Build GRCh38
rs545357670 Chr.14: 69191718-69191718 on Build GRCh38
rs916491543 Chr.14: 69191283-69191283 on Build GRCh38
rs1024514135 Chr.14: 69191458-69191458 on Build GRCh38
rs754278261 Chr.14: 69191278-69191278 on Build GRCh38
rs890003813 Chr.14: 69191414-69191414 on Build GRCh38
rs1013460238 Chr.14: 69191455-69191455 on Build GRCh38
rs567056126 Chr.14: 69191407-69191409 on Build GRCh38
rs1049725969 Chr.14: 69191684-69191684 on Build GRCh38
rs924385357 Chr.14: 69191662-69191662 on Build GRCh38
rs971461789 Chr.14: 69191465-69191465 on Build GRCh38
rs902367128 Chr.14: 69191307-69191307 on Build GRCh38
rs543196609 Chr.14: 69191436-69191436 on Build GRCh38
rs973339675 Chr.14: 69191341-69191341 on Build GRCh38
rs890162748 Chr.14: 69191364-69191364 on Build GRCh38
rs972897390 Chr.14: 69191451-69191451 on Build GRCh38
rs935642687 Chr.14: 69191683-69191683 on Build GRCh38
rs866953393 Chr.14: 69191334-69191334 on Build GRCh38
rs185981201 Chr.14: 69191347-69191347 on Build GRCh38
rs4899285 Chr.14: 69191723-69191723 on Build GRCh38
rs894713821 Chr.14: 69191709-69191709 on Build GRCh38

View Full Product Details