Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99218464-99219013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC23
Entrez Gene ID
64927
Gene Name
tetratricopeptide repeat domain 23
Gene Aliases
HCC-8
UniGene
Hs.513195
Gene Chromosome Location
Chr.15: 99136323-99251226 on Build GRCh38
Transcripts
NM_001288615.1 NM_001288616.1 NR_109947.1 XM_011521927.2 XM_011521929.1 XM_011521930.1 XM_011521931.1 XM_011521932.1 XM_011521933.1 XM_011521934.1 XM_011521935.1 XM_011521936.1 XM_011521937.1 XM_011521939.1 XM_011521940.2 XM_011521941.2 XM_011521943.2 XM_011521944.2 XM_011521945.2 XM_017022509.1 XM_017022510.1 XM_017022511.1 XM_017022512.1 XM_017022513.1 XM_017022514.1 XM_017022515.1 XM_017022516.1 XM_017022517.1 XM_017022518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7223773 c.522A>T p.G174G Chr.15: 99218647-99218647 on Build GRCh38
4737126 c.354C>T p.L118L Chr.15: 99218999-99218999 on Build GRCh38
5758618 c.455+1G>T p.? Chr.15: 99218897-99218897 on Build GRCh38
3505656 c.474G>A p.E158E Chr.15: 99218695-99218695 on Build GRCh38
6811030 c.458T>C p.F153S Chr.15: 99218711-99218711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036272304 Chr.15: 99218916-99218916 on Build GRCh38
rs745817434 Chr.15: 99218921-99218921 on Build GRCh38
rs771919342 Chr.15: 99218679-99218679 on Build GRCh38
rs757758331 Chr.15: 99218639-99218639 on Build GRCh38
rs760630999 Chr.15: 99218719-99218719 on Build GRCh38
rs776493308 Chr.15: 99218914-99218914 on Build GRCh38
rs541024445 Chr.15: 99218840-99218840 on Build GRCh38
rs772970039 Chr.15: 99218714-99218714 on Build GRCh38
rs970750478 Chr.15: 99218900-99218900 on Build GRCh38
rs188482812 Chr.15: 99218758-99218758 on Build GRCh38
rs1037009482 Chr.15: 99218843-99218843 on Build GRCh38
rs781743487 Chr.15: 99218555-99218555 on Build GRCh38
rs747783748 Chr.15: 99218678-99218678 on Build GRCh38
rs938020956 Chr.15: 99218832-99218832 on Build GRCh38
rs760889418 Chr.15: 99218964-99218964 on Build GRCh38
rs770375288 Chr.15: 99218781-99218781 on Build GRCh38
rs201945611 Chr.15: 99218610-99218610 on Build GRCh38
rs570698633 Chr.15: 99218793-99218793 on Build GRCh38
rs532474364 Chr.15: 99218976-99218976 on Build GRCh38
rs778479797 Chr.15: 99218890-99218890 on Build GRCh38
rs773190522 Chr.15: 99218596-99218596 on Build GRCh38
rs749016073 Chr.15: 99218562-99218562 on Build GRCh38
rs1026163750 Chr.15: 99218623-99218623 on Build GRCh38
rs764225935 Chr.15: 99218750-99218750 on Build GRCh38
rs766753255 Chr.15: 99218972-99218972 on Build GRCh38
rs751971098 Chr.15: 99218632-99218632 on Build GRCh38
rs369938828 Chr.15: 99218853-99218853 on Build GRCh38
rs371605515 Chr.15: 99218509-99218509 on Build GRCh38
rs116264426 Chr.15: 99218924-99218924 on Build GRCh38
rs921878289 Chr.15: 99218736-99218736 on Build GRCh38
rs139126638 Chr.15: 99218652-99218652 on Build GRCh38
rs1027220833 Chr.15: 99218885-99218885 on Build GRCh38
rs377161868 Chr.15: 99218648-99218648 on Build GRCh38
rs988420446 Chr.15: 99218974-99218974 on Build GRCh38
rs201175349 Chr.15: 99218770-99218770 on Build GRCh38
rs541924360 Chr.15: 99218857-99218857 on Build GRCh38
rs978439621 Chr.15: 99218994-99218994 on Build GRCh38
rs775108886 Chr.15: 99218492-99218492 on Build GRCh38
rs749798364 Chr.15: 99218841-99218841 on Build GRCh38
rs766726191 Chr.15: 99218872-99218872 on Build GRCh38
rs372195443 Chr.15: 99218546-99218546 on Build GRCh38
rs143602155 Chr.15: 99218630-99218630 on Build GRCh38
rs970238434 Chr.15: 99218552-99218552 on Build GRCh38
rs370447505 Chr.15: 99218896-99218896 on Build GRCh38
rs926615790 Chr.15: 99218817-99218817 on Build GRCh38
rs768358654 Chr.15: 99218655-99218655 on Build GRCh38
rs115374662 Chr.15: 99218558-99218558 on Build GRCh38
rs947278806 Chr.15: 99218733-99218733 on Build GRCh38
rs762886940 Chr.15: 99218749-99218749 on Build GRCh38
rs752165280 Chr.15: 99218988-99218988 on Build GRCh38
rs778222518 Chr.15: 99218543-99218543 on Build GRCh38
rs200167617 Chr.15: 99218645-99218645 on Build GRCh38
rs553848267 Chr.15: 99218640-99218640 on Build GRCh38
rs374117965 Chr.15: 99218875-99218875 on Build GRCh38
rs887652256 Chr.15: 99218858-99218858 on Build GRCh38
rs774229321 Chr.15: 99218579-99218579 on Build GRCh38
rs768804200 Chr.15: 99218564-99218565 on Build GRCh38
rs955635837 Chr.15: 99218760-99218760 on Build GRCh38
rs138663596 Chr.15: 99218938-99218938 on Build GRCh38
rs760600439 Chr.15: 99218609-99218609 on Build GRCh38
rs1042943809 Chr.15: 99218735-99218735 on Build GRCh38
rs774546992 Chr.15: 99218937-99218937 on Build GRCh38
rs141620949 Chr.15: 99218902-99218902 on Build GRCh38
rs565174983 Chr.15: 99218963-99218963 on Build GRCh38
rs1043853120 Chr.15: 99218812-99218812 on Build GRCh38
rs111836146 Chr.15: 99218803-99218803 on Build GRCh38
rs753401451 Chr.15: 99218874-99218874 on Build GRCh38
rs775645692 Chr.15: 99218742-99218742 on Build GRCh38
rs764750052 Chr.15: 99218978-99218978 on Build GRCh38
rs745751586 Chr.15: 99218539-99218539 on Build GRCh38
rs369025962 Chr.15: 99218570-99218570 on Build GRCh38
rs549762502 Chr.15: 99218496-99218496 on Build GRCh38
rs770914819 Chr.15: 99218909-99218909 on Build GRCh38
rs142730025 Chr.15: 99218943-99218943 on Build GRCh38
rs192800554 Chr.15: 99218806-99218806 on Build GRCh38
rs746739734 Chr.15: 99218907-99218907 on Build GRCh38
rs763163153 Chr.15: 99218957-99218957 on Build GRCh38
rs183073604 Chr.15: 99218491-99218491 on Build GRCh38
rs770734195 Chr.15: 99218724-99218724 on Build GRCh38
rs772009385 Chr.15: 99218588-99218588 on Build GRCh38
rs151021606 Chr.15: 99218989-99218989 on Build GRCh38
rs1001676750 Chr.15: 99218903-99218903 on Build GRCh38
rs577161045 Chr.15: 99218826-99218826 on Build GRCh38
rs897171665 Chr.15: 99218851-99218851 on Build GRCh38
rs1056489914 Chr.15: 99218833-99218833 on Build GRCh38
rs995776125 Chr.15: 99218550-99218550 on Build GRCh38
rs200054433 Chr.15: 99218551-99218551 on Build GRCh38
rs1008730084 Chr.15: 99218835-99218835 on Build GRCh38
rs144143144 Chr.15: 99218854-99218854 on Build GRCh38
rs778371332 Chr.15: 99218660-99218660 on Build GRCh38
rs767803690 Chr.15: 99218945-99218945 on Build GRCh38

View Full Product Details