Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71732678-71733235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1G1
Entrez Gene ID
164
Gene Name
adaptor related protein complex 1 gamma 1 subunit
Gene Aliases
ADTG, CLAPG1
UniGene
Hs.461253
Gene Chromosome Location
Chr.16: 71729000-71809073 on Build GRCh38
Transcripts
NM_001030007.1 NM_001128.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4784355 c.2393G>A p.R798Q Chr.16: 71733143-71733143 on Build GRCh38
6307614 c.2422C>A p.Q808K Chr.16: 71733105-71733105 on Build GRCh38
4784354 c.2453G>A p.R818Q Chr.16: 71733143-71733143 on Build GRCh38
5654318 c.2375T>G p.L792R Chr.16: 71733152-71733152 on Build GRCh38
6307613 c.2431C>A p.Q811K Chr.16: 71733105-71733105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056905240 Chr.16: 71733046-71733046 on Build GRCh38
rs750620151 Chr.16: 71732751-71732751 on Build GRCh38
rs898155602 Chr.16: 71732710-71732710 on Build GRCh38
rs749801429 Chr.16: 71733172-71733172 on Build GRCh38
rs899085166 Chr.16: 71733118-71733118 on Build GRCh38
rs118111752 Chr.16: 71733162-71733162 on Build GRCh38
rs1022079457 Chr.16: 71732811-71732811 on Build GRCh38
rs980208886 Chr.16: 71732821-71732821 on Build GRCh38
rs919620204 Chr.16: 71732953-71732953 on Build GRCh38
rs780767852 Chr.16: 71733169-71733169 on Build GRCh38
rs781693456 Chr.16: 71733139-71733139 on Build GRCh38
rs759024708 Chr.16: 71732786-71732786 on Build GRCh38
rs753761796 Chr.16: 71733100-71733100 on Build GRCh38
rs761467938 Chr.16: 71733057-71733057 on Build GRCh38
rs368796987 Chr.16: 71733109-71733109 on Build GRCh38
rs747784229 Chr.16: 71733184-71733184 on Build GRCh38
rs902882345 Chr.16: 71732976-71732976 on Build GRCh38
rs535337952 Chr.16: 71732801-71732801 on Build GRCh38
rs1038281123 Chr.16: 71732952-71732952 on Build GRCh38
rs563023607 Chr.16: 71732966-71732966 on Build GRCh38
rs138023626 Chr.16: 71732748-71732748 on Build GRCh38
rs752152814 Chr.16: 71732912-71732912 on Build GRCh38
rs574297350 Chr.16: 71732779-71732779 on Build GRCh38
rs778435079 Chr.16: 71733178-71733178 on Build GRCh38
rs956450644 Chr.16: 71732790-71732790 on Build GRCh38
rs530311104 Chr.16: 71732970-71732970 on Build GRCh38
rs772947407 Chr.16: 71733070-71733070 on Build GRCh38
rs577575699 Chr.16: 71732920-71732920 on Build GRCh38
rs555039498 Chr.16: 71732810-71732810 on Build GRCh38
rs912705603 Chr.16: 71732871-71732871 on Build GRCh38
rs780246974 Chr.16: 71732764-71732764 on Build GRCh38
rs1012270333 Chr.16: 71733067-71733067 on Build GRCh38
rs544927536 Chr.16: 71732963-71732963 on Build GRCh38
rs1046350 Chr.16: 71732731-71732731 on Build GRCh38
rs761947285 Chr.16: 71732719-71732719 on Build GRCh38
rs768203007 Chr.16: 71733047-71733047 on Build GRCh38
rs970034216 Chr.16: 71732820-71732820 on Build GRCh38
rs184500521 Chr.16: 71732859-71732859 on Build GRCh38
rs768442011 Chr.16: 71733032-71733033 on Build GRCh38
rs932301208 Chr.16: 71732958-71732958 on Build GRCh38
rs147519448 Chr.16: 71733061-71733061 on Build GRCh38
rs764166650 Chr.16: 71733126-71733126 on Build GRCh38
rs990414569 Chr.16: 71732791-71732791 on Build GRCh38
rs768058576 Chr.16: 71733175-71733175 on Build GRCh38
rs1021860243 Chr.16: 71733091-71733091 on Build GRCh38
rs763008013 Chr.16: 71733115-71733115 on Build GRCh38
rs755459557 Chr.16: 71732935-71732935 on Build GRCh38
rs573292602 Chr.16: 71732827-71732827 on Build GRCh38
rs766353180 Chr.16: 71733099-71733099 on Build GRCh38
rs375099704 Chr.16: 71733199-71733199 on Build GRCh38
rs758894758 Chr.16: 71732761-71732761 on Build GRCh38
rs773944366 Chr.16: 71733053-71733053 on Build GRCh38
rs781714640 Chr.16: 71733143-71733143 on Build GRCh38
rs527422540 Chr.16: 71733075-71733075 on Build GRCh38
rs939970704 Chr.16: 71732929-71732929 on Build GRCh38
rs771747624 Chr.16: 71733189-71733189 on Build GRCh38
rs565304077 Chr.16: 71732869-71732869 on Build GRCh38
rs998654102 Chr.16: 71732718-71732718 on Build GRCh38
rs992369333 Chr.16: 71732886-71732886 on Build GRCh38
rs960544621 Chr.16: 71732878-71732878 on Build GRCh38
rs974048079 Chr.16: 71732809-71732809 on Build GRCh38
rs200448545 Chr.16: 71733031-71733031 on Build GRCh38
rs756528630 Chr.16: 71733168-71733168 on Build GRCh38
rs1008357388 Chr.16: 71732972-71732972 on Build GRCh38
rs996583709 Chr.16: 71732714-71732714 on Build GRCh38
rs757454113 Chr.16: 71733142-71733142 on Build GRCh38
rs113530943 Chr.16: 71732980-71732980 on Build GRCh38
rs1042741484 Chr.16: 71732975-71732975 on Build GRCh38

View Full Product Details