Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 79206813-79207399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP9B
Entrez Gene ID
374868
Gene Name
ATPase phospholipid transporting 9B (putative)
Gene Aliases
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
UniGene
Hs.465475
Gene Chromosome Location
Chr.18: 79069275-79378391 on Build GRCh38
Transcripts
NM_001306085.1 NM_198531.4 XM_011525963.2 XM_011525964.2 XM_011525965.2 XM_011525966.2 XM_011525971.2 XM_011525972.2 XM_011525973.2 XM_011525974.2 XM_017025726.1 XM_017025727.1 XM_017025728.1 XM_017025729.1 XM_017025730.1 XM_017025731.1 XM_017025732.1 XM_017025733.1 XM_017025734.1 XM_017025735.1 XM_017025736.1 XM_017025737.1 XM_017025738.1 XM_017025739.1 XM_017025740.1 XM_017025741.1 XM_017025742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
990070 c.989G>A p.S330N Chr.18: 79206971-79206971 on Build GRCh38
6898148 c.1001C>T p.T334I Chr.18: 79206983-79206983 on Build GRCh38
990069 c.983G>A p.S328N Chr.18: 79206965-79206965 on Build GRCh38
6279233 c.1000A>G p.T334A Chr.18: 79206982-79206982 on Build GRCh38
1189731 c.983G>T p.S328I Chr.18: 79206965-79206965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758450595 Chr.18: 79206948-79206948 on Build GRCh38
rs948402802 Chr.18: 79207210-79207210 on Build GRCh38
rs954836385 Chr.18: 79207144-79207144 on Build GRCh38
rs150988101 Chr.18: 79207013-79207013 on Build GRCh38
rs984763328 Chr.18: 79206869-79206869 on Build GRCh38
rs767057337 Chr.18: 79206989-79206989 on Build GRCh38
rs764124977 Chr.18: 79207045-79207045 on Build GRCh38
rs961282889 Chr.18: 79207037-79207037 on Build GRCh38
rs769393044 Chr.18: 79206892-79206892 on Build GRCh38
rs1017657917 Chr.18: 79207114-79207114 on Build GRCh38
rs969682689 Chr.18: 79206866-79206866 on Build GRCh38
rs924537219 Chr.18: 79207187-79207187 on Build GRCh38
rs200201608 Chr.18: 79207050-79207050 on Build GRCh38
rs1016262351 Chr.18: 79206875-79206875 on Build GRCh38
rs923415204 Chr.18: 79206896-79206896 on Build GRCh38
rs780686047 Chr.18: 79207033-79207036 on Build GRCh38
rs761188722 Chr.18: 79206921-79206921 on Build GRCh38
rs1036858855 Chr.18: 79207231-79207231 on Build GRCh38
rs779107923 Chr.18: 79207218-79207218 on Build GRCh38
rs550436062 Chr.18: 79207111-79207111 on Build GRCh38
rs776882241 Chr.18: 79206981-79206981 on Build GRCh38
rs796377744 Chr.18: 79207141-79207141 on Build GRCh38
rs755468535 Chr.18: 79207139-79207139 on Build GRCh38
rs916954486 Chr.18: 79207223-79207223 on Build GRCh38
rs140482398 Chr.18: 79207033-79207033 on Build GRCh38
rs777169273 Chr.18: 79207024-79207024 on Build GRCh38
rs754447079 Chr.18: 79206929-79206929 on Build GRCh38
rs1054117101 Chr.18: 79206943-79206943 on Build GRCh38
rs766822305 Chr.18: 79206923-79206923 on Build GRCh38
rs938450970 Chr.18: 79207222-79207222 on Build GRCh38
rs1042976826 Chr.18: 79206951-79206951 on Build GRCh38
rs113989436 Chr.18: 79206969-79206969 on Build GRCh38
rs371402254 Chr.18: 79207026-79207026 on Build GRCh38
rs560676316 Chr.18: 79206993-79206993 on Build GRCh38
rs767233188 Chr.18: 79207023-79207023 on Build GRCh38
rs191700008 Chr.18: 79206991-79206991 on Build GRCh38
rs765681219 Chr.18: 79207029-79207029 on Build GRCh38
rs962064386 Chr.18: 79206890-79206890 on Build GRCh38
rs763657382 Chr.18: 79206906-79206906 on Build GRCh38
rs575066417 Chr.18: 79206970-79206970 on Build GRCh38
rs147002593 Chr.18: 79207272-79207272 on Build GRCh38
rs910367136 Chr.18: 79207158-79207158 on Build GRCh38
rs765738057 Chr.18: 79206940-79206940 on Build GRCh38
rs570810112 Chr.18: 79207245-79207245 on Build GRCh38
rs188823496 Chr.18: 79207308-79207308 on Build GRCh38
rs750750322 Chr.18: 79206913-79206913 on Build GRCh38
rs761536216 Chr.18: 79207000-79207000 on Build GRCh38
rs767528491 Chr.18: 79207211-79207211 on Build GRCh38
rs529831501 Chr.18: 79207232-79207233 on Build GRCh38
rs146565478 Chr.18: 79207157-79207157 on Build GRCh38
rs986211041 Chr.18: 79207098-79207098 on Build GRCh38
rs755880815 Chr.18: 79207053-79207053 on Build GRCh38
rs768209292 Chr.18: 79206995-79206995 on Build GRCh38
rs763393036 Chr.18: 79206884-79206884 on Build GRCh38
rs140379827 Chr.18: 79207254-79207254 on Build GRCh38
rs745771214 Chr.18: 79206886-79206886 on Build GRCh38
rs947260824 Chr.18: 79207159-79207159 on Build GRCh38
rs956420950 Chr.18: 79207310-79207310 on Build GRCh38
rs1024265691 Chr.18: 79206859-79206859 on Build GRCh38
rs966151308 Chr.18: 79207128-79207128 on Build GRCh38
rs774188389 Chr.18: 79206998-79206998 on Build GRCh38
rs752023651 Chr.18: 79207075-79207075 on Build GRCh38
rs757491511 Chr.18: 79207049-79207049 on Build GRCh38
rs146781201 Chr.18: 79207030-79207030 on Build GRCh38
rs749219146 Chr.18: 79207060-79207060 on Build GRCh38
rs755465563 Chr.18: 79206939-79206939 on Build GRCh38
rs186868522 Chr.18: 79206946-79206946 on Build GRCh38
rs566554563 Chr.18: 79207176-79207176 on Build GRCh38
rs149861710 Chr.18: 79206853-79206853 on Build GRCh38
rs1006461943 Chr.18: 79207130-79207130 on Build GRCh38
rs774135193 Chr.18: 79206917-79206917 on Build GRCh38
rs28669806 Chr.18: 79207281-79207281 on Build GRCh38
rs369957141 Chr.18: 79207032-79207032 on Build GRCh38
rs972068677 Chr.18: 79207138-79207138 on Build GRCh38
rs933115705 Chr.18: 79206898-79206898 on Build GRCh38
rs367655880 Chr.18: 79206950-79206950 on Build GRCh38
rs143130204 Chr.18: 79207179-79207179 on Build GRCh38
rs905391768 Chr.18: 79207123-79207123 on Build GRCh38
rs905359277 Chr.18: 79207236-79207236 on Build GRCh38
rs1023816493 Chr.18: 79207115-79207115 on Build GRCh38
rs1030345255 Chr.18: 79207280-79207280 on Build GRCh38
rs1044555728 Chr.18: 79206962-79206962 on Build GRCh38
rs142600945 Chr.18: 79206973-79206973 on Build GRCh38
rs895568416 Chr.18: 79207071-79207071 on Build GRCh38
rs1017675993 Chr.18: 79207291-79207291 on Build GRCh38
rs374816696 Chr.18: 79206893-79206893 on Build GRCh38
rs948486363 Chr.18: 79207233-79207233 on Build GRCh38
rs1037924641 Chr.18: 79207253-79207253 on Build GRCh38
rs148257468 Chr.18: 79207235-79207235 on Build GRCh38
rs762485029 Chr.18: 79206905-79206905 on Build GRCh38
rs184327585 Chr.18: 79207134-79207134 on Build GRCh38
rs995418722 Chr.18: 79207232-79207237 on Build GRCh38
rs990387130 Chr.18: 79207316-79207316 on Build GRCh38
rs536158169 Chr.18: 79207290-79207290 on Build GRCh38
rs779923776 Chr.18: 79207059-79207059 on Build GRCh38
rs751825810 Chr.18: 79207047-79207047 on Build GRCh38
rs906953971 Chr.18: 79207217-79207217 on Build GRCh38
rs1007982744 Chr.18: 79207259-79207259 on Build GRCh38
rs144211563 Chr.18: 79207133-79207133 on Build GRCh38
rs952167103 Chr.18: 79207089-79207089 on Build GRCh38
rs773759600 Chr.18: 79206879-79206879 on Build GRCh38
rs147600495 Chr.18: 79206873-79206873 on Build GRCh38

View Full Product Details