Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42296944-42297492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAFAH1B3
Entrez Gene ID
5050
Gene Name
platelet activating factor acetylhydrolase 1b catalytic subunit 3
Gene Aliases
PAFAHG
UniGene
Hs.466831
Gene Chromosome Location
Chr.19: 42297033-42302800 on Build GRCh38
Transcripts
NM_001145939.1 NM_001145940.1 NM_002573.3 XM_017026846.1 XM_017026847.1 XM_017026848.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4581118 c.688G>A p.A230T Chr.19: 42297086-42297086 on Build GRCh38
1660270 c.511T>A p.F171I Chr.19: 42297263-42297263 on Build GRCh38
7415848 c.521C>T p.A174V Chr.19: 42297253-42297253 on Build GRCh38
1241539 c.446G>A p.R149Q Chr.19: 42297328-42297328 on Build GRCh38
6317633 c.663C>A p.G221G Chr.19: 42297111-42297111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932202716 Chr.19: 42297075-42297075 on Build GRCh38
rs748929376 Chr.19: 42297376-42297376 on Build GRCh38
rs552766466 Chr.19: 42297034-42297034 on Build GRCh38
rs947635250 Chr.19: 42297152-42297152 on Build GRCh38
rs569009647 Chr.19: 42297296-42297296 on Build GRCh38
rs756687537 Chr.19: 42297046-42297046 on Build GRCh38
rs750490489 Chr.19: 42297344-42297344 on Build GRCh38
rs151099535 Chr.19: 42297306-42297306 on Build GRCh38
rs772476066 Chr.19: 42297221-42297221 on Build GRCh38
rs149072464 Chr.19: 42297291-42297291 on Build GRCh38
rs757242349 Chr.19: 42297282-42297282 on Build GRCh38
rs746347569 Chr.19: 42297095-42297095 on Build GRCh38
rs777799764 Chr.19: 42297272-42297272 on Build GRCh38
rs571001912 Chr.19: 42297040-42297040 on Build GRCh38
rs762308083 Chr.19: 42297249-42297249 on Build GRCh38
rs756193854 Chr.19: 42297094-42297094 on Build GRCh38
rs199558799 Chr.19: 42297181-42297181 on Build GRCh38
rs958860187 Chr.19: 42297300-42297300 on Build GRCh38
rs760479127 Chr.19: 42297325-42297325 on Build GRCh38
rs757764119 Chr.19: 42297044-42297045 on Build GRCh38
rs759759579 Chr.19: 42297098-42297098 on Build GRCh38
rs749898006 Chr.19: 42297138-42297138 on Build GRCh38
rs758920532 Chr.19: 42297352-42297352 on Build GRCh38
rs12429 Chr.19: 42297188-42297188 on Build GRCh38
rs756461132 Chr.19: 42297271-42297271 on Build GRCh38
rs921133234 Chr.19: 42297408-42297408 on Build GRCh38
rs781756687 Chr.19: 42297054-42297056 on Build GRCh38
rs145086603 Chr.19: 42297222-42297222 on Build GRCh38
rs572646376 Chr.19: 42297418-42297418 on Build GRCh38
rs780475032 Chr.19: 42297353-42297353 on Build GRCh38
rs759609760 Chr.19: 42297317-42297317 on Build GRCh38
rs766173888 Chr.19: 42297145-42297145 on Build GRCh38
rs774544378 Chr.19: 42297030-42297030 on Build GRCh38
rs947043354 Chr.19: 42296997-42296997 on Build GRCh38
rs761520518 Chr.19: 42297224-42297224 on Build GRCh38
rs372405011 Chr.19: 42297060-42297060 on Build GRCh38
rs914758641 Chr.19: 42297421-42297422 on Build GRCh38
rs772796445 Chr.19: 42297247-42297247 on Build GRCh38
rs531139156 Chr.19: 42296990-42296990 on Build GRCh38
rs143760547 Chr.19: 42297305-42297305 on Build GRCh38
rs947606209 Chr.19: 42297435-42297435 on Build GRCh38
rs570981778 Chr.19: 42297086-42297086 on Build GRCh38
rs771482720 Chr.19: 42297210-42297210 on Build GRCh38
rs534302050 Chr.19: 42296987-42296988 on Build GRCh38
rs749313761 Chr.19: 42297207-42297207 on Build GRCh38
rs776218743 Chr.19: 42297126-42297126 on Build GRCh38
rs1021817019 Chr.19: 42297329-42297329 on Build GRCh38
rs769423201 Chr.19: 42297229-42297229 on Build GRCh38
rs781602048 Chr.19: 42297355-42297355 on Build GRCh38
rs192553749 Chr.19: 42297379-42297379 on Build GRCh38
rs763835918 Chr.19: 42297411-42297411 on Build GRCh38
rs200573824 Chr.19: 42297157-42297157 on Build GRCh38
rs751376118 Chr.19: 42297148-42297148 on Build GRCh38
rs373974827 Chr.19: 42297385-42297385 on Build GRCh38
rs775528377 Chr.19: 42297387-42297387 on Build GRCh38
rs147763504 Chr.19: 42297087-42297087 on Build GRCh38
rs370064398 Chr.19: 42297334-42297334 on Build GRCh38
rs752552502 Chr.19: 42297321-42297321 on Build GRCh38
rs370017491 Chr.19: 42297133-42297133 on Build GRCh38
rs907892277 Chr.19: 42296976-42296976 on Build GRCh38
rs757472467 Chr.19: 42297337-42297337 on Build GRCh38
rs748830063 Chr.19: 42297068-42297068 on Build GRCh38
rs914789165 Chr.19: 42297139-42297139 on Build GRCh38
rs572765616 Chr.19: 42297213-42297213 on Build GRCh38
rs778929087 Chr.19: 42297339-42297339 on Build GRCh38
rs766373467 Chr.19: 42297251-42297251 on Build GRCh38
rs746770819 Chr.19: 42297067-42297067 on Build GRCh38
rs765182773 Chr.19: 42297048-42297048 on Build GRCh38
rs745316852 Chr.19: 42297384-42297384 on Build GRCh38
rs748427177 Chr.19: 42297167-42297167 on Build GRCh38
rs144947482 Chr.19: 42297252-42297252 on Build GRCh38
rs755709794 Chr.19: 42297017-42297017 on Build GRCh38
rs752531971 Chr.19: 42297267-42297267 on Build GRCh38
rs781585122 Chr.19: 42297158-42297158 on Build GRCh38
rs750411270 Chr.19: 42297056-42297056 on Build GRCh38
rs1054474234 Chr.19: 42297204-42297205 on Build GRCh38
rs778063234 Chr.19: 42297193-42297193 on Build GRCh38
rs770610163 Chr.19: 42297377-42297377 on Build GRCh38
rs758422229 Chr.19: 42297141-42297141 on Build GRCh38
rs139469632 Chr.19: 42297096-42297096 on Build GRCh38
rs756547283 Chr.19: 42297381-42297382 on Build GRCh38
rs977693644 Chr.19: 42297444-42297444 on Build GRCh38
rs374299820 Chr.19: 42297043-42297043 on Build GRCh38
rs773879190 Chr.19: 42297316-42297316 on Build GRCh38
rs371413124 Chr.19: 42297392-42297392 on Build GRCh38
rs772563976 Chr.19: 42297292-42297292 on Build GRCh38
rs914277058 Chr.19: 42296971-42296971 on Build GRCh38
rs11553635 Chr.19: 42297137-42297137 on Build GRCh38
rs746236712 Chr.19: 42296998-42296998 on Build GRCh38
rs773400010 Chr.19: 42297090-42297090 on Build GRCh38
rs1043818 Chr.19: 42297134-42297134 on Build GRCh38
rs117750674 Chr.19: 42297409-42297409 on Build GRCh38
rs767545895 Chr.19: 42297319-42297319 on Build GRCh38
rs375948081 Chr.19: 42297092-42297092 on Build GRCh38
rs770393032 Chr.19: 42297315-42297315 on Build GRCh38
rs923600427 Chr.19: 42297452-42297452 on Build GRCh38
rs753954983 Chr.19: 42297278-42297278 on Build GRCh38
rs199807899 Chr.19: 42297326-42297326 on Build GRCh38
rs763739992 Chr.19: 42297121-42297121 on Build GRCh38
rs759521771 Chr.19: 42297257-42297257 on Build GRCh38
rs557243754 Chr.19: 42297295-42297295 on Build GRCh38
rs768857831 Chr.19: 42297354-42297354 on Build GRCh38
rs924365327 Chr.19: 42297372-42297372 on Build GRCh38
rs779029713 Chr.19: 42297284-42297284 on Build GRCh38
rs753957344 Chr.19: 42297328-42297328 on Build GRCh38
rs896303499 Chr.19: 42297261-42297261 on Build GRCh38
rs779742816 Chr.19: 42297036-42297036 on Build GRCh38
rs369703171 Chr.19: 42297415-42297415 on Build GRCh38
rs539916888 Chr.19: 42297434-42297434 on Build GRCh38
rs369223278 Chr.19: 42297265-42297265 on Build GRCh38
rs770016763 Chr.19: 42297197-42297197 on Build GRCh38
rs763721979 Chr.19: 42297037-42297037 on Build GRCh38
rs774652545 Chr.19: 42297211-42297211 on Build GRCh38

View Full Product Details