Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51880161-51880760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF577
Entrez Gene ID
84765
Gene Name
zinc finger protein 577
Gene Aliases
-
UniGene
Hs.723015
Gene Chromosome Location
Chr.19: 51855803-51887976 on Build GRCh38
Transcripts
NM_001135590.1 NM_032679.2 NR_024181.1 XM_006723432.3 XM_006723433.2 XM_011527408.2 XM_017027387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1612679 c.24G>A p.M8I Chr.19: 51880359-51880359 on Build GRCh38
4000856 c.60+7T>C p.? Chr.19: 51880316-51880316 on Build GRCh38
3389286 c.6A>C p.K2N Chr.19: 51880377-51880377 on Build GRCh38
7218449 c.60+7T>G p.? Chr.19: 51880316-51880316 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1040372710 Chr.19: 51880602-51880602 on Build GRCh38
rs763136435 Chr.19: 51880385-51880385 on Build GRCh38
rs924904620 Chr.19: 51880207-51880207 on Build GRCh38
rs753747730 Chr.19: 51880353-51880353 on Build GRCh38
rs115833063 Chr.19: 51880438-51880438 on Build GRCh38
rs774365172 Chr.19: 51880424-51880424 on Build GRCh38
rs73576171 Chr.19: 51880190-51880190 on Build GRCh38
rs112415723 Chr.19: 51880361-51880361 on Build GRCh38
rs147711365 Chr.19: 51880530-51880530 on Build GRCh38
rs374050006 Chr.19: 51880312-51880312 on Build GRCh38
rs568212318 Chr.19: 51880728-51880728 on Build GRCh38
rs750074838 Chr.19: 51880614-51880614 on Build GRCh38
rs992552811 Chr.19: 51880662-51880662 on Build GRCh38
rs755492831 Chr.19: 51880660-51880660 on Build GRCh38
rs202085303 Chr.19: 51880363-51880363 on Build GRCh38
rs944755745 Chr.19: 51880577-51880577 on Build GRCh38
rs8110461 Chr.19: 51880316-51880316 on Build GRCh38
rs200598804 Chr.19: 51880372-51880372 on Build GRCh38
rs8110531 Chr.19: 51880257-51880257 on Build GRCh38
rs978834273 Chr.19: 51880724-51880724 on Build GRCh38
rs752895966 Chr.19: 51880340-51880340 on Build GRCh38
rs751232642 Chr.19: 51880348-51880348 on Build GRCh38
rs760759642 Chr.19: 51880341-51880341 on Build GRCh38
rs544223035 Chr.19: 51880640-51880640 on Build GRCh38
rs926211217 Chr.19: 51880496-51880496 on Build GRCh38
rs771391119 Chr.19: 51880319-51880319 on Build GRCh38
rs544445052 Chr.19: 51880441-51880441 on Build GRCh38
rs547828946 Chr.19: 51880211-51880211 on Build GRCh38
rs117092369 Chr.19: 51880697-51880697 on Build GRCh38
rs954514930 Chr.19: 51880677-51880677 on Build GRCh38
rs773538969 Chr.19: 51880345-51880345 on Build GRCh38
rs8113768 Chr.19: 51880685-51880685 on Build GRCh38
rs778724188 Chr.19: 51880280-51880280 on Build GRCh38
rs28658468 Chr.19: 51880414-51880414 on Build GRCh38
rs748706773 Chr.19: 51880694-51880694 on Build GRCh38
rs566479016 Chr.19: 51880583-51880583 on Build GRCh38
rs573124584 Chr.19: 51880404-51880404 on Build GRCh38
rs757024072 Chr.19: 51880277-51880277 on Build GRCh38
rs145580196 Chr.19: 51880330-51880330 on Build GRCh38
rs888368515 Chr.19: 51880622-51880622 on Build GRCh38
rs562741545 Chr.19: 51880473-51880473 on Build GRCh38
rs533896902 Chr.19: 51880258-51880258 on Build GRCh38
rs530331155 Chr.19: 51880476-51880476 on Build GRCh38
rs891885561 Chr.19: 51880576-51880576 on Build GRCh38
rs1030565544 Chr.19: 51880664-51880664 on Build GRCh38
rs772464374 Chr.19: 51880524-51880525 on Build GRCh38
rs8111551 Chr.19: 51880396-51880396 on Build GRCh38
rs8110546 Chr.19: 51880303-51880303 on Build GRCh38
rs993729054 Chr.19: 51880401-51880401 on Build GRCh38
rs919957896 Chr.19: 51880442-51880442 on Build GRCh38
rs924821458 Chr.19: 51880475-51880475 on Build GRCh38
rs531652737 Chr.19: 51880432-51880432 on Build GRCh38
rs542140196 Chr.19: 51880366-51880366 on Build GRCh38
rs772623028 Chr.19: 51880382-51880383 on Build GRCh38
rs376402835 Chr.19: 51880336-51880336 on Build GRCh38
rs750177133 Chr.19: 51880368-51880368 on Build GRCh38
rs8113685 Chr.19: 51880637-51880637 on Build GRCh38
rs8113584 Chr.19: 51880621-51880621 on Build GRCh38
rs761121497 Chr.19: 51880403-51880404 on Build GRCh38
rs917454603 Chr.19: 51880522-51880522 on Build GRCh38
rs193095967 Chr.19: 51880318-51880318 on Build GRCh38
rs1007363969 Chr.19: 51880678-51880678 on Build GRCh38
rs753796526 Chr.19: 51880275-51880275 on Build GRCh38
rs746460016 Chr.19: 51880305-51880305 on Build GRCh38
rs17835286 Chr.19: 51880606-51880606 on Build GRCh38
rs774732209 Chr.19: 51880427-51880427 on Build GRCh38
rs766807052 Chr.19: 51880406-51880407 on Build GRCh38
rs779713194 Chr.19: 51880380-51880380 on Build GRCh38
rs774921008 Chr.19: 51880329-51880329 on Build GRCh38
rs752417468 Chr.19: 51880407-51880407 on Build GRCh38
rs1057479696 Chr.19: 51880509-51880509 on Build GRCh38
rs916879722 Chr.19: 51880474-51880474 on Build GRCh38
rs773529529 Chr.19: 51880394-51880395 on Build GRCh38
rs961118310 Chr.19: 51880659-51880659 on Build GRCh38
rs367722442 Chr.19: 51880301-51880301 on Build GRCh38
rs113714800 Chr.19: 51880282-51880282 on Build GRCh38
rs746652257 Chr.19: 51880611-51880611 on Build GRCh38
rs745506169 Chr.19: 51880299-51880299 on Build GRCh38
rs770101696 Chr.19: 51880324-51880324 on Build GRCh38
rs375688340 Chr.19: 51880254-51880254 on Build GRCh38
rs754511775 Chr.19: 51880399-51880399 on Build GRCh38

View Full Product Details