Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98985954-98986514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL15A1
Entrez Gene ID
1306
Gene Name
collagen type XV alpha 1 chain
Gene Aliases
-
UniGene
Hs.409034
Gene Chromosome Location
Chr.9: 98943392-99070792 on Build GRCh38
Transcripts
NM_001855.4 XM_011518214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7192119 c.585T>G p.A195A Chr.9: 98986049-98986049 on Build GRCh38
4519226 c.558_559CT>TC p.F187L Chr.9: 98986022-98986023 on Build GRCh38
6258128 c.576G>T p.Q192H Chr.9: 98986040-98986040 on Build GRCh38
6394464 c.646A>T p.T216S Chr.9: 98986110-98986110 on Build GRCh38
4909966 c.558C>A p.P186P Chr.9: 98986022-98986022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775618011 Chr.9: 98986016-98986016 on Build GRCh38
rs533629578 Chr.9: 98986061-98986061 on Build GRCh38
rs138366672 Chr.9: 98986015-98986015 on Build GRCh38
rs752484757 Chr.9: 98986067-98986067 on Build GRCh38
rs780107118 Chr.9: 98986192-98986192 on Build GRCh38
rs541160594 Chr.9: 98986433-98986433 on Build GRCh38
rs755963031 Chr.9: 98986070-98986070 on Build GRCh38
rs564912343 Chr.9: 98986387-98986387 on Build GRCh38
rs144123375 Chr.9: 98986101-98986101 on Build GRCh38
rs367577178 Chr.9: 98986121-98986121 on Build GRCh38
rs969598038 Chr.9: 98986365-98986365 on Build GRCh38
rs781377732 Chr.9: 98986132-98986132 on Build GRCh38
rs765891236 Chr.9: 98986060-98986060 on Build GRCh38
rs923521913 Chr.9: 98986379-98986379 on Build GRCh38
rs763468006 Chr.9: 98986163-98986163 on Build GRCh38
rs2075663 Chr.9: 98986074-98986074 on Build GRCh38
rs750622764 Chr.9: 98986002-98986004 on Build GRCh38
rs746005741 Chr.9: 98986014-98986014 on Build GRCh38
rs1036057893 Chr.9: 98986474-98986474 on Build GRCh38
rs747201542 Chr.9: 98986017-98986017 on Build GRCh38
rs953622536 Chr.9: 98986383-98986383 on Build GRCh38
rs956703056 Chr.9: 98986350-98986350 on Build GRCh38
rs34485462 Chr.9: 98986102-98986103 on Build GRCh38
rs770381847 Chr.9: 98986479-98986479 on Build GRCh38
rs75461182 Chr.9: 98986007-98986007 on Build GRCh38
rs751192017 Chr.9: 98985992-98985992 on Build GRCh38
rs773702295 Chr.9: 98986052-98986052 on Build GRCh38
rs908965098 Chr.9: 98986198-98986198 on Build GRCh38
rs576422294 Chr.9: 98986236-98986236 on Build GRCh38
rs976056036 Chr.9: 98986373-98986373 on Build GRCh38
rs372275873 Chr.9: 98986135-98986135 on Build GRCh38
rs558739936 Chr.9: 98986031-98986031 on Build GRCh38
rs770341356 Chr.9: 98986046-98986046 on Build GRCh38
rs762408701 Chr.9: 98986054-98986054 on Build GRCh38
rs768805861 Chr.9: 98986120-98986120 on Build GRCh38
rs781362380 Chr.9: 98986301-98986301 on Build GRCh38
rs541439391 Chr.9: 98986229-98986229 on Build GRCh38
rs751875256 Chr.9: 98986196-98986196 on Build GRCh38
rs116393661 Chr.9: 98986264-98986264 on Build GRCh38
rs753527788 Chr.9: 98986100-98986100 on Build GRCh38
rs187697004 Chr.9: 98986030-98986030 on Build GRCh38
rs34312437 Chr.9: 98986194-98986194 on Build GRCh38
rs772923849 Chr.9: 98986089-98986090 on Build GRCh38
rs941799156 Chr.9: 98986469-98986469 on Build GRCh38
rs941358806 Chr.9: 98986461-98986461 on Build GRCh38
rs1002774943 Chr.9: 98986249-98986249 on Build GRCh38
rs754779649 Chr.9: 98985993-98985993 on Build GRCh38
rs756050932 Chr.9: 98986009-98986009 on Build GRCh38
rs894356406 Chr.9: 98986281-98986281 on Build GRCh38
rs113791685 Chr.9: 98986035-98986035 on Build GRCh38
rs956964338 Chr.9: 98986123-98986123 on Build GRCh38
rs758460554 Chr.9: 98986080-98986080 on Build GRCh38
rs1052021357 Chr.9: 98986168-98986168 on Build GRCh38
rs184236472 Chr.9: 98986414-98986414 on Build GRCh38
rs1000937173 Chr.9: 98986330-98986330 on Build GRCh38
rs948226567 Chr.9: 98986034-98986034 on Build GRCh38
rs61999303 Chr.9: 98986062-98986062 on Build GRCh38
rs748577355 Chr.9: 98986147-98986147 on Build GRCh38
rs1013171660 Chr.9: 98986285-98986285 on Build GRCh38
rs1037623118 Chr.9: 98986160-98986160 on Build GRCh38
rs1010489487 Chr.9: 98986289-98986289 on Build GRCh38
rs1023861543 Chr.9: 98986339-98986342 on Build GRCh38
rs968576874 Chr.9: 98986329-98986329 on Build GRCh38
rs748128818 Chr.9: 98986008-98986008 on Build GRCh38
rs1031035320 Chr.9: 98986333-98986333 on Build GRCh38
rs941783499 Chr.9: 98986214-98986214 on Build GRCh38
rs781086307 Chr.9: 98986000-98986000 on Build GRCh38
rs556457015 Chr.9: 98986230-98986230 on Build GRCh38
rs968124809 Chr.9: 98986084-98986084 on Build GRCh38
rs192623412 Chr.9: 98986164-98986164 on Build GRCh38
rs923487492 Chr.9: 98986170-98986170 on Build GRCh38
rs574015279 Chr.9: 98986419-98986419 on Build GRCh38
rs1021450864 Chr.9: 98986311-98986311 on Build GRCh38
rs756150683 Chr.9: 98986076-98986076 on Build GRCh38
rs753786778 Chr.9: 98986077-98986077 on Build GRCh38
rs12238644 Chr.9: 98986275-98986275 on Build GRCh38
rs999682253 Chr.9: 98986206-98986206 on Build GRCh38
rs779150801 Chr.9: 98986010-98986010 on Build GRCh38
rs918978912 Chr.9: 98986489-98986489 on Build GRCh38
rs773757306 Chr.9: 98986158-98986158 on Build GRCh38

View Full Product Details