Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113407095-113407503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLE3
Entrez Gene ID
54107
Gene Name
DNA polymerase epsilon 3, accessory subunit
Gene Aliases
CHARAC17, CHRAC17, YBL1, p17
UniGene
Hs.108112
Gene Chromosome Location
Chr.9: 113407235-113410749 on Build GRCh38
Transcripts
NM_001278255.1 NM_017443.4 NR_027261.1

SNP Detail

SNP ID Chromosome Location
rs796296606 Chr.9: 113407422-113407422 on Build GRCh38
rs1000857194 Chr.9: 113407345-113407345 on Build GRCh38
rs937859925 Chr.9: 113407266-113407266 on Build GRCh38
rs896578527 Chr.9: 113407298-113407298 on Build GRCh38
rs982422331 Chr.9: 113407157-113407157 on Build GRCh38
rs955088084 Chr.9: 113407153-113407153 on Build GRCh38
rs2792813 Chr.9: 113407435-113407435 on Build GRCh38
rs548242264 Chr.9: 113407396-113407396 on Build GRCh38
rs941035633 Chr.9: 113407191-113407191 on Build GRCh38
rs537858771 Chr.9: 113407385-113407385 on Build GRCh38
rs945477102 Chr.9: 113407319-113407319 on Build GRCh38
rs757049008 Chr.9: 113407347-113407347 on Build GRCh38
rs959284127 Chr.9: 113407462-113407462 on Build GRCh38
rs965273638 Chr.9: 113407143-113407143 on Build GRCh38
rs903875151 Chr.9: 113407326-113407326 on Build GRCh38
rs992068246 Chr.9: 113407470-113407470 on Build GRCh38
rs1003958612 Chr.9: 113407386-113407386 on Build GRCh38
rs908250568 Chr.9: 113407184-113407184 on Build GRCh38
rs973739656 Chr.9: 113407442-113407442 on Build GRCh38
rs369661173 Chr.9: 113407325-113407325 on Build GRCh38
rs557780183 Chr.9: 113407340-113407340 on Build GRCh38
rs539466788 Chr.9: 113407331-113407331 on Build GRCh38
rs764330323 Chr.9: 113407152-113407152 on Build GRCh38
rs926547858 Chr.9: 113407210-113407210 on Build GRCh38
rs34006719 Chr.9: 113407421-113407421 on Build GRCh38
rs1017834659 Chr.9: 113407121-113407121 on Build GRCh38
rs1030425478 Chr.9: 113407370-113407370 on Build GRCh38
rs866206083 Chr.9: 113407304-113407304 on Build GRCh38
rs997643615 Chr.9: 113407359-113407359 on Build GRCh38
rs36080340 Chr.9: 113407453-113407453 on Build GRCh38
rs1015279165 Chr.9: 113407416-113407416 on Build GRCh38
rs760934789 Chr.9: 113407197-113407198 on Build GRCh38
rs147331290 Chr.9: 113407398-113407398 on Build GRCh38
rs751362026 Chr.9: 113407274-113407274 on Build GRCh38
rs568953557 Chr.9: 113407194-113407194 on Build GRCh38

View Full Product Details