Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132165971-132166508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTNG2
Entrez Gene ID
84628
Gene Name
netrin G2
Gene Aliases
LHLL9381, Lmnt2, NTNG1, bA479K20.1
UniGene
Hs.163642
Gene Chromosome Location
Chr.9: 132161234-132244534 on Build GRCh38
Transcripts
NM_032536.2 XM_006717304.3 XM_011519094.2 XM_011519096.2 XM_011519097.2 XM_011519098.2 XM_011519099.2 XM_011519100.2 XM_011519102.2 XM_011519103.2 XM_011519104.2 XM_011519105.2 XM_011519106.2 XM_011519107.2 XM_011519108.2 XM_011519109.2 XM_011519110.2 XM_017015212.1 XM_017015213.1 XM_017015214.1 XM_017015216.1

SNP Detail

SNP ID Chromosome Location
rs767135849 Chr.9: 132166165-132166167 on Build GRCh38
rs538522470 Chr.9: 132166165-132166165 on Build GRCh38
rs774518120 Chr.9: 132166031-132166031 on Build GRCh38
rs543116369 Chr.9: 132166368-132166368 on Build GRCh38
rs895941290 Chr.9: 132166355-132166355 on Build GRCh38
rs141066789 Chr.9: 132166171-132166171 on Build GRCh38
rs1037185187 Chr.9: 132166140-132166140 on Build GRCh38
rs575312190 Chr.9: 132166294-132166294 on Build GRCh38
rs565258510 Chr.9: 132166471-132166471 on Build GRCh38
rs200913269 Chr.9: 132166008-132166009 on Build GRCh38
rs752954662 Chr.9: 132166472-132166472 on Build GRCh38
rs560163156 Chr.9: 132166125-132166125 on Build GRCh38
rs751327929 Chr.9: 132165996-132165997 on Build GRCh38
rs908445479 Chr.9: 132166206-132166206 on Build GRCh38
rs768847860 Chr.9: 132165995-132165996 on Build GRCh38
rs760002393 Chr.9: 132166321-132166321 on Build GRCh38
rs77874701 Chr.9: 132166409-132166409 on Build GRCh38
rs939955018 Chr.9: 132166232-132166232 on Build GRCh38
rs1014437415 Chr.9: 132166368-132166372 on Build GRCh38
rs1053298817 Chr.9: 132166389-132166389 on Build GRCh38
rs1041078100 Chr.9: 132166233-132166233 on Build GRCh38
rs1049483949 Chr.9: 132166328-132166328 on Build GRCh38
rs776457277 Chr.9: 132166454-132166454 on Build GRCh38
rs1024322513 Chr.9: 132165994-132165994 on Build GRCh38
rs1014516988 Chr.9: 132166412-132166412 on Build GRCh38
rs776868360 Chr.9: 132166279-132166279 on Build GRCh38
rs540667354 Chr.9: 132166432-132166432 on Build GRCh38
rs939797192 Chr.9: 132166137-132166137 on Build GRCh38
rs920716214 Chr.9: 132166120-132166120 on Build GRCh38
rs532341162 Chr.9: 132166020-132166020 on Build GRCh38
rs554812615 Chr.9: 132166381-132166381 on Build GRCh38
rs151089458 Chr.9: 132166085-132166085 on Build GRCh38
rs766301826 Chr.9: 132166382-132166382 on Build GRCh38
rs552539292 Chr.9: 132166062-132166062 on Build GRCh38
rs965553614 Chr.9: 132166468-132166468 on Build GRCh38
rs761991881 Chr.9: 132166212-132166212 on Build GRCh38
rs779462641 Chr.9: 132165997-132165998 on Build GRCh38

View Full Product Details