Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 37846795-37847371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGM2
Entrez Gene ID
55276
Gene Name
phosphoglucomutase 2
Gene Aliases
MSTP006
UniGene
Hs.23363
Gene Chromosome Location
Chr.4: 37826660-37862938 on Build GRCh38
Transcripts
NM_018290.3 XM_011513711.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3825794 c.1217G>A p.G406E Chr.4: 37847230-37847230 on Build GRCh38
1310031 c.1180C>T p.H394Y Chr.4: 37847103-37847103 on Build GRCh38
5726426 c.1242C>T p.D414D Chr.4: 37847255-37847255 on Build GRCh38
733545 c.1019G>A p.R340K Chr.4: 37846942-37846942 on Build GRCh38
6197959 c.1008-7C>T p.? Chr.4: 37846924-37846924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757622084 Chr.4: 37846912-37846913 on Build GRCh38
rs1008921547 Chr.4: 37846994-37846994 on Build GRCh38
rs773567034 Chr.4: 37847222-37847222 on Build GRCh38
rs772018544 Chr.4: 37846906-37846906 on Build GRCh38
rs541207553 Chr.4: 37847330-37847330 on Build GRCh38
rs970740476 Chr.4: 37847073-37847073 on Build GRCh38
rs768378896 Chr.4: 37847085-37847085 on Build GRCh38
rs373785173 Chr.4: 37847057-37847057 on Build GRCh38
rs981536661 Chr.4: 37847136-37847136 on Build GRCh38
rs865910041 Chr.4: 37847133-37847133 on Build GRCh38
rs770928939 Chr.4: 37846995-37846995 on Build GRCh38
rs768837181 Chr.4: 37847023-37847023 on Build GRCh38
rs775973803 Chr.4: 37846935-37846935 on Build GRCh38
rs776207778 Chr.4: 37846915-37846915 on Build GRCh38
rs146573335 Chr.4: 37847039-37847039 on Build GRCh38
rs1038160707 Chr.4: 37847007-37847007 on Build GRCh38
rs765465119 Chr.4: 37846855-37846855 on Build GRCh38
rs914382793 Chr.4: 37847241-37847241 on Build GRCh38
rs112186779 Chr.4: 37846913-37846914 on Build GRCh38
rs750266558 Chr.4: 37847309-37847309 on Build GRCh38
rs142630273 Chr.4: 37847025-37847025 on Build GRCh38
rs765543372 Chr.4: 37847170-37847170 on Build GRCh38
rs762388948 Chr.4: 37846944-37846944 on Build GRCh38
rs566155120 Chr.4: 37846872-37846872 on Build GRCh38
rs201260848 Chr.4: 37847201-37847201 on Build GRCh38
rs779663411 Chr.4: 37847188-37847188 on Build GRCh38
rs377534332 Chr.4: 37847104-37847104 on Build GRCh38
rs778634036 Chr.4: 37846894-37846894 on Build GRCh38
rs748588862 Chr.4: 37847119-37847119 on Build GRCh38
rs142887287 Chr.4: 37846846-37846846 on Build GRCh38
rs769513810 Chr.4: 37847141-37847141 on Build GRCh38
rs751390154 Chr.4: 37847047-37847047 on Build GRCh38
rs528886092 Chr.4: 37846881-37846881 on Build GRCh38
rs950779110 Chr.4: 37847162-37847162 on Build GRCh38
rs368899552 Chr.4: 37846914-37846914 on Build GRCh38
rs147924614 Chr.4: 37847076-37847076 on Build GRCh38
rs187076049 Chr.4: 37846957-37846957 on Build GRCh38
rs959408762 Chr.4: 37846842-37846842 on Build GRCh38
rs373567431 Chr.4: 37847071-37847071 on Build GRCh38
rs370977104 Chr.4: 37847314-37847314 on Build GRCh38
rs774560479 Chr.4: 37847094-37847094 on Build GRCh38
rs750473953 Chr.4: 37847038-37847038 on Build GRCh38
rs1027022584 Chr.4: 37846840-37846840 on Build GRCh38
rs773016358 Chr.4: 37847241-37847248 on Build GRCh38
rs776548771 Chr.4: 37847256-37847256 on Build GRCh38
rs140033585 Chr.4: 37847214-37847214 on Build GRCh38
rs924549438 Chr.4: 37847102-37847102 on Build GRCh38
rs771447058 Chr.4: 37847237-37847237 on Build GRCh38
rs775838196 Chr.4: 37847291-37847291 on Build GRCh38
rs779716563 Chr.4: 37847080-37847080 on Build GRCh38
rs143879563 Chr.4: 37846940-37846940 on Build GRCh38
rs762179521 Chr.4: 37847028-37847028 on Build GRCh38
rs746512066 Chr.4: 37846912-37846912 on Build GRCh38
rs754558839 Chr.4: 37847197-37847197 on Build GRCh38
rs758181941 Chr.4: 37846978-37846978 on Build GRCh38
rs765371340 Chr.4: 37847271-37847271 on Build GRCh38
rs959460933 Chr.4: 37847049-37847049 on Build GRCh38
rs934504892 Chr.4: 37846913-37846913 on Build GRCh38
rs767548263 Chr.4: 37847173-37847173 on Build GRCh38
rs999533492 Chr.4: 37846861-37846861 on Build GRCh38
rs747471327 Chr.4: 37847230-37847230 on Build GRCh38
rs762070661 Chr.4: 37847137-37847137 on Build GRCh38
rs990312162 Chr.4: 37846883-37846883 on Build GRCh38
rs371051463 Chr.4: 37847158-37847158 on Build GRCh38
rs778696706 Chr.4: 37847200-37847200 on Build GRCh38
rs368033914 Chr.4: 37846962-37846962 on Build GRCh38
rs538140719 Chr.4: 37846942-37846942 on Build GRCh38
rs148604949 Chr.4: 37846986-37846986 on Build GRCh38
rs755229310 Chr.4: 37847043-37847043 on Build GRCh38
rs759699982 Chr.4: 37847266-37847266 on Build GRCh38
rs1032376735 Chr.4: 37847052-37847052 on Build GRCh38
rs151106378 Chr.4: 37847019-37847019 on Build GRCh38
rs199864815 Chr.4: 37847135-37847135 on Build GRCh38
rs1031897108 Chr.4: 37846863-37846863 on Build GRCh38
rs199503228 Chr.4: 37846924-37846924 on Build GRCh38
rs28662352 Chr.4: 37847328-37847328 on Build GRCh38
rs756152375 Chr.4: 37847180-37847180 on Build GRCh38
rs144871946 Chr.4: 37847035-37847035 on Build GRCh38
rs759625293 Chr.4: 37847164-37847164 on Build GRCh38
rs776773826 Chr.4: 37847105-37847105 on Build GRCh38
rs576708418 Chr.4: 37847141-37847141 on Build GRCh38
rs748853806 Chr.4: 37847082-37847082 on Build GRCh38
rs781317423 Chr.4: 37847058-37847058 on Build GRCh38
rs769539200 Chr.4: 37847008-37847008 on Build GRCh38
rs2290742 Chr.4: 37846834-37846834 on Build GRCh38
rs750319699 Chr.4: 37847174-37847174 on Build GRCh38
rs766375682 Chr.4: 37847152-37847152 on Build GRCh38
rs6814767 Chr.4: 37846909-37846909 on Build GRCh38
rs767457128 Chr.4: 37846945-37846945 on Build GRCh38
rs780798440 Chr.4: 37847003-37847003 on Build GRCh38
rs751953753 Chr.4: 37846943-37846943 on Build GRCh38
rs767380668 Chr.4: 37847304-37847304 on Build GRCh38
rs761180811 Chr.4: 37847147-37847147 on Build GRCh38
rs753998019 Chr.4: 37847041-37847041 on Build GRCh38
rs756309156 Chr.4: 37846961-37846961 on Build GRCh38
rs374140583 Chr.4: 37847172-37847172 on Build GRCh38
rs28544509 Chr.4: 37846914-37846914 on Build GRCh38
rs368412134 Chr.4: 37846982-37846982 on Build GRCh38
rs142978012 Chr.4: 37847010-37847010 on Build GRCh38
rs752488441 Chr.4: 37846969-37846969 on Build GRCh38
rs377562170 Chr.4: 37847310-37847310 on Build GRCh38
rs141684020 Chr.4: 37847292-37847292 on Build GRCh38
rs950026070 Chr.4: 37846874-37846874 on Build GRCh38
rs762895957 Chr.4: 37847262-37847262 on Build GRCh38
rs768850346 Chr.4: 37846924-37846924 on Build GRCh38
rs973902645 Chr.4: 37847261-37847261 on Build GRCh38
rs745417818 Chr.4: 37847005-37847005 on Build GRCh38
rs747988518 Chr.4: 37846897-37846897 on Build GRCh38
rs752392054 Chr.4: 37846907-37846909 on Build GRCh38

View Full Product Details